Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 158938319-158938831 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PYHIN1
Entrez Gene ID
149628
Gene Name
pyrin and HIN domain family member 1
Gene Aliases
IFIX
UniGene
Hs.710248
Gene Chromosome Location
Chr.1: 158931177-159010083 on Build GRCh38
Transcripts
NM_152501.4 NM_198928.4 NM_198929.4 NM_198930.3 XM_005244930.1 XM_011509242.2 XM_011509243.2 XM_017000463.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1987056 c.334G>T p.V112L Chr.1: 158938465-158938465 on Build GRCh38
338232 c.389C>G p.A130G Chr.1: 158938520-158938520 on Build GRCh38
4484766 c.284C>T p.S95F Chr.1: 158938415-158938415 on Build GRCh38
7138793 c.403G>C p.G135R Chr.1: 158938534-158938534 on Build GRCh38
4023889 c.410A>G p.Q137R Chr.1: 158938541-158938541 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs563195017 Chr.1: 158938396-158938396 on Build GRCh38
rs76450859 Chr.1: 158938767-158938767 on Build GRCh38
rs945291096 Chr.1: 158938375-158938375 on Build GRCh38
rs1006956724 Chr.1: 158938419-158938419 on Build GRCh38
rs746725907 Chr.1: 158938524-158938524 on Build GRCh38
rs570328049 Chr.1: 158938715-158938715 on Build GRCh38
rs373565343 Chr.1: 158938358-158938358 on Build GRCh38
rs989902884 Chr.1: 158938621-158938621 on Build GRCh38
rs112374116 Chr.1: 158938553-158938553 on Build GRCh38
rs201414654 Chr.1: 158938503-158938503 on Build GRCh38
rs898485966 Chr.1: 158938458-158938458 on Build GRCh38
rs750107406 Chr.1: 158938359-158938359 on Build GRCh38
rs761830753 Chr.1: 158938534-158938534 on Build GRCh38
rs368112979 Chr.1: 158938420-158938420 on Build GRCh38
rs766178626 Chr.1: 158938489-158938489 on Build GRCh38
rs776359395 Chr.1: 158938532-158938532 on Build GRCh38
rs776645629 Chr.1: 158938432-158938432 on Build GRCh38
rs772242282 Chr.1: 158938513-158938513 on Build GRCh38
rs756816142 Chr.1: 158938352-158938352 on Build GRCh38
rs759748194 Chr.1: 158938400-158938400 on Build GRCh38
rs781683802 Chr.1: 158938370-158938370 on Build GRCh38
rs1025945833 Chr.1: 158938510-158938510 on Build GRCh38
rs775242535 Chr.1: 158938472-158938472 on Build GRCh38
rs945732247 Chr.1: 158938714-158938714 on Build GRCh38
rs765894188 Chr.1: 158938574-158938574 on Build GRCh38
rs778371600 Chr.1: 158938385-158938385 on Build GRCh38
rs770166039 Chr.1: 158938374-158938374 on Build GRCh38
rs764838755 Chr.1: 158938442-158938442 on Build GRCh38
rs1016545316 Chr.1: 158938593-158938593 on Build GRCh38
rs1043656884 Chr.1: 158938387-158938387 on Build GRCh38
rs529485492 Chr.1: 158938730-158938730 on Build GRCh38
rs267598111 Chr.1: 158938415-158938415 on Build GRCh38
rs933212737 Chr.1: 158938716-158938717 on Build GRCh38
rs1030931791 Chr.1: 158938795-158938795 on Build GRCh38
rs1050243426 Chr.1: 158938764-158938764 on Build GRCh38
rs971604199 Chr.1: 158938506-158938506 on Build GRCh38
rs140897285 Chr.1: 158938423-158938423 on Build GRCh38
rs768503732 Chr.1: 158938525-158938525 on Build GRCh38
rs549083684 Chr.1: 158938769-158938769 on Build GRCh38
rs745995286 Chr.1: 158938511-158938511 on Build GRCh38
rs765318978 Chr.1: 158938543-158938543 on Build GRCh38
rs1038449372 Chr.1: 158938790-158938790 on Build GRCh38
rs769303303 Chr.1: 158938452-158938452 on Build GRCh38
rs954760724 Chr.1: 158938801-158938801 on Build GRCh38
rs745721384 Chr.1: 158938449-158938449 on Build GRCh38
rs201214617 Chr.1: 158938584-158938584 on Build GRCh38
rs775124794 Chr.1: 158938418-158938418 on Build GRCh38
rs745903018 Chr.1: 158938363-158938363 on Build GRCh38
rs764133904 Chr.1: 158938347-158938347 on Build GRCh38
rs199928433 Chr.1: 158938561-158938561 on Build GRCh38
rs138593148 Chr.1: 158938493-158938493 on Build GRCh38
rs752959693 Chr.1: 158938491-158938491 on Build GRCh38
rs376999934 Chr.1: 158938372-158938372 on Build GRCh38
rs981619976 Chr.1: 158938520-158938520 on Build GRCh38
rs375461756 Chr.1: 158938693-158938693 on Build GRCh38
rs371608217 Chr.1: 158938389-158938389 on Build GRCh38
rs761341948 Chr.1: 158938437-158938437 on Build GRCh38
rs758193865 Chr.1: 158938364-158938364 on Build GRCh38
rs757825470 Chr.1: 158938502-158938502 on Build GRCh38
rs775803496 Chr.1: 158938521-158938521 on Build GRCh38
rs199804938 Chr.1: 158938399-158938399 on Build GRCh38
rs531466227 Chr.1: 158938382-158938382 on Build GRCh38
rs563757453 Chr.1: 158938501-158938501 on Build GRCh38
rs753247820 Chr.1: 158938351-158938351 on Build GRCh38
rs914256751 Chr.1: 158938670-158938670 on Build GRCh38
rs748746522 Chr.1: 158938422-158938422 on Build GRCh38
rs151006388 Chr.1: 158938410-158938410 on Build GRCh38

View Full Product Details