Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 159307561-159308057 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCER1A
Entrez Gene ID
2205
Gene Name
Fc fragment of IgE receptor Ia
Gene Aliases
FCE1A, FcERI
UniGene
Hs.897
Gene Chromosome Location
Chr.1: 159283888-159308224 on Build GRCh38
Transcripts
NM_002001.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
94303 c.724G>A p.G242S Chr.1: 159307882-159307882 on Build GRCh38
1206923 c.758A>C p.N253T Chr.1: 159307916-159307916 on Build GRCh38
4023950 c.610C>T p.L204L Chr.1: 159307768-159307768 on Build GRCh38
4169453 c.655G>C p.D219H Chr.1: 159307813-159307813 on Build GRCh38
675975 c.660A>G p.T220T Chr.1: 159307818-159307818 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780188505 Chr.1: 159307746-159307746 on Build GRCh38
rs777728646 Chr.1: 159307933-159307933 on Build GRCh38
rs898522031 Chr.1: 159308015-159308015 on Build GRCh38
rs747241420 Chr.1: 159307814-159307814 on Build GRCh38
rs965531666 Chr.1: 159307589-159307589 on Build GRCh38
rs201635034 Chr.1: 159307883-159307883 on Build GRCh38
rs757267193 Chr.1: 159307731-159307731 on Build GRCh38
rs79965525 Chr.1: 159308014-159308014 on Build GRCh38
rs556615494 Chr.1: 159307969-159307969 on Build GRCh38
rs867893538 Chr.1: 159307869-159307869 on Build GRCh38
rs371852080 Chr.1: 159307850-159307850 on Build GRCh38
rs779231575 Chr.1: 159307891-159307891 on Build GRCh38
rs544883122 Chr.1: 159307958-159307958 on Build GRCh38
rs190576386 Chr.1: 159307994-159307994 on Build GRCh38
rs748787041 Chr.1: 159307863-159307863 on Build GRCh38
rs764816198 Chr.1: 159307921-159307921 on Build GRCh38
rs780281986 Chr.1: 159307812-159307812 on Build GRCh38
rs961351521 Chr.1: 159308006-159308006 on Build GRCh38
rs760662246 Chr.1: 159307943-159307943 on Build GRCh38
rs763898435 Chr.1: 159307708-159307709 on Build GRCh38
rs754255545 Chr.1: 159307790-159307790 on Build GRCh38
rs767054118 Chr.1: 159307693-159307693 on Build GRCh38
rs574517469 Chr.1: 159307605-159307609 on Build GRCh38
rs41264475 Chr.1: 159307899-159307899 on Build GRCh38
rs747269017 Chr.1: 159307923-159307923 on Build GRCh38
rs371500826 Chr.1: 159307834-159307834 on Build GRCh38
rs1028467946 Chr.1: 159307955-159307955 on Build GRCh38
rs756661811 Chr.1: 159307632-159307632 on Build GRCh38
rs143576640 Chr.1: 159307892-159307892 on Build GRCh38
rs766035632 Chr.1: 159307771-159307771 on Build GRCh38
rs773076918 Chr.1: 159307860-159307860 on Build GRCh38
rs200302228 Chr.1: 159307784-159307784 on Build GRCh38
rs553543242 Chr.1: 159307782-159307782 on Build GRCh38
rs754345322 Chr.1: 159307725-159307725 on Build GRCh38
rs150421183 Chr.1: 159307909-159307909 on Build GRCh38
rs996907435 Chr.1: 159307736-159307736 on Build GRCh38
rs781609732 Chr.1: 159307910-159307910 on Build GRCh38
rs751196521 Chr.1: 159307774-159307774 on Build GRCh38
rs761856898 Chr.1: 159307757-159307757 on Build GRCh38
rs758438640 Chr.1: 159307898-159307898 on Build GRCh38
rs750349173 Chr.1: 159307972-159307972 on Build GRCh38
rs370625938 Chr.1: 159307745-159307745 on Build GRCh38
rs778206897 Chr.1: 159307724-159307724 on Build GRCh38
rs200009003 Chr.1: 159307769-159307769 on Build GRCh38
rs769678039 Chr.1: 159307762-159307762 on Build GRCh38
rs763119646 Chr.1: 159307778-159307778 on Build GRCh38
rs773933049 Chr.1: 159307873-159307873 on Build GRCh38
rs181227379 Chr.1: 159307723-159307723 on Build GRCh38
rs535656043 Chr.1: 159307793-159307793 on Build GRCh38
rs77006202 Chr.1: 159307609-159307609 on Build GRCh38
rs759172009 Chr.1: 159307880-159307880 on Build GRCh38
rs531235531 Chr.1: 159307633-159307633 on Build GRCh38
rs146511084 Chr.1: 159307882-159307882 on Build GRCh38
rs754763948 Chr.1: 159307830-159307830 on Build GRCh38
rs17851282 Chr.1: 159307847-159307847 on Build GRCh38
rs896349320 Chr.1: 159307611-159307611 on Build GRCh38
rs560372743 Chr.1: 159308008-159308008 on Build GRCh38
rs779033762 Chr.1: 159307734-159307734 on Build GRCh38
rs375475641 Chr.1: 159307861-159307861 on Build GRCh38
rs753537203 Chr.1: 159307773-159307774 on Build GRCh38
rs373904317 Chr.1: 159307751-159307751 on Build GRCh38
rs750492996 Chr.1: 159307805-159307805 on Build GRCh38
rs749193181 Chr.1: 159307935-159307935 on Build GRCh38
rs868534170 Chr.1: 159308028-159308028 on Build GRCh38
rs757753621 Chr.1: 159307796-159307796 on Build GRCh38
rs188354096 Chr.1: 159307682-159307682 on Build GRCh38
rs761450888 Chr.1: 159307863-159307865 on Build GRCh38
rs770382474 Chr.1: 159307865-159307865 on Build GRCh38
rs904455516 Chr.1: 159307846-159307846 on Build GRCh38
rs185718654 Chr.1: 159307989-159307989 on Build GRCh38
rs755540332 Chr.1: 159307753-159307753 on Build GRCh38
rs148446616 Chr.1: 159307780-159307780 on Build GRCh38
rs367698700 Chr.1: 159307741-159307741 on Build GRCh38
rs142312060 Chr.1: 159307752-159307752 on Build GRCh38
rs770955683 Chr.1: 159307937-159307937 on Build GRCh38
rs771690270 Chr.1: 159307970-159307970 on Build GRCh38
rs887075647 Chr.1: 159307659-159307659 on Build GRCh38
rs769743599 Chr.1: 159307852-159307852 on Build GRCh38
rs375089556 Chr.1: 159307947-159307947 on Build GRCh38
rs758427988 Chr.1: 159307810-159307810 on Build GRCh38
rs751747623 Chr.1: 159307907-159307907 on Build GRCh38
rs143419433 Chr.1: 159307919-159307919 on Build GRCh38
rs755886643 Chr.1: 159307926-159307926 on Build GRCh38
rs772778349 Chr.1: 159307768-159307768 on Build GRCh38

View Full Product Details