Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 160039230-160039738 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNJ10
Entrez Gene ID
3766
Gene Name
potassium voltage-gated channel subfamily J member 10
Gene Aliases
BIRK-10, KCNJ13-PEN, KIR1.2, KIR4.1, SESAME
UniGene
Hs.408960
Gene Chromosome Location
Chr.1: 160037467-160070261 on Build GRCh38
Transcripts
NM_002241.4

SNP Detail

SNP ID Chromosome Location
rs961371438 Chr.1: 160039538-160039538 on Build GRCh38
rs932788482 Chr.1: 160039573-160039573 on Build GRCh38
rs1053074 Chr.1: 160039331-160039331 on Build GRCh38
rs1054377761 Chr.1: 160039642-160039642 on Build GRCh38
rs1036784286 Chr.1: 160039258-160039258 on Build GRCh38
rs886045402 Chr.1: 160039373-160039374 on Build GRCh38
rs868740933 Chr.1: 160039387-160039387 on Build GRCh38
rs138511291 Chr.1: 160039554-160039554 on Build GRCh38
rs188142576 Chr.1: 160039261-160039261 on Build GRCh38
rs781009174 Chr.1: 160039584-160039584 on Build GRCh38
rs932485495 Chr.1: 160039266-160039266 on Build GRCh38
rs996400298 Chr.1: 160039502-160039502 on Build GRCh38
rs908525660 Chr.1: 160039521-160039521 on Build GRCh38
rs756042800 Chr.1: 160039643-160039643 on Build GRCh38
rs1032109837 Chr.1: 160039427-160039427 on Build GRCh38
rs1054345320 Chr.1: 160039362-160039362 on Build GRCh38
rs891031616 Chr.1: 160039313-160039313 on Build GRCh38
rs1013504556 Chr.1: 160039484-160039484 on Build GRCh38
rs1040776576 Chr.1: 160039681-160039681 on Build GRCh38
rs552915175 Chr.1: 160039459-160039459 on Build GRCh38
rs912425295 Chr.1: 160039583-160039583 on Build GRCh38
rs777546653 Chr.1: 160039380-160039387 on Build GRCh38
rs763135560 Chr.1: 160039472-160039472 on Build GRCh38
rs540860788 Chr.1: 160039431-160039431 on Build GRCh38
rs1049626955 Chr.1: 160039302-160039302 on Build GRCh38
rs886045403 Chr.1: 160039374-160039381 on Build GRCh38
rs764202643 Chr.1: 160039478-160039478 on Build GRCh38
rs182896549 Chr.1: 160039491-160039491 on Build GRCh38
rs374594439 Chr.1: 160039374-160039377 on Build GRCh38
rs2486253 Chr.1: 160039629-160039629 on Build GRCh38
rs771719546 Chr.1: 160039374-160039387 on Build GRCh38
rs886045405 Chr.1: 160039538-160039538 on Build GRCh38
rs916950905 Chr.1: 160039324-160039324 on Build GRCh38
rs891822007 Chr.1: 160039406-160039406 on Build GRCh38
rs550315680 Chr.1: 160039605-160039605 on Build GRCh38
rs542638955 Chr.1: 160039604-160039604 on Build GRCh38
rs920012947 Chr.1: 160039555-160039555 on Build GRCh38
rs779982323 Chr.1: 160039662-160039662 on Build GRCh38
rs946104495 Chr.1: 160039655-160039655 on Build GRCh38
rs1000595958 Chr.1: 160039392-160039392 on Build GRCh38
rs201506426 Chr.1: 160039509-160039509 on Build GRCh38
rs886045404 Chr.1: 160039374-160039379 on Build GRCh38
rs979961092 Chr.1: 160039620-160039620 on Build GRCh38
rs1000100515 Chr.1: 160039362-160039365 on Build GRCh38
rs192835895 Chr.1: 160039359-160039359 on Build GRCh38
rs374034792 Chr.1: 160039625-160039625 on Build GRCh38
rs971614621 Chr.1: 160039654-160039654 on Build GRCh38
rs146422203 Chr.1: 160039374-160039383 on Build GRCh38
rs1010615689 Chr.1: 160039423-160039423 on Build GRCh38
rs78280261 Chr.1: 160039580-160039580 on Build GRCh38
rs886045406 Chr.1: 160039566-160039566 on Build GRCh38
rs530473210 Chr.1: 160039581-160039581 on Build GRCh38
rs972048863 Chr.1: 160039492-160039492 on Build GRCh38
rs569957581 Chr.1: 160039271-160039271 on Build GRCh38
rs56656397 Chr.1: 160039424-160039425 on Build GRCh38
rs68110005 Chr.1: 160039417-160039424 on Build GRCh38
rs2486254 Chr.1: 160039353-160039353 on Build GRCh38
rs981746645 Chr.1: 160039495-160039495 on Build GRCh38
rs768453943 Chr.1: 160039685-160039685 on Build GRCh38
rs894885843 Chr.1: 160039630-160039630 on Build GRCh38
rs748999196 Chr.1: 160039675-160039675 on Build GRCh38
rs1042348021 Chr.1: 160039597-160039597 on Build GRCh38
rs1023542913 Chr.1: 160039647-160039647 on Build GRCh38
rs72013077 Chr.1: 160039374-160039375 on Build GRCh38

View Full Product Details