Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 160041158-160041654 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNJ10
Entrez Gene ID
3766
Gene Name
potassium voltage-gated channel subfamily J member 10
Gene Aliases
BIRK-10, KCNJ13-PEN, KIR1.2, KIR4.1, SESAME
UniGene
Hs.408960
Gene Chromosome Location
Chr.1: 160037467-160070261 on Build GRCh38
Transcripts
NM_002241.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5881546 c.1028G>A p.R343H Chr.1: 160041505-160041505 on Build GRCh38
4444229 c.898T>C p.Y300H Chr.1: 160041635-160041635 on Build GRCh38
5892596 c.1085G>A p.R362K Chr.1: 160041448-160041448 on Build GRCh38
3376801 c.1032C>G p.D344E Chr.1: 160041501-160041501 on Build GRCh38
75083 c.984C>A p.S328R Chr.1: 160041549-160041549 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762083144 Chr.1: 160041322-160041322 on Build GRCh38
rs368793191 Chr.1: 160041603-160041603 on Build GRCh38
rs1052473157 Chr.1: 160041272-160041272 on Build GRCh38
rs115130978 Chr.1: 160041304-160041304 on Build GRCh38
rs770365142 Chr.1: 160041576-160041576 on Build GRCh38
rs749370465 Chr.1: 160041598-160041598 on Build GRCh38
rs867965809 Chr.1: 160041626-160041626 on Build GRCh38
rs796052605 Chr.1: 160041512-160041512 on Build GRCh38
rs747349113 Chr.1: 160041369-160041369 on Build GRCh38
rs942950199 Chr.1: 160041240-160041240 on Build GRCh38
rs148441646 Chr.1: 160041341-160041341 on Build GRCh38
rs888967595 Chr.1: 160041479-160041479 on Build GRCh38
rs926668743 Chr.1: 160041552-160041552 on Build GRCh38
rs748547971 Chr.1: 160041560-160041560 on Build GRCh38
rs375092803 Chr.1: 160041496-160041496 on Build GRCh38
rs778505490 Chr.1: 160041526-160041526 on Build GRCh38
rs767462808 Chr.1: 160041429-160041429 on Build GRCh38
rs555848512 Chr.1: 160041296-160041296 on Build GRCh38
rs557098814 Chr.1: 160041290-160041290 on Build GRCh38
rs986014209 Chr.1: 160041224-160041224 on Build GRCh38
rs756846428 Chr.1: 160041449-160041449 on Build GRCh38
rs760522990 Chr.1: 160041610-160041610 on Build GRCh38
rs142596580 Chr.1: 160041472-160041472 on Build GRCh38
rs114052043 Chr.1: 160041300-160041300 on Build GRCh38
rs886044169 Chr.1: 160041539-160041539 on Build GRCh38
rs200185135 Chr.1: 160041425-160041425 on Build GRCh38
rs777227509 Chr.1: 160041432-160041432 on Build GRCh38
rs764034536 Chr.1: 160041505-160041505 on Build GRCh38
rs748553015 Chr.1: 160041484-160041484 on Build GRCh38
rs554662066 Chr.1: 160041319-160041319 on Build GRCh38
rs961651783 Chr.1: 160041368-160041368 on Build GRCh38
rs184428585 Chr.1: 160041320-160041320 on Build GRCh38
rs894051645 Chr.1: 160041370-160041370 on Build GRCh38
rs368537434 Chr.1: 160041486-160041486 on Build GRCh38
rs1007318109 Chr.1: 160041480-160041480 on Build GRCh38
rs763224969 Chr.1: 160041405-160041405 on Build GRCh38
rs1016110817 Chr.1: 160041229-160041229 on Build GRCh38
rs760787513 Chr.1: 160041366-160041366 on Build GRCh38
rs377330985 Chr.1: 160041431-160041431 on Build GRCh38
rs777070823 Chr.1: 160041377-160041377 on Build GRCh38
rs751338154 Chr.1: 160041410-160041410 on Build GRCh38
rs374990648 Chr.1: 160041430-160041430 on Build GRCh38
rs754783764 Chr.1: 160041427-160041427 on Build GRCh38
rs781370143 Chr.1: 160041553-160041553 on Build GRCh38
rs775091787 Chr.1: 160041606-160041606 on Build GRCh38
rs771187154 Chr.1: 160041599-160041599 on Build GRCh38
rs61821988 Chr.1: 160041406-160041406 on Build GRCh38
rs893885507 Chr.1: 160041299-160041299 on Build GRCh38
rs748892244 Chr.1: 160041355-160041355 on Build GRCh38
rs370664864 Chr.1: 160041464-160041464 on Build GRCh38
rs770602948 Chr.1: 160041363-160041363 on Build GRCh38
rs768946463 Chr.1: 160041251-160041251 on Build GRCh38
rs756827951 Chr.1: 160041513-160041513 on Build GRCh38
rs1054054995 Chr.1: 160041344-160041344 on Build GRCh38
rs753814824 Chr.1: 160041629-160041629 on Build GRCh38
rs189596680 Chr.1: 160041409-160041409 on Build GRCh38
rs750091894 Chr.1: 160041535-160041535 on Build GRCh38
rs377168333 Chr.1: 160041524-160041524 on Build GRCh38
rs1034857467 Chr.1: 160041521-160041521 on Build GRCh38
rs901146903 Chr.1: 160041242-160041242 on Build GRCh38
rs571511216 Chr.1: 160041217-160041217 on Build GRCh38
rs371239938 Chr.1: 160041386-160041386 on Build GRCh38
rs146396982 Chr.1: 160041490-160041490 on Build GRCh38
rs868795052 Chr.1: 160041422-160041422 on Build GRCh38
rs372774976 Chr.1: 160041396-160041396 on Build GRCh38
rs377019389 Chr.1: 160041372-160041372 on Build GRCh38
rs761289460 Chr.1: 160041630-160041630 on Build GRCh38
rs1041714461 Chr.1: 160041241-160041241 on Build GRCh38
rs759356991 Chr.1: 160041497-160041497 on Build GRCh38
rs778478217 Chr.1: 160041585-160041585 on Build GRCh38
rs200737764 Chr.1: 160041605-160041605 on Build GRCh38
rs767170923 Chr.1: 160041498-160041498 on Build GRCh38
rs137853074 Chr.1: 160041491-160041491 on Build GRCh38
rs186165644 Chr.1: 160041501-160041501 on Build GRCh38
rs770453248 Chr.1: 160041485-160041485 on Build GRCh38
rs373899425 Chr.1: 160041482-160041482 on Build GRCh38
rs115725914 Chr.1: 160041256-160041256 on Build GRCh38
rs145588542 Chr.1: 160041441-160041441 on Build GRCh38
rs764014198 Chr.1: 160041615-160041615 on Build GRCh38
rs539110996 Chr.1: 160041222-160041222 on Build GRCh38
rs752571720 Chr.1: 160041500-160041500 on Build GRCh38
rs747395965 Chr.1: 160041468-160041468 on Build GRCh38
rs773446859 Chr.1: 160041399-160041399 on Build GRCh38
rs758090376 Chr.1: 160041550-160041550 on Build GRCh38
rs753333164 Chr.1: 160041506-160041506 on Build GRCh38
rs911410860 Chr.1: 160041235-160041235 on Build GRCh38

View Full Product Details