Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 160351616-160352117 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCSTN
Entrez Gene ID
23385
Gene Name
nicastrin
Gene Aliases
ATAG1874
UniGene
Hs.517249
Gene Chromosome Location
Chr.1: 160343056-160358952 on Build GRCh38
Transcripts
NM_001290184.1 NM_001290186.1 NM_015331.2 XM_005245053.4 XM_011509363.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7104265 c.759C>A p.D253E Chr.1: 160351781-160351781 on Build GRCh38
7104264 c.819C>A p.D273E Chr.1: 160351781-160351781 on Build GRCh38
4706035 c.794G>A p.R265H Chr.1: 160352064-160352064 on Build GRCh38
6494306 c.747C>T p.D249D Chr.1: 160351709-160351709 on Build GRCh38
2085683 c.739G>A p.V247I Chr.1: 160351701-160351701 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754637223 Chr.1: 160351647-160351647 on Build GRCh38
rs865874437 Chr.1: 160351794-160351794 on Build GRCh38
rs746388807 Chr.1: 160351832-160351832 on Build GRCh38
rs1023313115 Chr.1: 160351869-160351869 on Build GRCh38
rs981923091 Chr.1: 160351947-160351947 on Build GRCh38
rs908535441 Chr.1: 160351949-160351949 on Build GRCh38
rs868397928 Chr.1: 160351890-160351890 on Build GRCh38
rs762074777 Chr.1: 160351856-160351856 on Build GRCh38
rs889757743 Chr.1: 160351779-160351779 on Build GRCh38
rs757609494 Chr.1: 160351829-160351829 on Build GRCh38
rs746362895 Chr.1: 160351683-160351683 on Build GRCh38
rs200362821 Chr.1: 160351839-160351839 on Build GRCh38
rs867650316 Chr.1: 160352088-160352088 on Build GRCh38
rs773020426 Chr.1: 160352009-160352009 on Build GRCh38
rs931337815 Chr.1: 160351738-160351738 on Build GRCh38
rs772178774 Chr.1: 160351685-160351685 on Build GRCh38
rs148530123 Chr.1: 160351778-160351778 on Build GRCh38
rs201989886 Chr.1: 160351846-160351846 on Build GRCh38
rs762952922 Chr.1: 160351749-160351749 on Build GRCh38
rs1035174616 Chr.1: 160351982-160351982 on Build GRCh38
rs200203877 Chr.1: 160351853-160351853 on Build GRCh38
rs150605943 Chr.1: 160351691-160351691 on Build GRCh38
rs750378021 Chr.1: 160351740-160351740 on Build GRCh38
rs778304828 Chr.1: 160351658-160351658 on Build GRCh38
rs759185792 Chr.1: 160352049-160352049 on Build GRCh38
rs531877039 Chr.1: 160352089-160352089 on Build GRCh38
rs766018225 Chr.1: 160351761-160351761 on Build GRCh38
rs532533391 Chr.1: 160351837-160351837 on Build GRCh38
rs200575152 Chr.1: 160351675-160351675 on Build GRCh38
rs11547485 Chr.1: 160351712-160351712 on Build GRCh38
rs952839356 Chr.1: 160351929-160351929 on Build GRCh38
rs201298374 Chr.1: 160351699-160351699 on Build GRCh38
rs957958414 Chr.1: 160351865-160351865 on Build GRCh38
rs201725255 Chr.1: 160352004-160352004 on Build GRCh38
rs755748959 Chr.1: 160352068-160352068 on Build GRCh38
rs767029348 Chr.1: 160352063-160352063 on Build GRCh38
rs776577215 Chr.1: 160351714-160351714 on Build GRCh38
rs778081884 Chr.1: 160352020-160352020 on Build GRCh38
rs973293410 Chr.1: 160352062-160352062 on Build GRCh38
rs201597243 Chr.1: 160351655-160351655 on Build GRCh38
rs919982186 Chr.1: 160351909-160351909 on Build GRCh38
rs969420360 Chr.1: 160351876-160351876 on Build GRCh38
rs867036355 Chr.1: 160351700-160351700 on Build GRCh38
rs982233280 Chr.1: 160351677-160351677 on Build GRCh38
rs748015366 Chr.1: 160351832-160351840 on Build GRCh38
rs773294945 Chr.1: 160351944-160351944 on Build GRCh38
rs371680708 Chr.1: 160351844-160351844 on Build GRCh38
rs376416053 Chr.1: 160352002-160352002 on Build GRCh38
rs368085541 Chr.1: 160351662-160351662 on Build GRCh38
rs752329948 Chr.1: 160352064-160352064 on Build GRCh38
rs750728892 Chr.1: 160352093-160352093 on Build GRCh38
rs373915943 Chr.1: 160351687-160351687 on Build GRCh38
rs931295906 Chr.1: 160351666-160351666 on Build GRCh38
rs768805561 Chr.1: 160351694-160351694 on Build GRCh38
rs202069681 Chr.1: 160351701-160351701 on Build GRCh38
rs200043700 Chr.1: 160351665-160351665 on Build GRCh38
rs6664627 Chr.1: 160351709-160351709 on Build GRCh38
rs368412577 Chr.1: 160351843-160351843 on Build GRCh38
rs371663092 Chr.1: 160351983-160351983 on Build GRCh38
rs577452809 Chr.1: 160352046-160352046 on Build GRCh38
rs780645547 Chr.1: 160351804-160351804 on Build GRCh38
rs754694506 Chr.1: 160351790-160351790 on Build GRCh38
rs202165621 Chr.1: 160351799-160351799 on Build GRCh38
rs1041066 Chr.1: 160351866-160351866 on Build GRCh38
rs763011396 Chr.1: 160352013-160352013 on Build GRCh38
rs975910747 Chr.1: 160351907-160351907 on Build GRCh38
rs201353599 Chr.1: 160351682-160351682 on Build GRCh38
rs372365642 Chr.1: 160351735-160351735 on Build GRCh38
rs201381789 Chr.1: 160351845-160351845 on Build GRCh38
rs751121713 Chr.1: 160352041-160352041 on Build GRCh38
rs758055411 Chr.1: 160351906-160351906 on Build GRCh38
rs766364792 Chr.1: 160352026-160352026 on Build GRCh38
rs752392004 Chr.1: 160351812-160351812 on Build GRCh38

View Full Product Details