Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100884234-100884742 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SRRT
Entrez Gene ID
51593
Gene Name
serrate, RNA effector molecule
Gene Aliases
ARS2, ASR2, serrate
UniGene
Hs.111801
Gene Chromosome Location
Chr.7: 100874971-100888664 on Build GRCh38
Transcripts
NM_001128852.1 NM_001128853.1 NM_001128854.1 NM_015908.5 XM_005250405.2 XM_005250406.2 XM_005250407.2 XM_005250408.2 XM_017012290.1 XM_017012291.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6219521 c.799A>G p.I267V Chr.7: 100884409-100884409 on Build GRCh38
5959455 c.808_809insAGG p.E275_Q276insE Chr.7: 100884418-100884419 on Build GRCh38
3374420 c.855G>C p.K285N Chr.7: 100884465-100884465 on Build GRCh38
6228323 c.759C>T p.A253A Chr.7: 100884369-100884369 on Build GRCh38
5455528 c.816G>C p.E272D Chr.7: 100884426-100884426 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779052415 Chr.7: 100884402-100884402 on Build GRCh38
rs749020577 Chr.7: 100884467-100884469 on Build GRCh38
rs749835079 Chr.7: 100884393-100884393 on Build GRCh38
rs756753215 Chr.7: 100884278-100884278 on Build GRCh38
rs747988425 Chr.7: 100884702-100884702 on Build GRCh38
rs140745082 Chr.7: 100884509-100884509 on Build GRCh38
rs772508733 Chr.7: 100884346-100884346 on Build GRCh38
rs771695055 Chr.7: 100884289-100884289 on Build GRCh38
rs757573671 Chr.7: 100884404-100884404 on Build GRCh38
rs77638633 Chr.7: 100884313-100884313 on Build GRCh38
rs574539657 Chr.7: 100884583-100884583 on Build GRCh38
rs759248839 Chr.7: 100884717-100884717 on Build GRCh38
rs975635781 Chr.7: 100884701-100884701 on Build GRCh38
rs758076420 Chr.7: 100884355-100884355 on Build GRCh38
rs764546628 Chr.7: 100884561-100884561 on Build GRCh38
rs755952551 Chr.7: 100884418-100884419 on Build GRCh38
rs769829227 Chr.7: 100884539-100884539 on Build GRCh38
rs372458329 Chr.7: 100884638-100884638 on Build GRCh38
rs770407592 Chr.7: 100884715-100884715 on Build GRCh38
rs368044223 Chr.7: 100884398-100884398 on Build GRCh38
rs780280009 Chr.7: 100884515-100884515 on Build GRCh38
rs371531627 Chr.7: 100884461-100884461 on Build GRCh38
rs143459147 Chr.7: 100884369-100884369 on Build GRCh38
rs780973393 Chr.7: 100884527-100884527 on Build GRCh38
rs765811537 Chr.7: 100884566-100884566 on Build GRCh38
rs753164592 Chr.7: 100884365-100884365 on Build GRCh38
rs777726773 Chr.7: 100884705-100884705 on Build GRCh38
rs770380395 Chr.7: 100884556-100884556 on Build GRCh38
rs778464271 Chr.7: 100884392-100884392 on Build GRCh38
rs201456567 Chr.7: 100884274-100884274 on Build GRCh38
rs1007801397 Chr.7: 100884708-100884708 on Build GRCh38
rs776506765 Chr.7: 100884456-100884456 on Build GRCh38
rs777986982 Chr.7: 100884281-100884281 on Build GRCh38
rs775860815 Chr.7: 100884318-100884318 on Build GRCh38
rs775122762 Chr.7: 100884407-100884407 on Build GRCh38
rs777681259 Chr.7: 100884394-100884398 on Build GRCh38
rs896313092 Chr.7: 100884520-100884520 on Build GRCh38
rs1032491540 Chr.7: 100884311-100884311 on Build GRCh38
rs780955235 Chr.7: 100884596-100884596 on Build GRCh38
rs765301567 Chr.7: 100884264-100884264 on Build GRCh38
rs774069942 Chr.7: 100884716-100884716 on Build GRCh38
rs761757230 Chr.7: 100884459-100884459 on Build GRCh38
rs560845666 Chr.7: 100884286-100884286 on Build GRCh38
rs750126093 Chr.7: 100884256-100884256 on Build GRCh38
rs80278106 Chr.7: 100884429-100884429 on Build GRCh38
rs755262650 Chr.7: 100884590-100884590 on Build GRCh38
rs946466700 Chr.7: 100884444-100884444 on Build GRCh38
rs374697648 Chr.7: 100884714-100884714 on Build GRCh38
rs187232903 Chr.7: 100884314-100884314 on Build GRCh38
rs375882161 Chr.7: 100884508-100884508 on Build GRCh38
rs370063149 Chr.7: 100884473-100884473 on Build GRCh38
rs565636722 Chr.7: 100884334-100884334 on Build GRCh38
rs116129589 Chr.7: 100884519-100884519 on Build GRCh38
rs772392508 Chr.7: 100884406-100884406 on Build GRCh38
rs150930514 Chr.7: 100884481-100884481 on Build GRCh38
rs55833792 Chr.7: 100884718-100884718 on Build GRCh38
rs1043371329 Chr.7: 100884462-100884462 on Build GRCh38
rs1027630985 Chr.7: 100884672-100884672 on Build GRCh38
rs758451016 Chr.7: 100884570-100884570 on Build GRCh38
rs138621054 Chr.7: 100884294-100884294 on Build GRCh38
rs569140627 Chr.7: 100884444-100884449 on Build GRCh38
rs376926696 Chr.7: 100884342-100884342 on Build GRCh38
rs767770114 Chr.7: 100884356-100884356 on Build GRCh38
rs766105030 Chr.7: 100884478-100884478 on Build GRCh38
rs748538857 Chr.7: 100884276-100884276 on Build GRCh38
rs1057428345 Chr.7: 100884257-100884257 on Build GRCh38
rs758711103 Chr.7: 100884331-100884332 on Build GRCh38
rs778740620 Chr.7: 100884699-100884700 on Build GRCh38
rs937914351 Chr.7: 100884484-100884484 on Build GRCh38
rs373668899 Chr.7: 100884502-100884502 on Build GRCh38
rs920891164 Chr.7: 100884399-100884399 on Build GRCh38
rs769587965 Chr.7: 100884703-100884703 on Build GRCh38
rs183733865 Chr.7: 100884308-100884308 on Build GRCh38
rs774351392 Chr.7: 100884450-100884450 on Build GRCh38
rs780014188 Chr.7: 100884577-100884577 on Build GRCh38
rs768467767 Chr.7: 100884435-100884435 on Build GRCh38
rs751707381 Chr.7: 100884419-100884421 on Build GRCh38
rs775419276 Chr.7: 100884331-100884331 on Build GRCh38
rs749989925 Chr.7: 100884351-100884351 on Build GRCh38
rs372411794 Chr.7: 100884543-100884543 on Build GRCh38
rs541799837 Chr.7: 100884645-100884645 on Build GRCh38
rs547079893 Chr.7: 100884320-100884320 on Build GRCh38
rs774385955 Chr.7: 100884284-100884284 on Build GRCh38
rs376597506 Chr.7: 100884465-100884465 on Build GRCh38
rs199702658 Chr.7: 100884507-100884507 on Build GRCh38
rs531290886 Chr.7: 100884533-100884533 on Build GRCh38
rs368358714 Chr.7: 100884606-100884606 on Build GRCh38
rs140231361 Chr.7: 100884337-100884337 on Build GRCh38
rs200228895 Chr.7: 100884282-100884282 on Build GRCh38
rs15624 Chr.7: 100884405-100884405 on Build GRCh38
rs200398833 Chr.7: 100884557-100884557 on Build GRCh38
rs750920298 Chr.7: 100884569-100884569 on Build GRCh38
rs764566925 Chr.7: 100884501-100884501 on Build GRCh38
rs756497699 Chr.7: 100884499-100884499 on Build GRCh38
rs371129874 Chr.7: 100884275-100884275 on Build GRCh38
rs139372604 Chr.7: 100884544-100884544 on Build GRCh38
rs748076523 Chr.7: 100884531-100884531 on Build GRCh38
rs758513653 Chr.7: 100884511-100884511 on Build GRCh38
rs768457440 Chr.7: 100884530-100884532 on Build GRCh38
rs1037273810 Chr.7: 100884552-100884552 on Build GRCh38
rs754310318 Chr.7: 100884563-100884563 on Build GRCh38
rs372442352 Chr.7: 100884490-100884490 on Build GRCh38
rs762384483 Chr.7: 100884564-100884564 on Build GRCh38

View Full Product Details