Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100892253-100892754 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACHE
Entrez Gene ID
43
Gene Name
acetylcholinesterase (Cartwright blood group)
Gene Aliases
ACEE, ARACHE, N-ACHE, YT
UniGene
Hs.154495
Gene Chromosome Location
Chr.7: 100889994-100896133 on Build GRCh38
Transcripts
NM_000665.4 NM_001282449.1 NM_001302621.1 NM_001302622.1 NM_015831.2 XM_006715995.2 XM_011516225.1 XM_011516226.1 XM_011516228.2 XM_011516229.2 XM_017012219.1 XM_017012220.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1742682 c.1244A>G p.D415G Chr.7: 100892643-100892643 on Build GRCh38
6592018 c.1517G>A p.R506Q Chr.7: 100892370-100892370 on Build GRCh38
5871347 c.1516C>T p.R506* Chr.7: 100892371-100892371 on Build GRCh38
3081082 c.1401G>A p.T467T Chr.7: 100892486-100892486 on Build GRCh38
3381743 c.1327G>T p.A443S Chr.7: 100892560-100892560 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs938295495 Chr.7: 100892594-100892594 on Build GRCh38
rs370541497 Chr.7: 100892286-100892286 on Build GRCh38
rs761313292 Chr.7: 100892617-100892617 on Build GRCh38
rs867153702 Chr.7: 100892403-100892403 on Build GRCh38
rs201546756 Chr.7: 100892523-100892523 on Build GRCh38
rs139429533 Chr.7: 100892394-100892394 on Build GRCh38
rs766774822 Chr.7: 100892409-100892409 on Build GRCh38
rs41281001 Chr.7: 100892701-100892701 on Build GRCh38
rs377196885 Chr.7: 100892573-100892573 on Build GRCh38
rs992009907 Chr.7: 100892530-100892530 on Build GRCh38
rs377100607 Chr.7: 100892285-100892285 on Build GRCh38
rs372952913 Chr.7: 100892690-100892690 on Build GRCh38
rs781114815 Chr.7: 100892703-100892703 on Build GRCh38
rs769372048 Chr.7: 100892584-100892584 on Build GRCh38
rs759952889 Chr.7: 100892522-100892522 on Build GRCh38
rs749124873 Chr.7: 100892284-100892284 on Build GRCh38
rs745696709 Chr.7: 100892486-100892486 on Build GRCh38
rs200942047 Chr.7: 100892298-100892298 on Build GRCh38
rs1017295576 Chr.7: 100892447-100892447 on Build GRCh38
rs768103053 Chr.7: 100892441-100892441 on Build GRCh38
rs915997320 Chr.7: 100892544-100892544 on Build GRCh38
rs371841362 Chr.7: 100892572-100892572 on Build GRCh38
rs776324892 Chr.7: 100892402-100892402 on Build GRCh38
rs772297940 Chr.7: 100892727-100892727 on Build GRCh38
rs370742086 Chr.7: 100892652-100892652 on Build GRCh38
rs746488915 Chr.7: 100892502-100892502 on Build GRCh38
rs145048252 Chr.7: 100892438-100892438 on Build GRCh38
rs752650772 Chr.7: 100892444-100892444 on Build GRCh38
rs145811992 Chr.7: 100892472-100892472 on Build GRCh38
rs61729577 Chr.7: 100892550-100892550 on Build GRCh38
rs374012624 Chr.7: 100892591-100892591 on Build GRCh38
rs761506155 Chr.7: 100892513-100892513 on Build GRCh38
rs774542226 Chr.7: 100892516-100892516 on Build GRCh38
rs7636 Chr.7: 100892456-100892456 on Build GRCh38
rs751422399 Chr.7: 100892340-100892340 on Build GRCh38
rs370969881 Chr.7: 100892310-100892310 on Build GRCh38
rs145683970 Chr.7: 100892531-100892531 on Build GRCh38
rs199565888 Chr.7: 100892429-100892429 on Build GRCh38
rs752130937 Chr.7: 100892306-100892306 on Build GRCh38
rs775874742 Chr.7: 100892730-100892730 on Build GRCh38
rs779225043 Chr.7: 100892554-100892554 on Build GRCh38
rs371120982 Chr.7: 100892590-100892590 on Build GRCh38
rs113533105 Chr.7: 100892510-100892510 on Build GRCh38
rs535618245 Chr.7: 100892539-100892539 on Build GRCh38
rs199872491 Chr.7: 100892630-100892630 on Build GRCh38
rs766862660 Chr.7: 100892305-100892305 on Build GRCh38
rs760239221 Chr.7: 100892396-100892396 on Build GRCh38
rs751381796 Chr.7: 100892699-100892699 on Build GRCh38
rs934885760 Chr.7: 100892722-100892722 on Build GRCh38
rs763680727 Chr.7: 100892397-100892397 on Build GRCh38
rs754089513 Chr.7: 100892535-100892535 on Build GRCh38
rs149813374 Chr.7: 100892459-100892459 on Build GRCh38
rs772928592 Chr.7: 100892585-100892585 on Build GRCh38
rs550436794 Chr.7: 100892564-100892564 on Build GRCh38
rs756253277 Chr.7: 100892337-100892337 on Build GRCh38
rs750130512 Chr.7: 100892555-100892555 on Build GRCh38
rs770906989 Chr.7: 100892363-100892363 on Build GRCh38
rs148991713 Chr.7: 100892515-100892515 on Build GRCh38
rs753301602 Chr.7: 100892526-100892526 on Build GRCh38
rs771862488 Chr.7: 100892487-100892487 on Build GRCh38
rs774442656 Chr.7: 100892287-100892287 on Build GRCh38
rs758779224 Chr.7: 100892299-100892299 on Build GRCh38
rs781179503 Chr.7: 100892319-100892319 on Build GRCh38
rs151107784 Chr.7: 100892453-100892453 on Build GRCh38
rs778143437 Chr.7: 100892339-100892339 on Build GRCh38
rs376838625 Chr.7: 100892616-100892616 on Build GRCh38
rs771585174 Chr.7: 100892382-100892382 on Build GRCh38
rs199754698 Chr.7: 100892288-100892288 on Build GRCh38
rs947520697 Chr.7: 100892326-100892326 on Build GRCh38
rs774422002 Chr.7: 100892370-100892370 on Build GRCh38
rs367977631 Chr.7: 100892563-100892563 on Build GRCh38
rs753928751 Chr.7: 100892627-100892627 on Build GRCh38
rs750362321 Chr.7: 100892711-100892711 on Build GRCh38
rs903399345 Chr.7: 100892341-100892341 on Build GRCh38
rs745829096 Chr.7: 100892372-100892372 on Build GRCh38
rs747602194 Chr.7: 100892704-100892704 on Build GRCh38
rs748284210 Chr.7: 100892321-100892321 on Build GRCh38
rs1030385619 Chr.7: 100892377-100892377 on Build GRCh38
rs374462106 Chr.7: 100892406-100892406 on Build GRCh38
rs765366984 Chr.7: 100892651-100892651 on Build GRCh38
rs780056308 Chr.7: 100892496-100892496 on Build GRCh38
rs866284496 Chr.7: 100892620-100892620 on Build GRCh38
rs760828686 Chr.7: 100892528-100892528 on Build GRCh38
rs367893261 Chr.7: 100892291-100892291 on Build GRCh38
rs546907479 Chr.7: 100892509-100892509 on Build GRCh38
rs755706015 Chr.7: 100892705-100892705 on Build GRCh38
rs74418820 Chr.7: 100892621-100892621 on Build GRCh38
rs758098245 Chr.7: 100892678-100892678 on Build GRCh38
rs755962641 Chr.7: 100892723-100892723 on Build GRCh38
rs914753658 Chr.7: 100892548-100892548 on Build GRCh38
rs749032132 Chr.7: 100892361-100892361 on Build GRCh38
rs528356898 Chr.7: 100892597-100892597 on Build GRCh38
rs760291349 Chr.7: 100892293-100892293 on Build GRCh38
rs11554090 Chr.7: 100892368-100892368 on Build GRCh38
rs1007745520 Chr.7: 100892432-100892432 on Build GRCh38
rs766999179 Chr.7: 100892289-100892289 on Build GRCh38
rs763756408 Chr.7: 100892296-100892296 on Build GRCh38

View Full Product Details