Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 103102131-103102677 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NAPEPLD
Entrez Gene ID
222236
Gene Name
N-acyl phosphatidylethanolamine phospholipase D
Gene Aliases
FMP30, NAPE-PLD
UniGene
Hs.324271
Gene Chromosome Location
Chr.7: 103099576-103149560 on Build GRCh38
Transcripts
NM_001122838.1 NM_198990.4 XM_005250214.4 XM_005250215.2 XM_005250218.2 XM_011515933.2 XM_011515934.2 XM_017011854.1 XM_017011855.1

SNP Detail

SNP ID Chromosome Location
rs189342666 Chr.7: 103102653-103102653 on Build GRCh38
rs918444163 Chr.7: 103102168-103102168 on Build GRCh38
rs1012575026 Chr.7: 103102236-103102236 on Build GRCh38
rs765959698 Chr.7: 103102498-103102498 on Build GRCh38
rs1042623413 Chr.7: 103102318-103102318 on Build GRCh38
rs1053770495 Chr.7: 103102257-103102257 on Build GRCh38
rs938114486 Chr.7: 103102239-103102239 on Build GRCh38
rs893842893 Chr.7: 103102307-103102307 on Build GRCh38
rs777270539 Chr.7: 103102628-103102628 on Build GRCh38
rs1036431633 Chr.7: 103102160-103102160 on Build GRCh38
rs372924237 Chr.7: 103102172-103102172 on Build GRCh38
rs901425633 Chr.7: 103102355-103102355 on Build GRCh38
rs895024039 Chr.7: 103102216-103102216 on Build GRCh38
rs1030684054 Chr.7: 103102322-103102322 on Build GRCh38
rs374231229 Chr.7: 103102200-103102200 on Build GRCh38
rs766563080 Chr.7: 103102387-103102387 on Build GRCh38
rs928704903 Chr.7: 103102558-103102558 on Build GRCh38
rs926468100 Chr.7: 103102482-103102482 on Build GRCh38
rs138191761 Chr.7: 103102497-103102497 on Build GRCh38
rs534716852 Chr.7: 103102496-103102496 on Build GRCh38
rs756352942 Chr.7: 103102267-103102267 on Build GRCh38
rs966623565 Chr.7: 103102375-103102375 on Build GRCh38
rs919186242 Chr.7: 103102164-103102164 on Build GRCh38
rs538497411 Chr.7: 103102446-103102446 on Build GRCh38
rs757849633 Chr.7: 103102535-103102535 on Build GRCh38
rs915349436 Chr.7: 103102547-103102547 on Build GRCh38
rs779356448 Chr.7: 103102528-103102528 on Build GRCh38
rs569965324 Chr.7: 103102311-103102311 on Build GRCh38
rs754204895 Chr.7: 103102475-103102475 on Build GRCh38
rs1056795477 Chr.7: 103102212-103102213 on Build GRCh38
rs938066765 Chr.7: 103102483-103102483 on Build GRCh38
rs926685048 Chr.7: 103102204-103102204 on Build GRCh38
rs530401291 Chr.7: 103102374-103102374 on Build GRCh38
rs566139321 Chr.7: 103102635-103102635 on Build GRCh38
rs1046664981 Chr.7: 103102196-103102196 on Build GRCh38
rs1056338972 Chr.7: 103102607-103102607 on Build GRCh38
rs764516311 Chr.7: 103102611-103102613 on Build GRCh38
rs945358493 Chr.7: 103102575-103102575 on Build GRCh38
rs997660674 Chr.7: 103102431-103102431 on Build GRCh38
rs1028944720 Chr.7: 103102362-103102362 on Build GRCh38
rs998331766 Chr.7: 103102361-103102361 on Build GRCh38
rs971085294 Chr.7: 103102466-103102466 on Build GRCh38
rs1042790841 Chr.7: 103102593-103102593 on Build GRCh38
rs201353155 Chr.7: 103102247-103102247 on Build GRCh38
rs1015979798 Chr.7: 103102399-103102399 on Build GRCh38
rs751223328 Chr.7: 103102502-103102502 on Build GRCh38
rs186440657 Chr.7: 103102419-103102419 on Build GRCh38
rs909240390 Chr.7: 103102512-103102513 on Build GRCh38
rs750603801 Chr.7: 103102240-103102240 on Build GRCh38
rs962017368 Chr.7: 103102518-103102518 on Build GRCh38
rs919145062 Chr.7: 103102459-103102459 on Build GRCh38
rs548901609 Chr.7: 103102541-103102542 on Build GRCh38

View Full Product Details