Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 103363880-103364399 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSMC2
Entrez Gene ID
5701
Gene Name
proteasome 26S subunit, ATPase 2
Gene Aliases
MSS1, Nbla10058, S7
UniGene
Hs.437366
Gene Chromosome Location
Chr.7: 103344405-103369395 on Build GRCh38
Transcripts
NM_002803.3 NM_001321787.1 NM_206883.2 NM_206884.2 NM_206885.2 NR_135802.1

Gene Symbol
SLC26A5
Entrez Gene ID
375611
Gene Name
solute carrier family 26 member 5
Gene Aliases
DFNB61, PRES
UniGene
Hs.585146
Gene Chromosome Location
Chr.7: 103352598-103446177 on Build GRCh38
Transcripts
NM_002803.3 NM_001321787.1 NM_206883.2 NM_206884.2 NM_206885.2 NR_135802.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3876799 c.677C>T p.A226V Chr.7: 103364228-103364228 on Build GRCh38
1568182 c.633C>T p.G211G Chr.7: 103364184-103364184 on Build GRCh38
3876800 c.688G>C p.A230P Chr.7: 103364239-103364239 on Build GRCh38
3876801 c.705G>A p.A235A Chr.7: 103364256-103364256 on Build GRCh38
1083570 c.750C>T p.V250V Chr.7: 103364301-103364301 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374959956 Chr.7: 103364150-103364150 on Build GRCh38
rs774084514 Chr.7: 103364245-103364245 on Build GRCh38
rs556239820 Chr.7: 103364029-103364029 on Build GRCh38
rs545478396 Chr.7: 103364376-103364376 on Build GRCh38
rs1054191925 Chr.7: 103364235-103364235 on Build GRCh38
rs751235684 Chr.7: 103364105-103364105 on Build GRCh38
rs750961758 Chr.7: 103364347-103364347 on Build GRCh38
rs1013520746 Chr.7: 103363931-103363931 on Build GRCh38
rs376164420 Chr.7: 103364211-103364211 on Build GRCh38
rs746224068 Chr.7: 103364136-103364136 on Build GRCh38
rs201639323 Chr.7: 103364333-103364333 on Build GRCh38
rs1009847555 Chr.7: 103364330-103364330 on Build GRCh38
rs144680798 Chr.7: 103364256-103364256 on Build GRCh38
rs748982263 Chr.7: 103364363-103364363 on Build GRCh38
rs541830606 Chr.7: 103364045-103364045 on Build GRCh38
rs758727100 Chr.7: 103364352-103364352 on Build GRCh38
rs956043497 Chr.7: 103364350-103364350 on Build GRCh38
rs1045617774 Chr.7: 103364193-103364193 on Build GRCh38
rs756709619 Chr.7: 103363957-103363957 on Build GRCh38
rs1018813358 Chr.7: 103363953-103363953 on Build GRCh38
rs988710880 Chr.7: 103363941-103363941 on Build GRCh38
rs922044412 Chr.7: 103364027-103364027 on Build GRCh38
rs960195465 Chr.7: 103363909-103363909 on Build GRCh38
rs141423831 Chr.7: 103364128-103364128 on Build GRCh38
rs61733321 Chr.7: 103364205-103364205 on Build GRCh38
rs988038101 Chr.7: 103364016-103364016 on Build GRCh38
rs767092398 Chr.7: 103364259-103364259 on Build GRCh38
rs199759890 Chr.7: 103364167-103364167 on Build GRCh38
rs377670982 Chr.7: 103364104-103364104 on Build GRCh38
rs955719003 Chr.7: 103363940-103363940 on Build GRCh38
rs186622343 Chr.7: 103363923-103363923 on Build GRCh38
rs752093947 Chr.7: 103363956-103363956 on Build GRCh38
rs750692030 Chr.7: 103364229-103364229 on Build GRCh38
rs780322948 Chr.7: 103364184-103364184 on Build GRCh38
rs780295964 Chr.7: 103364354-103364354 on Build GRCh38
rs547017233 Chr.7: 103364071-103364073 on Build GRCh38
rs776715911 Chr.7: 103364213-103364213 on Build GRCh38
rs369074674 Chr.7: 103364057-103364057 on Build GRCh38
rs956243814 Chr.7: 103363992-103363992 on Build GRCh38
rs778150181 Chr.7: 103364134-103364134 on Build GRCh38
rs574445637 Chr.7: 103364036-103364036 on Build GRCh38
rs147608353 Chr.7: 103364298-103364298 on Build GRCh38
rs199732205 Chr.7: 103364110-103364110 on Build GRCh38
rs978863024 Chr.7: 103363979-103363979 on Build GRCh38
rs974512403 Chr.7: 103363997-103363997 on Build GRCh38
rs780859411 Chr.7: 103364114-103364114 on Build GRCh38
rs370246510 Chr.7: 103364220-103364220 on Build GRCh38
rs1033061191 Chr.7: 103363926-103363926 on Build GRCh38
rs940654559 Chr.7: 103364088-103364088 on Build GRCh38
rs780658965 Chr.7: 103363990-103363990 on Build GRCh38
rs1057360971 Chr.7: 103364131-103364132 on Build GRCh38
rs764449288 Chr.7: 103363912-103363912 on Build GRCh38
rs752992304 Chr.7: 103363983-103363983 on Build GRCh38
rs568232731 Chr.7: 103364223-103364223 on Build GRCh38
rs897454161 Chr.7: 103364121-103364122 on Build GRCh38
rs755225826 Chr.7: 103364358-103364358 on Build GRCh38
rs755737635 Chr.7: 103364301-103364301 on Build GRCh38

View Full Product Details