Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 113080499-113081015 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR85
Entrez Gene ID
54329
Gene Name
G protein-coupled receptor 85
Gene Aliases
SREB, SREB2
UniGene
Hs.152009
Gene Chromosome Location
Chr.7: 113080413-113087778 on Build GRCh38
Transcripts
NM_001146265.1 NM_001146266.1 NM_001146267.1 NM_018970.6

SNP Detail

SNP ID Chromosome Location
rs114486665 Chr.7: 113080661-113080661 on Build GRCh38
rs879250486 Chr.7: 113080825-113080825 on Build GRCh38
rs977220093 Chr.7: 113080605-113080605 on Build GRCh38
rs570427934 Chr.7: 113080888-113080888 on Build GRCh38
rs943627670 Chr.7: 113080622-113080622 on Build GRCh38
rs527597095 Chr.7: 113080826-113080826 on Build GRCh38
rs765552097 Chr.7: 113080584-113080584 on Build GRCh38
rs988229674 Chr.7: 113080858-113080858 on Build GRCh38
rs763635742 Chr.7: 113080579-113080579 on Build GRCh38
rs780132191 Chr.7: 113080898-113080898 on Build GRCh38
rs893824217 Chr.7: 113080582-113080582 on Build GRCh38
rs62488674 Chr.7: 113080677-113080677 on Build GRCh38
rs184515966 Chr.7: 113080540-113080540 on Build GRCh38
rs973099587 Chr.7: 113080755-113080755 on Build GRCh38
rs775025410 Chr.7: 113080531-113080531 on Build GRCh38
rs758322686 Chr.7: 113080833-113080833 on Build GRCh38
rs1039672418 Chr.7: 113080842-113080842 on Build GRCh38
rs2255594 Chr.7: 113080578-113080578 on Build GRCh38
rs1035003510 Chr.7: 113080811-113080811 on Build GRCh38
rs188185275 Chr.7: 113080600-113080600 on Build GRCh38
rs910730009 Chr.7: 113080929-113080929 on Build GRCh38
rs778626564 Chr.7: 113080894-113080894 on Build GRCh38
rs539467570 Chr.7: 113080668-113080668 on Build GRCh38
rs763603472 Chr.7: 113080606-113080606 on Build GRCh38
rs1022804767 Chr.7: 113080785-113080785 on Build GRCh38
rs943596447 Chr.7: 113080934-113080934 on Build GRCh38
rs760318679 Chr.7: 113080536-113080536 on Build GRCh38
rs2293369 Chr.7: 113080769-113080769 on Build GRCh38
rs961014070 Chr.7: 113080817-113080817 on Build GRCh38
rs370517183 Chr.7: 113080550-113080550 on Build GRCh38
rs929425596 Chr.7: 113080538-113080538 on Build GRCh38
rs912094941 Chr.7: 113080809-113080809 on Build GRCh38
rs755235105 Chr.7: 113080673-113080673 on Build GRCh38
rs1003215118 Chr.7: 113080810-113080810 on Build GRCh38
rs966765679 Chr.7: 113080592-113080592 on Build GRCh38
rs551856447 Chr.7: 113080882-113080882 on Build GRCh38
rs1042407108 Chr.7: 113080899-113080899 on Build GRCh38
rs993239083 Chr.7: 113080726-113080726 on Build GRCh38
rs542076932 Chr.7: 113080761-113080761 on Build GRCh38
rs1008260386 Chr.7: 113080560-113080560 on Build GRCh38
rs1041045473 Chr.7: 113080961-113080961 on Build GRCh38
rs902596116 Chr.7: 113080913-113080913 on Build GRCh38
rs746979617 Chr.7: 113080914-113080914 on Build GRCh38
rs1047961858 Chr.7: 113080735-113080735 on Build GRCh38
rs112486695 Chr.7: 113080990-113080990 on Build GRCh38
rs74798468 Chr.7: 113080545-113080545 on Build GRCh38
rs929080006 Chr.7: 113080775-113080775 on Build GRCh38
rs1051926795 Chr.7: 113080778-113080778 on Build GRCh38
rs529914408 Chr.7: 113080729-113080729 on Build GRCh38
rs553946190 Chr.7: 113080585-113080585 on Build GRCh38
rs781414262 Chr.7: 113080752-113080752 on Build GRCh38
rs56080411 Chr.7: 113080660-113080660 on Build GRCh38

View Full Product Details