Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 123954065-123954565 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPAM1
Entrez Gene ID
6677
Gene Name
sperm adhesion molecule 1
Gene Aliases
HEL-S-96n, HYA1, HYAL1, HYAL3, HYAL5, PH-20, PH20, SPAG15
UniGene
Hs.121494
Gene Chromosome Location
Chr.7: 123925232-123971410 on Build GRCh38
Transcripts
NM_001174044.1 NM_001174045.1 NM_001174046.1 NM_003117.4 NM_153189.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5586658 c.649G>A p.G217S Chr.7: 123954219-123954219 on Build GRCh38
3366755 c.907T>A p.Y303N Chr.7: 123954477-123954477 on Build GRCh38
3632757 c.797A>T p.N266I Chr.7: 123954367-123954367 on Build GRCh38
3632740 c.591G>A p.K197K Chr.7: 123954161-123954161 on Build GRCh38
3928799 c.686A>T p.Y229F Chr.7: 123954256-123954256 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375604421 Chr.7: 123954272-123954272 on Build GRCh38
rs769222158 Chr.7: 123954236-123954236 on Build GRCh38
rs149929945 Chr.7: 123954138-123954138 on Build GRCh38
rs868193749 Chr.7: 123954356-123954356 on Build GRCh38
rs763650203 Chr.7: 123954290-123954290 on Build GRCh38
rs151176936 Chr.7: 123954345-123954345 on Build GRCh38
rs969456380 Chr.7: 123954400-123954400 on Build GRCh38
rs762640029 Chr.7: 123954412-123954412 on Build GRCh38
rs754183378 Chr.7: 123954390-123954390 on Build GRCh38
rs746068102 Chr.7: 123954227-123954227 on Build GRCh38
rs113752093 Chr.7: 123954190-123954190 on Build GRCh38
rs755292470 Chr.7: 123954477-123954477 on Build GRCh38
rs773490507 Chr.7: 123954266-123954266 on Build GRCh38
rs758872341 Chr.7: 123954478-123954478 on Build GRCh38
rs561593355 Chr.7: 123954186-123954186 on Build GRCh38
rs1010819869 Chr.7: 123954442-123954442 on Build GRCh38
rs765285640 Chr.7: 123954443-123954447 on Build GRCh38
rs751473529 Chr.7: 123954298-123954298 on Build GRCh38
rs2285996 Chr.7: 123954152-123954152 on Build GRCh38
rs779630580 Chr.7: 123954407-123954408 on Build GRCh38
rs544888620 Chr.7: 123954166-123954166 on Build GRCh38
rs773877910 Chr.7: 123954432-123954432 on Build GRCh38
rs185873814 Chr.7: 123954444-123954444 on Build GRCh38
rs1035171373 Chr.7: 123954419-123954419 on Build GRCh38
rs188850653 Chr.7: 123954499-123954499 on Build GRCh38
rs749343447 Chr.7: 123954235-123954235 on Build GRCh38
rs370092959 Chr.7: 123954098-123954098 on Build GRCh38
rs952962850 Chr.7: 123954311-123954311 on Build GRCh38
rs748725260 Chr.7: 123954256-123954256 on Build GRCh38
rs769135908 Chr.7: 123954221-123954221 on Build GRCh38
rs940693148 Chr.7: 123954200-123954200 on Build GRCh38
rs775468710 Chr.7: 123954102-123954102 on Build GRCh38
rs757030216 Chr.7: 123954220-123954220 on Build GRCh38
rs776334838 Chr.7: 123954194-123954194 on Build GRCh38
rs532636427 Chr.7: 123954406-123954406 on Build GRCh38
rs747607749 Chr.7: 123954483-123954483 on Build GRCh38
rs376151172 Chr.7: 123954198-123954198 on Build GRCh38
rs748896299 Chr.7: 123954518-123954518 on Build GRCh38
rs993551021 Chr.7: 123954525-123954525 on Build GRCh38
rs772738728 Chr.7: 123954405-123954405 on Build GRCh38
rs760385447 Chr.7: 123954459-123954459 on Build GRCh38
rs781010587 Chr.7: 123954221-123954221 on Build GRCh38
rs769313920 Chr.7: 123954151-123954151 on Build GRCh38
rs190469752 Chr.7: 123954289-123954289 on Build GRCh38
rs771392653 Chr.7: 123954408-123954408 on Build GRCh38
rs146942265 Chr.7: 123954399-123954399 on Build GRCh38
rs1035151771 Chr.7: 123954497-123954497 on Build GRCh38
rs368148854 Chr.7: 123954280-123954280 on Build GRCh38
rs148629332 Chr.7: 123954222-123954222 on Build GRCh38
rs774997522 Chr.7: 123954286-123954286 on Build GRCh38
rs748635413 Chr.7: 123954404-123954404 on Build GRCh38
rs200315691 Chr.7: 123954265-123954265 on Build GRCh38
rs377689190 Chr.7: 123954201-123954201 on Build GRCh38
rs984286192 Chr.7: 123954340-123954340 on Build GRCh38
rs371105891 Chr.7: 123954202-123954202 on Build GRCh38
rs769274268 Chr.7: 123954537-123954537 on Build GRCh38
rs142644076 Chr.7: 123954443-123954443 on Build GRCh38
rs142125830 Chr.7: 123954264-123954264 on Build GRCh38
rs780137128 Chr.7: 123954230-123954230 on Build GRCh38
rs746131722 Chr.7: 123954093-123954093 on Build GRCh38
rs150235912 Chr.7: 123954484-123954484 on Build GRCh38
rs768314840 Chr.7: 123954520-123954520 on Build GRCh38
rs756316980 Chr.7: 123954228-123954228 on Build GRCh38
rs773720006 Chr.7: 123954153-123954153 on Build GRCh38
rs552422838 Chr.7: 123954409-123954409 on Build GRCh38
rs982591467 Chr.7: 123954457-123954457 on Build GRCh38
rs996310163 Chr.7: 123954249-123954249 on Build GRCh38
rs867117620 Chr.7: 123954121-123954121 on Build GRCh38
rs868062838 Chr.7: 123954355-123954355 on Build GRCh38
rs774996576 Chr.7: 123954238-123954238 on Build GRCh38
rs754091551 Chr.7: 123954466-123954466 on Build GRCh38
rs778138042 Chr.7: 123954481-123954481 on Build GRCh38
rs141109529 Chr.7: 123954381-123954381 on Build GRCh38
rs750170851 Chr.7: 123954448-123954448 on Build GRCh38
rs961989569 Chr.7: 123954257-123954257 on Build GRCh38
rs939146862 Chr.7: 123954149-123954149 on Build GRCh38
rs766257029 Chr.7: 123954463-123954463 on Build GRCh38
rs780147908 Chr.7: 123954384-123954384 on Build GRCh38
rs755097882 Chr.7: 123954393-123954393 on Build GRCh38
rs762275983 Chr.7: 123954156-123954156 on Build GRCh38
rs761776007 Chr.7: 123954322-123954322 on Build GRCh38
rs893737869 Chr.7: 123954163-123954163 on Build GRCh38
rs750204136 Chr.7: 123954363-123954363 on Build GRCh38
rs147745114 Chr.7: 123954418-123954418 on Build GRCh38

View Full Product Details