Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 124764590-124765081 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR37
Entrez Gene ID
2861
Gene Name
G protein-coupled receptor 37
Gene Aliases
EDNRBL, PAELR, hET(B)R-LP
UniGene
Hs.406094
Gene Chromosome Location
Chr.7: 124745601-124766025 on Build GRCh38
Transcripts
NM_005302.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4540302 c.281G>A p.R94H Chr.7: 124764696-124764696 on Build GRCh38
3877765 c.275C>T p.P92L Chr.7: 124764702-124764702 on Build GRCh38
5963303 c.180A>T p.G60G Chr.7: 124764797-124764797 on Build GRCh38
5011852 c.332G>C p.G111A Chr.7: 124764645-124764645 on Build GRCh38
6674137 c.137C>A p.P46H Chr.7: 124764840-124764840 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750797121 Chr.7: 124764931-124764931 on Build GRCh38
rs775239448 Chr.7: 124764765-124764765 on Build GRCh38
rs767987181 Chr.7: 124764775-124764775 on Build GRCh38
rs778606606 Chr.7: 124764939-124764939 on Build GRCh38
rs542190726 Chr.7: 124765003-124765003 on Build GRCh38
rs990289975 Chr.7: 124764731-124764731 on Build GRCh38
rs764215446 Chr.7: 124764984-124764984 on Build GRCh38
rs62638682 Chr.7: 124764654-124764654 on Build GRCh38
rs769553821 Chr.7: 124764656-124764656 on Build GRCh38
rs770645657 Chr.7: 124764904-124764904 on Build GRCh38
rs992789587 Chr.7: 124764959-124764959 on Build GRCh38
rs755125416 Chr.7: 124764637-124764637 on Build GRCh38
rs779080776 Chr.7: 124764720-124764720 on Build GRCh38
rs767160980 Chr.7: 124764673-124764673 on Build GRCh38
rs754962917 Chr.7: 124764711-124764711 on Build GRCh38
rs377727646 Chr.7: 124764669-124764669 on Build GRCh38
rs188114495 Chr.7: 124765035-124765035 on Build GRCh38
rs62638681 Chr.7: 124764929-124764929 on Build GRCh38
rs147396306 Chr.7: 124764815-124764815 on Build GRCh38
rs754108421 Chr.7: 124764707-124764707 on Build GRCh38
rs767439352 Chr.7: 124764810-124764810 on Build GRCh38
rs766183726 Chr.7: 124764822-124764822 on Build GRCh38
rs373034842 Chr.7: 124764658-124764658 on Build GRCh38
rs763276308 Chr.7: 124764970-124764970 on Build GRCh38
rs548740719 Chr.7: 124764874-124764874 on Build GRCh38
rs996129085 Chr.7: 124764738-124764738 on Build GRCh38
rs376765252 Chr.7: 124764650-124764650 on Build GRCh38
rs370030753 Chr.7: 124764701-124764701 on Build GRCh38
rs1008493878 Chr.7: 124764628-124764628 on Build GRCh38
rs779265027 Chr.7: 124764641-124764641 on Build GRCh38
rs761290228 Chr.7: 124764804-124764804 on Build GRCh38
rs1003898 Chr.7: 124765054-124765054 on Build GRCh38
rs763686517 Chr.7: 124764665-124764665 on Build GRCh38
rs1003897 Chr.7: 124765056-124765056 on Build GRCh38
rs893943695 Chr.7: 124764998-124764998 on Build GRCh38
rs1032466965 Chr.7: 124764693-124764693 on Build GRCh38
rs533008895 Chr.7: 124764732-124764732 on Build GRCh38
rs1041021316 Chr.7: 124764943-124764943 on Build GRCh38
rs547579249 Chr.7: 124764675-124764675 on Build GRCh38
rs760441116 Chr.7: 124764818-124764818 on Build GRCh38
rs759127920 Chr.7: 124764903-124764916 on Build GRCh38
rs750929196 Chr.7: 124764652-124764652 on Build GRCh38
rs749004173 Chr.7: 124764767-124764767 on Build GRCh38
rs147161580 Chr.7: 124764935-124764935 on Build GRCh38
rs749366749 Chr.7: 124764623-124764623 on Build GRCh38
rs773084533 Chr.7: 124764898-124764898 on Build GRCh38
rs747370351 Chr.7: 124764746-124764746 on Build GRCh38
rs760503220 Chr.7: 124764648-124764648 on Build GRCh38
rs759342244 Chr.7: 124764952-124764952 on Build GRCh38
rs781609406 Chr.7: 124764930-124764930 on Build GRCh38
rs750206983 Chr.7: 124764674-124764674 on Build GRCh38
rs568704083 Chr.7: 124764891-124764891 on Build GRCh38
rs957174572 Chr.7: 124764695-124764695 on Build GRCh38
rs760581985 Chr.7: 124764846-124764846 on Build GRCh38
rs768344239 Chr.7: 124764894-124764894 on Build GRCh38
rs773928348 Chr.7: 124764801-124764801 on Build GRCh38
rs771458806 Chr.7: 124764763-124764763 on Build GRCh38
rs775727862 Chr.7: 124764966-124764966 on Build GRCh38
rs747665043 Chr.7: 124764895-124764895 on Build GRCh38
rs1037581854 Chr.7: 124764712-124764712 on Build GRCh38
rs752155979 Chr.7: 124764858-124764858 on Build GRCh38
rs200282466 Chr.7: 124764696-124764696 on Build GRCh38
rs561953129 Chr.7: 124765026-124765026 on Build GRCh38
rs750943043 Chr.7: 124765002-124765002 on Build GRCh38
rs373879644 Chr.7: 124764946-124764946 on Build GRCh38
rs141580656 Chr.7: 124764996-124764996 on Build GRCh38
rs759841234 Chr.7: 124764886-124764886 on Build GRCh38
rs747778141 Chr.7: 124764944-124764944 on Build GRCh38
rs143391902 Chr.7: 124764906-124764906 on Build GRCh38
rs765490264 Chr.7: 124764910-124764910 on Build GRCh38
rs149569764 Chr.7: 124764737-124764737 on Build GRCh38
rs556616401 Chr.7: 124764949-124764949 on Build GRCh38
rs969032618 Chr.7: 124764614-124764614 on Build GRCh38
rs138132243 Chr.7: 124764915-124764915 on Build GRCh38
rs776497146 Chr.7: 124764951-124764951 on Build GRCh38
rs944623116 Chr.7: 124765049-124765049 on Build GRCh38
rs771620776 Chr.7: 124764897-124764897 on Build GRCh38
rs766685596 Chr.7: 124765012-124765012 on Build GRCh38
rs931777346 Chr.7: 124765017-124765017 on Build GRCh38
rs769522179 Chr.7: 124764962-124764962 on Build GRCh38
rs1050371605 Chr.7: 124765025-124765025 on Build GRCh38
rs764832581 Chr.7: 124764977-124764977 on Build GRCh38
rs751679891 Chr.7: 124764989-124764989 on Build GRCh38
rs752806538 Chr.7: 124764849-124764849 on Build GRCh38
rs753965814 Chr.7: 124764834-124764834 on Build GRCh38
rs781696266 Chr.7: 124764880-124764880 on Build GRCh38
rs201716246 Chr.7: 124764907-124764907 on Build GRCh38
rs758470818 Chr.7: 124764853-124764853 on Build GRCh38
rs755231420 Chr.7: 124764843-124764843 on Build GRCh38
rs1011337202 Chr.7: 124765037-124765037 on Build GRCh38
rs199529926 Chr.7: 124764857-124764857 on Build GRCh38
rs780313583 Chr.7: 124764615-124764615 on Build GRCh38
rs58723361 Chr.7: 124764925-124764925 on Build GRCh38
rs778478649 Chr.7: 124764740-124764740 on Build GRCh38
rs763099360 Chr.7: 124764917-124764917 on Build GRCh38
rs760637240 Chr.7: 124764903-124764903 on Build GRCh38
rs112834318 Chr.7: 124764666-124764666 on Build GRCh38

View Full Product Details