Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 151213455-151213961 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCF2
Entrez Gene ID
10061
Gene Name
ATP binding cassette subfamily F member 2
Gene Aliases
ABC28, EST133090, HUSSY-18, HUSSY18
UniGene
Hs.654958
Gene Chromosome Location
Chr.7: 151207837-151227374 on Build GRCh38
Transcripts
NM_005692.4 NM_007189.2 XM_017011639.1 XM_017011640.1

SNP Detail

SNP ID Chromosome Location
rs80135613 Chr.7: 151213478-151213478 on Build GRCh38
rs925047192 Chr.7: 151213536-151213539 on Build GRCh38
rs529458759 Chr.7: 151213492-151213492 on Build GRCh38
rs970398429 Chr.7: 151213838-151213838 on Build GRCh38
rs2415 Chr.7: 151213549-151213549 on Build GRCh38
rs549601247 Chr.7: 151213550-151213550 on Build GRCh38
rs1048684841 Chr.7: 151213501-151213501 on Build GRCh38
rs905670578 Chr.7: 151213939-151213939 on Build GRCh38
rs66608097 Chr.7: 151213658-151213658 on Build GRCh38
rs575092214 Chr.7: 151213888-151213888 on Build GRCh38
rs138085673 Chr.7: 151213492-151213495 on Build GRCh38
rs1030246757 Chr.7: 151213491-151213492 on Build GRCh38
rs764496570 Chr.7: 151213601-151213601 on Build GRCh38
rs773513688 Chr.7: 151213924-151213924 on Build GRCh38
rs1052623725 Chr.7: 151213839-151213839 on Build GRCh38
rs984704888 Chr.7: 151213890-151213890 on Build GRCh38
rs964632389 Chr.7: 151213488-151213488 on Build GRCh38
rs370478187 Chr.7: 151213910-151213910 on Build GRCh38
rs1023336426 Chr.7: 151213783-151213784 on Build GRCh38
rs949638727 Chr.7: 151213589-151213589 on Build GRCh38
rs571639383 Chr.7: 151213696-151213696 on Build GRCh38
rs905098405 Chr.7: 151213893-151213893 on Build GRCh38
rs993355174 Chr.7: 151213660-151213660 on Build GRCh38
rs1030693092 Chr.7: 151213701-151213701 on Build GRCh38
rs998367131 Chr.7: 151213568-151213568 on Build GRCh38
rs763565746 Chr.7: 151213595-151213595 on Build GRCh38
rs933750679 Chr.7: 151213831-151213831 on Build GRCh38
rs1035286537 Chr.7: 151213651-151213651 on Build GRCh38
rs942962941 Chr.7: 151213738-151213738 on Build GRCh38
rs569725194 Chr.7: 151213520-151213520 on Build GRCh38
rs902408850 Chr.7: 151213706-151213706 on Build GRCh38
rs145275975 Chr.7: 151213644-151213644 on Build GRCh38
rs778407938 Chr.7: 151213645-151213645 on Build GRCh38
rs370355313 Chr.7: 151213614-151213614 on Build GRCh38
rs966979689 Chr.7: 151213518-151213521 on Build GRCh38
rs572783518 Chr.7: 151213813-151213813 on Build GRCh38
rs1010548313 Chr.7: 151213884-151213884 on Build GRCh38
rs990488579 Chr.7: 151213756-151213756 on Build GRCh38
rs147217005 Chr.7: 151213771-151213771 on Build GRCh38
rs892168526 Chr.7: 151213600-151213600 on Build GRCh38
rs1047123208 Chr.7: 151213607-151213607 on Build GRCh38
rs148624345 Chr.7: 151213907-151213907 on Build GRCh38
rs557134991 Chr.7: 151213751-151213751 on Build GRCh38
rs2414 Chr.7: 151213556-151213556 on Build GRCh38
rs766036183 Chr.7: 151213695-151213695 on Build GRCh38
rs1022173497 Chr.7: 151213615-151213615 on Build GRCh38
rs1039855167 Chr.7: 151213795-151213795 on Build GRCh38
rs930204175 Chr.7: 151213490-151213490 on Build GRCh38
rs866670949 Chr.7: 151213672-151213672 on Build GRCh38
rs544876375 Chr.7: 151213833-151213833 on Build GRCh38
rs984348390 Chr.7: 151213699-151213699 on Build GRCh38
rs942729423 Chr.7: 151213650-151213650 on Build GRCh38
rs981784241 Chr.7: 151213843-151213843 on Build GRCh38
rs930162523 Chr.7: 151213632-151213632 on Build GRCh38
rs999782737 Chr.7: 151213708-151213708 on Build GRCh38
rs539557178 Chr.7: 151213781-151213781 on Build GRCh38
rs564088149 Chr.7: 151213715-151213715 on Build GRCh38
rs753312719 Chr.7: 151213665-151213665 on Build GRCh38
rs942865300 Chr.7: 151213521-151213521 on Build GRCh38
rs957759148 Chr.7: 151213539-151213539 on Build GRCh38
rs910013864 Chr.7: 151213892-151213892 on Build GRCh38
rs992886086 Chr.7: 151213920-151213920 on Build GRCh38
rs192541577 Chr.7: 151213783-151213783 on Build GRCh38
rs1030665801 Chr.7: 151213576-151213576 on Build GRCh38
rs552775130 Chr.7: 151213807-151213807 on Build GRCh38
rs1034849940 Chr.7: 151213906-151213906 on Build GRCh38
rs373554417 Chr.7: 151213649-151213649 on Build GRCh38
rs186621515 Chr.7: 151213489-151213489 on Build GRCh38
rs1025751272 Chr.7: 151213921-151213921 on Build GRCh38
rs769878110 Chr.7: 151213551-151213551 on Build GRCh38
rs1002567562 Chr.7: 151213899-151213899 on Build GRCh38
rs957789750 Chr.7: 151213740-151213740 on Build GRCh38
rs934008565 Chr.7: 151213937-151213937 on Build GRCh38
rs564841294 Chr.7: 151213861-151213861 on Build GRCh38

View Full Product Details