Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 151380936-151381444 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR86
Entrez Gene ID
349136
Gene Name
WD repeat domain 86
Gene Aliases
-
UniGene
Hs.647083
Gene Chromosome Location
Chr.7: 151377974-151410705 on Build GRCh38
Transcripts
NM_001284260.1 NM_001284261.1 NM_001284262.1 NM_198285.2 XM_005249989.3 XM_005249990.4 XM_011516144.2 XM_011516145.2 XM_011516146.2 XM_011516147.2 XM_011516148.1 XM_011516150.1 XM_011516151.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5595560 c.826C>T p.P276S Chr.7: 151381367-151381367 on Build GRCh38
6994413 c.815C>T p.P272L Chr.7: 151381378-151381378 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1038439864 Chr.7: 151381244-151381244 on Build GRCh38
rs2286418 Chr.7: 151381132-151381132 on Build GRCh38
rs926359168 Chr.7: 151381126-151381126 on Build GRCh38
rs934933579 Chr.7: 151381377-151381377 on Build GRCh38
rs979208627 Chr.7: 151381106-151381106 on Build GRCh38
rs750184938 Chr.7: 151381186-151381186 on Build GRCh38
rs999021860 Chr.7: 151381042-151381042 on Build GRCh38
rs751563083 Chr.7: 151381013-151381013 on Build GRCh38
rs551152986 Chr.7: 151381277-151381277 on Build GRCh38
rs115584628 Chr.7: 151381068-151381068 on Build GRCh38
rs1051319839 Chr.7: 151380964-151380964 on Build GRCh38
rs113513284 Chr.7: 151381382-151381382 on Build GRCh38
rs560737402 Chr.7: 151380960-151380960 on Build GRCh38
rs752742859 Chr.7: 151381408-151381408 on Build GRCh38
rs1033459214 Chr.7: 151381347-151381347 on Build GRCh38
rs746190846 Chr.7: 151380967-151380967 on Build GRCh38
rs905949634 Chr.7: 151381400-151381400 on Build GRCh38
rs1053308666 Chr.7: 151381044-151381044 on Build GRCh38
rs146159843 Chr.7: 151381267-151381267 on Build GRCh38
rs941895280 Chr.7: 151381415-151381415 on Build GRCh38
rs533432620 Chr.7: 151381390-151381390 on Build GRCh38
rs112828444 Chr.7: 151381416-151381416 on Build GRCh38
rs578123028 Chr.7: 151381321-151381321 on Build GRCh38
rs962842354 Chr.7: 151381230-151381230 on Build GRCh38
rs756349543 Chr.7: 151381250-151381250 on Build GRCh38
rs996814008 Chr.7: 151381272-151381272 on Build GRCh38
rs62490281 Chr.7: 151380985-151380985 on Build GRCh38
rs555478075 Chr.7: 151381372-151381372 on Build GRCh38
rs959082794 Chr.7: 151381352-151381352 on Build GRCh38
rs527461801 Chr.7: 151381146-151381146 on Build GRCh38
rs755066114 Chr.7: 151381301-151381301 on Build GRCh38
rs767425678 Chr.7: 151381177-151381177 on Build GRCh38
rs1021500960 Chr.7: 151381098-151381098 on Build GRCh38
rs941867064 Chr.7: 151381240-151381240 on Build GRCh38
rs917853306 Chr.7: 151381159-151381159 on Build GRCh38
rs960619714 Chr.7: 151381151-151381151 on Build GRCh38
rs2286417 Chr.7: 151381285-151381285 on Build GRCh38
rs776043337 Chr.7: 151381102-151381102 on Build GRCh38
rs546141021 Chr.7: 151381041-151381041 on Build GRCh38
rs773034957 Chr.7: 151381386-151381386 on Build GRCh38
rs955782120 Chr.7: 151381093-151381093 on Build GRCh38
rs968659691 Chr.7: 151381305-151381305 on Build GRCh38
rs547310783 Chr.7: 151381157-151381157 on Build GRCh38
rs535569719 Chr.7: 151381308-151381308 on Build GRCh38
rs77335389 Chr.7: 151381103-151381103 on Build GRCh38
rs34686318 Chr.7: 151381158-151381159 on Build GRCh38
rs929193229 Chr.7: 151381167-151381167 on Build GRCh38
rs529421651 Chr.7: 151381235-151381235 on Build GRCh38
rs983074948 Chr.7: 151381173-151381173 on Build GRCh38
rs1051020080 Chr.7: 151381276-151381276 on Build GRCh38
rs570371747 Chr.7: 151381249-151381249 on Build GRCh38
rs1002923458 Chr.7: 151381401-151381401 on Build GRCh38
rs1029896801 Chr.7: 151381084-151381084 on Build GRCh38
rs531883378 Chr.7: 151381094-151381094 on Build GRCh38
rs548384351 Chr.7: 151381121-151381121 on Build GRCh38
rs991843683 Chr.7: 151381358-151381358 on Build GRCh38
rs1044525833 Chr.7: 151381399-151381399 on Build GRCh38
rs1021197580 Chr.7: 151381299-151381299 on Build GRCh38
rs909130301 Chr.7: 151381412-151381412 on Build GRCh38
rs1047992050 Chr.7: 151380979-151380979 on Build GRCh38
rs558648916 Chr.7: 151381337-151381340 on Build GRCh38
rs887220186 Chr.7: 151380984-151380984 on Build GRCh38
rs774360904 Chr.7: 151381322-151381322 on Build GRCh38
rs59360596 Chr.7: 151381201-151381201 on Build GRCh38

View Full Product Details