Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 156681587-156682098 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LMBR1
Entrez Gene ID
64327
Gene Name
limb development membrane protein 1
Gene Aliases
ACHP, C7orf2, DIF14, LSS, PPD2, THYP, TPT, ZRS
UniGene
Hs.209989
Gene Chromosome Location
Chr.7: 156673354-156893208 on Build GRCh38
Transcripts
NM_022458.3 XM_005249555.2 XM_005249558.2 XM_017012507.1 XM_017012509.1 XM_017012514.1 XM_017012515.1 XM_017012517.1 XM_017012518.1

SNP Detail

SNP ID Chromosome Location
rs535905178 Chr.7: 156681646-156681646 on Build GRCh38
rs918492359 Chr.7: 156681817-156681817 on Build GRCh38
rs1031206410 Chr.7: 156681731-156681731 on Build GRCh38
rs1039240027 Chr.7: 156681859-156681859 on Build GRCh38
rs983385501 Chr.7: 156681654-156681654 on Build GRCh38
rs1001084120 Chr.7: 156681724-156681724 on Build GRCh38
rs116292915 Chr.7: 156681920-156681920 on Build GRCh38
rs1035084510 Chr.7: 156681659-156681659 on Build GRCh38
rs911647400 Chr.7: 156681816-156681816 on Build GRCh38
rs988483978 Chr.7: 156682060-156682060 on Build GRCh38
rs113867033 Chr.7: 156681997-156681997 on Build GRCh38
rs911144502 Chr.7: 156681615-156681615 on Build GRCh38
rs1056117291 Chr.7: 156681842-156681842 on Build GRCh38
rs573456996 Chr.7: 156681802-156681802 on Build GRCh38
rs529448700 Chr.7: 156682062-156682062 on Build GRCh38
rs548910349 Chr.7: 156682055-156682055 on Build GRCh38
rs1038448890 Chr.7: 156681641-156681641 on Build GRCh38
rs569266479 Chr.7: 156681740-156681740 on Build GRCh38
rs992681772 Chr.7: 156681667-156681667 on Build GRCh38
rs199726319 Chr.7: 156681981-156681981 on Build GRCh38
rs970281844 Chr.7: 156681998-156681998 on Build GRCh38
rs1023775800 Chr.7: 156681683-156681683 on Build GRCh38
rs976037528 Chr.7: 156681742-156681742 on Build GRCh38
rs573402696 Chr.7: 156681898-156681898 on Build GRCh38
rs985708342 Chr.7: 156681780-156681780 on Build GRCh38
rs934329042 Chr.7: 156681793-156681793 on Build GRCh38
rs17837686 Chr.7: 156681989-156681989 on Build GRCh38
rs929849277 Chr.7: 156682030-156682030 on Build GRCh38
rs904109452 Chr.7: 156681687-156681687 on Build GRCh38
rs557438593 Chr.7: 156681678-156681678 on Build GRCh38
rs560208746 Chr.7: 156682012-156682012 on Build GRCh38
rs961032830 Chr.7: 156681666-156681666 on Build GRCh38
rs373702651 Chr.7: 156681633-156681633 on Build GRCh38
rs116037533 Chr.7: 156681880-156681880 on Build GRCh38
rs146931391 Chr.7: 156682013-156682013 on Build GRCh38
rs953437380 Chr.7: 156681773-156681773 on Build GRCh38
rs970039026 Chr.7: 156681730-156681730 on Build GRCh38
rs776468876 Chr.7: 156681982-156681982 on Build GRCh38
rs540163305 Chr.7: 156681765-156681765 on Build GRCh38
rs941190756 Chr.7: 156681786-156681786 on Build GRCh38
rs761541056 Chr.7: 156681853-156681853 on Build GRCh38
rs367713862 Chr.7: 156681781-156681781 on Build GRCh38
rs371502278 Chr.7: 156681841-156681841 on Build GRCh38
rs540488086 Chr.7: 156681835-156681835 on Build GRCh38
rs918523318 Chr.7: 156682019-156682019 on Build GRCh38
rs1004841100 Chr.7: 156681974-156681974 on Build GRCh38
rs185766032 Chr.7: 156681990-156681990 on Build GRCh38
rs896217189 Chr.7: 156681677-156681677 on Build GRCh38
rs956734936 Chr.7: 156681738-156681738 on Build GRCh38
rs944420082 Chr.7: 156681852-156681852 on Build GRCh38
rs886667761 Chr.7: 156681971-156681971 on Build GRCh38
rs543898101 Chr.7: 156681899-156681899 on Build GRCh38
rs567467797 Chr.7: 156682058-156682058 on Build GRCh38
rs181517016 Chr.7: 156681626-156681626 on Build GRCh38
rs904059102 Chr.7: 156681886-156681886 on Build GRCh38
rs915721896 Chr.7: 156681728-156681728 on Build GRCh38
rs776143065 Chr.7: 156681662-156681662 on Build GRCh38
rs950703982 Chr.7: 156681638-156681638 on Build GRCh38
rs1034947639 Chr.7: 156681976-156681976 on Build GRCh38
rs892695469 Chr.7: 156681947-156681947 on Build GRCh38

View Full Product Details