Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 157174824-157175327 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBE3C
Entrez Gene ID
9690
Gene Name
ubiquitin protein ligase E3C
Gene Aliases
HECTH2
UniGene
Hs.118351
Gene Chromosome Location
Chr.7: 157138952-157269372 on Build GRCh38
Transcripts
NM_014671.2 XM_005249564.4 XM_017012818.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6022839 c.383_386delTTGT p.F128fs*16 Chr.7: 157174959-157174962 on Build GRCh38
4614047 c.380_383delTGTT p.F128fs*16 Chr.7: 157174956-157174959 on Build GRCh38
6367851 c.346T>C p.W116R Chr.7: 157174922-157174922 on Build GRCh38
485132 c.343A>G p.I115V Chr.7: 157174919-157174919 on Build GRCh38
3636746 c.379C>T p.L127L Chr.7: 157174955-157174955 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761050736 Chr.7: 157175212-157175212 on Build GRCh38
rs548988985 Chr.7: 157175295-157175295 on Build GRCh38
rs185553405 Chr.7: 157174871-157174871 on Build GRCh38
rs766260700 Chr.7: 157174883-157174883 on Build GRCh38
rs56852731 Chr.7: 157175158-157175159 on Build GRCh38
rs1003720929 Chr.7: 157174980-157174980 on Build GRCh38
rs778065398 Chr.7: 157175203-157175204 on Build GRCh38
rs1054034613 Chr.7: 157175143-157175143 on Build GRCh38
rs747148978 Chr.7: 157175080-157175080 on Build GRCh38
rs74965044 Chr.7: 157174908-157174908 on Build GRCh38
rs761066149 Chr.7: 157174856-157174856 on Build GRCh38
rs748339231 Chr.7: 157174875-157174875 on Build GRCh38
rs780509161 Chr.7: 157174947-157174947 on Build GRCh38
rs201353044 Chr.7: 157175046-157175046 on Build GRCh38
rs752883213 Chr.7: 157175079-157175079 on Build GRCh38
rs891494527 Chr.7: 157175150-157175150 on Build GRCh38
rs776509878 Chr.7: 157174993-157174993 on Build GRCh38
rs932372220 Chr.7: 157174895-157174895 on Build GRCh38
rs377043435 Chr.7: 157175023-157175023 on Build GRCh38
rs775012941 Chr.7: 157174876-157174876 on Build GRCh38
rs199504577 Chr.7: 157174982-157174982 on Build GRCh38
rs576218768 Chr.7: 157174969-157174969 on Build GRCh38
rs398048213 Chr.7: 157175139-157175140 on Build GRCh38
rs774923493 Chr.7: 157175012-157175012 on Build GRCh38
rs748251254 Chr.7: 157174965-157174965 on Build GRCh38
rs761578147 Chr.7: 157175049-157175049 on Build GRCh38
rs771218878 Chr.7: 157175081-157175081 on Build GRCh38
rs928238833 Chr.7: 157175298-157175298 on Build GRCh38
rs966662132 Chr.7: 157175138-157175141 on Build GRCh38
rs758521131 Chr.7: 157175074-157175074 on Build GRCh38
rs953849067 Chr.7: 157175230-157175230 on Build GRCh38
rs766610071 Chr.7: 157175056-157175056 on Build GRCh38
rs779544581 Chr.7: 157175069-157175069 on Build GRCh38
rs901646536 Chr.7: 157175185-157175185 on Build GRCh38
rs149473604 Chr.7: 157174970-157174970 on Build GRCh38
rs182171073 Chr.7: 157175061-157175061 on Build GRCh38
rs575632831 Chr.7: 157174967-157174967 on Build GRCh38
rs867944377 Chr.7: 157174923-157174923 on Build GRCh38
rs146568780 Chr.7: 157175179-157175179 on Build GRCh38
rs936272791 Chr.7: 157175141-157175141 on Build GRCh38
rs753458978 Chr.7: 157175258-157175258 on Build GRCh38
rs759565162 Chr.7: 157174919-157174919 on Build GRCh38
rs756431361 Chr.7: 157174933-157174933 on Build GRCh38
rs755270074 Chr.7: 157174912-157174912 on Build GRCh38
rs760370872 Chr.7: 157174882-157174882 on Build GRCh38
rs972779478 Chr.7: 157175271-157175271 on Build GRCh38
rs78558289 Chr.7: 157174861-157174861 on Build GRCh38
rs755571467 Chr.7: 157174950-157174950 on Build GRCh38
rs553328316 Chr.7: 157174953-157174953 on Build GRCh38
rs752238975 Chr.7: 157174924-157174924 on Build GRCh38
rs369411959 Chr.7: 157175159-157175159 on Build GRCh38
rs560850820 Chr.7: 157175187-157175187 on Build GRCh38
rs373648534 Chr.7: 157174870-157174870 on Build GRCh38
rs765977513 Chr.7: 157175220-157175220 on Build GRCh38
rs763664474 Chr.7: 157175031-157175031 on Build GRCh38
rs1055662114 Chr.7: 157174981-157174981 on Build GRCh38
rs998766430 Chr.7: 157175145-157175145 on Build GRCh38
rs759353206 Chr.7: 157174996-157174996 on Build GRCh38
rs150362446 Chr.7: 157174949-157174949 on Build GRCh38
rs370142937 Chr.7: 157174931-157174931 on Build GRCh38
rs1016976082 Chr.7: 157174929-157174929 on Build GRCh38
rs753322507 Chr.7: 157175073-157175073 on Build GRCh38
rs777956614 Chr.7: 157175076-157175076 on Build GRCh38
rs764680507 Chr.7: 157174921-157174921 on Build GRCh38
rs763754352 Chr.7: 157174928-157174928 on Build GRCh38
rs750730070 Chr.7: 157175060-157175060 on Build GRCh38
rs140273785 Chr.7: 157174925-157174925 on Build GRCh38
rs909589154 Chr.7: 157175275-157175275 on Build GRCh38
rs939660380 Chr.7: 157175287-157175287 on Build GRCh38
rs11771975 Chr.7: 157175201-157175201 on Build GRCh38
rs773082996 Chr.7: 157174879-157174879 on Build GRCh38
rs1016942155 Chr.7: 157175266-157175266 on Build GRCh38
rs527640343 Chr.7: 157175291-157175291 on Build GRCh38
rs533678511 Chr.7: 157175166-157175166 on Build GRCh38
rs945762705 Chr.7: 157175156-157175156 on Build GRCh38
rs770607677 Chr.7: 157174990-157174990 on Build GRCh38
rs961370762 Chr.7: 157175270-157175270 on Build GRCh38
rs867745928 Chr.7: 157175138-157175138 on Build GRCh38
rs769728213 Chr.7: 157175000-157175000 on Build GRCh38
rs776968248 Chr.7: 157174956-157174959 on Build GRCh38
rs137977548 Chr.7: 157174957-157174957 on Build GRCh38

View Full Product Details