Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 157183766-157184272 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBE3C
Entrez Gene ID
9690
Gene Name
ubiquitin protein ligase E3C
Gene Aliases
HECTH2
UniGene
Hs.118351
Gene Chromosome Location
Chr.7: 157138952-157269372 on Build GRCh38
Transcripts
NM_014671.2 XM_005249564.4 XM_017012818.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4612213 c.1061_1062delTC p.P356fs*11 Chr.7: 157183947-157183948 on Build GRCh38
3411865 c.1004A>T p.E335V Chr.7: 157183890-157183890 on Build GRCh38
4826296 c.1099G>A p.D367N Chr.7: 157183985-157183985 on Build GRCh38
745709 c.1009G>C p.G337R Chr.7: 157183895-157183895 on Build GRCh38
4456240 c.1001C>T p.S334F Chr.7: 157183887-157183887 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771312830 Chr.7: 157183851-157183852 on Build GRCh38
rs370796468 Chr.7: 157183935-157183935 on Build GRCh38
rs770697534 Chr.7: 157184012-157184012 on Build GRCh38
rs980535421 Chr.7: 157183963-157183963 on Build GRCh38
rs890541608 Chr.7: 157183875-157183875 on Build GRCh38
rs954416504 Chr.7: 157183835-157183835 on Build GRCh38
rs967858853 Chr.7: 157184200-157184200 on Build GRCh38
rs950931331 Chr.7: 157184105-157184105 on Build GRCh38
rs748427123 Chr.7: 157183906-157183906 on Build GRCh38
rs893633335 Chr.7: 157183809-157183809 on Build GRCh38
rs927337474 Chr.7: 157184163-157184163 on Build GRCh38
rs548533331 Chr.7: 157183793-157183793 on Build GRCh38
rs765339316 Chr.7: 157184058-157184058 on Build GRCh38
rs750028071 Chr.7: 157184158-157184158 on Build GRCh38
rs752326793 Chr.7: 157183942-157183942 on Build GRCh38
rs530197091 Chr.7: 157183790-157183790 on Build GRCh38
rs182681920 Chr.7: 157183844-157183844 on Build GRCh38
rs780696191 Chr.7: 157183967-157183967 on Build GRCh38
rs777371276 Chr.7: 157184005-157184005 on Build GRCh38
rs374198589 Chr.7: 157183962-157183962 on Build GRCh38
rs757024206 Chr.7: 157184073-157184073 on Build GRCh38
rs890232612 Chr.7: 157184051-157184052 on Build GRCh38
rs759519325 Chr.7: 157184023-157184023 on Build GRCh38
rs146755594 Chr.7: 157183961-157183961 on Build GRCh38
rs372529835 Chr.7: 157183866-157183866 on Build GRCh38
rs920807707 Chr.7: 157183927-157183927 on Build GRCh38
rs936018754 Chr.7: 157184174-157184174 on Build GRCh38
rs755635215 Chr.7: 157184171-157184171 on Build GRCh38
rs937961251 Chr.7: 157183990-157183990 on Build GRCh38
rs750846760 Chr.7: 157183854-157183854 on Build GRCh38
rs1027863920 Chr.7: 157184104-157184104 on Build GRCh38
rs965266443 Chr.7: 157183898-157183898 on Build GRCh38
rs779242914 Chr.7: 157183897-157183897 on Build GRCh38
rs772339443 Chr.7: 157183982-157183982 on Build GRCh38
rs923799948 Chr.7: 157184206-157184206 on Build GRCh38
rs779936054 Chr.7: 157183880-157183880 on Build GRCh38
rs1044052198 Chr.7: 157183794-157183794 on Build GRCh38
rs572476948 Chr.7: 157184182-157184182 on Build GRCh38
rs371072828 Chr.7: 157184038-157184038 on Build GRCh38
rs759537194 Chr.7: 157183817-157183817 on Build GRCh38
rs377393340 Chr.7: 157183973-157183973 on Build GRCh38
rs375495846 Chr.7: 157184080-157184080 on Build GRCh38
rs1005096762 Chr.7: 157184106-157184106 on Build GRCh38
rs1031710414 Chr.7: 157183863-157183863 on Build GRCh38
rs752414869 Chr.7: 157183840-157183840 on Build GRCh38
rs529008923 Chr.7: 157184227-157184227 on Build GRCh38
rs902451019 Chr.7: 157183799-157183799 on Build GRCh38
rs775013451 Chr.7: 157184028-157184028 on Build GRCh38
rs960378195 Chr.7: 157184129-157184129 on Build GRCh38
rs778052560 Chr.7: 157184068-157184068 on Build GRCh38
rs768917749 Chr.7: 157183883-157183883 on Build GRCh38
rs375850651 Chr.7: 157183913-157183913 on Build GRCh38
rs944503195 Chr.7: 157184235-157184235 on Build GRCh38
rs1024411608 Chr.7: 157184108-157184108 on Build GRCh38
rs944675755 Chr.7: 157184242-157184242 on Build GRCh38
rs369070971 Chr.7: 157183857-157183857 on Build GRCh38
rs951033281 Chr.7: 157183960-157183960 on Build GRCh38
rs564041011 Chr.7: 157184137-157184137 on Build GRCh38
rs558082258 Chr.7: 157184114-157184114 on Build GRCh38
rs753810727 Chr.7: 157184245-157184245 on Build GRCh38
rs746804527 Chr.7: 157184011-157184011 on Build GRCh38
rs370416956 Chr.7: 157183834-157183834 on Build GRCh38
rs767109840 Chr.7: 157184157-157184157 on Build GRCh38
rs375693521 Chr.7: 157184048-157184048 on Build GRCh38
rs201036596 Chr.7: 157184062-157184062 on Build GRCh38
rs553479269 Chr.7: 157183916-157183916 on Build GRCh38
rs781657429 Chr.7: 157183853-157183853 on Build GRCh38
rs762689579 Chr.7: 157183944-157183944 on Build GRCh38
rs372209828 Chr.7: 157184079-157184079 on Build GRCh38
rs776519809 Chr.7: 157184019-157184019 on Build GRCh38
rs754465613 Chr.7: 157183971-157183971 on Build GRCh38
rs79812505 Chr.7: 157183811-157183811 on Build GRCh38
rs759041063 Chr.7: 157183931-157183931 on Build GRCh38
rs535022918 Chr.7: 157183881-157183881 on Build GRCh38
rs570309027 Chr.7: 157183797-157183797 on Build GRCh38
rs927637942 Chr.7: 157183979-157183979 on Build GRCh38
rs758464378 Chr.7: 157184067-157184067 on Build GRCh38
rs201956089 Chr.7: 157183902-157183902 on Build GRCh38
rs754343076 Chr.7: 157184057-157184057 on Build GRCh38
rs202243973 Chr.7: 157184039-157184039 on Build GRCh38
rs145359477 Chr.7: 157184077-157184077 on Build GRCh38
rs779531494 Chr.7: 157184218-157184218 on Build GRCh38
rs766896010 Chr.7: 157184052-157184052 on Build GRCh38
rs12112208 Chr.7: 157184072-157184072 on Build GRCh38
rs769247187 Chr.7: 157184026-157184026 on Build GRCh38
rs1042231507 Chr.7: 157184050-157184050 on Build GRCh38
rs539858129 Chr.7: 157184205-157184205 on Build GRCh38
rs755083822 Chr.7: 157183972-157183972 on Build GRCh38
rs777120853 Chr.7: 157184052-157184052 on Build GRCh38
rs756678763 Chr.7: 157183879-157183879 on Build GRCh38
rs776605724 Chr.7: 157183917-157183917 on Build GRCh38
rs374920083 Chr.7: 157184076-157184076 on Build GRCh38
rs771711166 Chr.7: 157183909-157183909 on Build GRCh38
rs767735059 Chr.7: 157183948-157183948 on Build GRCh38
rs561429220 Chr.7: 157184222-157184222 on Build GRCh38
rs890283603 Chr.7: 157184217-157184217 on Build GRCh38
rs1002110244 Chr.7: 157183794-157183795 on Build GRCh38
rs912506089 Chr.7: 157184228-157184228 on Build GRCh38
rs754564797 Chr.7: 157183830-157183830 on Build GRCh38
rs763824304 Chr.7: 157184047-157184047 on Build GRCh38
rs753349610 Chr.7: 157183829-157183829 on Build GRCh38
rs868644837 Chr.7: 157183816-157183816 on Build GRCh38

View Full Product Details