Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 157568822-157569317 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPRN2
Entrez Gene ID
5799
Gene Name
protein tyrosine phosphatase, receptor type N2
Gene Aliases
IA-2beta, IAR, ICAAR, PTPRP, R-PTP-N2
UniGene
Hs.490789
Gene Chromosome Location
Chr.7: 157539052-158587802 on Build GRCh38
Transcripts
NM_001308267.1 NM_001308268.1 NM_002847.4 NM_130842.3 NM_130843.3 XM_017012475.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1647993 c.2922G>A p.R974R Chr.7: 157568951-157568951 on Build GRCh38
745701 c.2853G>A p.R951R Chr.7: 157568951-157568951 on Build GRCh38
7111931 c.2922G>C p.R974R Chr.7: 157568951-157568951 on Build GRCh38
1647992 c.2921G>A p.R974Q Chr.7: 157568952-157568952 on Build GRCh38
745699 c.2852G>A p.R951Q Chr.7: 157568952-157568952 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775364404 Chr.7: 157568945-157568945 on Build GRCh38
rs973545255 Chr.7: 157569080-157569080 on Build GRCh38
rs750453472 Chr.7: 157568935-157568935 on Build GRCh38
rs989391618 Chr.7: 157568983-157568983 on Build GRCh38
rs552998917 Chr.7: 157569041-157569041 on Build GRCh38
rs757598649 Chr.7: 157568850-157568850 on Build GRCh38
rs79679565 Chr.7: 157569229-157569229 on Build GRCh38
rs1056411223 Chr.7: 157569236-157569236 on Build GRCh38
rs999348625 Chr.7: 157569167-157569167 on Build GRCh38
rs572885113 Chr.7: 157569284-157569284 on Build GRCh38
rs762274961 Chr.7: 157568910-157568910 on Build GRCh38
rs894999102 Chr.7: 157568926-157568926 on Build GRCh38
rs532180970 Chr.7: 157568954-157568954 on Build GRCh38
rs946575092 Chr.7: 157569143-157569143 on Build GRCh38
rs894969639 Chr.7: 157569130-157569130 on Build GRCh38
rs867851806 Chr.7: 157568952-157568952 on Build GRCh38
rs780384663 Chr.7: 157568854-157568854 on Build GRCh38
rs763261513 Chr.7: 157568908-157568908 on Build GRCh38
rs112570350 Chr.7: 157569060-157569061 on Build GRCh38
rs762163628 Chr.7: 157569070-157569070 on Build GRCh38
rs556736303 Chr.7: 157568895-157568895 on Build GRCh38
rs201741114 Chr.7: 157568875-157568875 on Build GRCh38
rs760300360 Chr.7: 157568887-157568887 on Build GRCh38
rs1031648602 Chr.7: 157569168-157569168 on Build GRCh38
rs34992438 Chr.7: 157569112-157569113 on Build GRCh38
rs910990096 Chr.7: 157569199-157569199 on Build GRCh38
rs983103444 Chr.7: 157569097-157569097 on Build GRCh38
rs985147773 Chr.7: 157569195-157569195 on Build GRCh38
rs890474317 Chr.7: 157569189-157569189 on Build GRCh38
rs146088797 Chr.7: 157569231-157569231 on Build GRCh38
rs535280952 Chr.7: 157569075-157569075 on Build GRCh38
rs922806348 Chr.7: 157568996-157568996 on Build GRCh38
rs79866755 Chr.7: 157569232-157569232 on Build GRCh38
rs762918278 Chr.7: 157568897-157568898 on Build GRCh38
rs922757014 Chr.7: 157569148-157569148 on Build GRCh38
rs144237368 Chr.7: 157569008-157569008 on Build GRCh38
rs768578494 Chr.7: 157568873-157568873 on Build GRCh38
rs144485708 Chr.7: 157568933-157568933 on Build GRCh38
rs534567674 Chr.7: 157569009-157569009 on Build GRCh38
rs770391264 Chr.7: 157568882-157568882 on Build GRCh38
rs1043531689 Chr.7: 157569149-157569149 on Build GRCh38
rs774691690 Chr.7: 157569060-157569060 on Build GRCh38
rs7789467 Chr.7: 157568872-157568872 on Build GRCh38
rs368578089 Chr.7: 157568982-157568982 on Build GRCh38
rs993983313 Chr.7: 157569044-157569044 on Build GRCh38
rs956480909 Chr.7: 157569112-157569112 on Build GRCh38
rs776269736 Chr.7: 157568900-157568900 on Build GRCh38
rs896342720 Chr.7: 157569062-157569062 on Build GRCh38
rs973632971 Chr.7: 157569283-157569283 on Build GRCh38
rs372046530 Chr.7: 157569016-157569016 on Build GRCh38
rs970021101 Chr.7: 157569084-157569084 on Build GRCh38
rs939245403 Chr.7: 157569108-157569108 on Build GRCh38
rs372883822 Chr.7: 157569000-157569000 on Build GRCh38
rs760634608 Chr.7: 157569017-157569017 on Build GRCh38
rs1054977535 Chr.7: 157569111-157569111 on Build GRCh38
rs778830037 Chr.7: 157568964-157568964 on Build GRCh38
rs221294 Chr.7: 157569042-157569042 on Build GRCh38
rs756281977 Chr.7: 157568851-157568851 on Build GRCh38
rs371671584 Chr.7: 157568846-157568846 on Build GRCh38
rs964906449 Chr.7: 157569048-157569048 on Build GRCh38
rs920397857 Chr.7: 157569086-157569086 on Build GRCh38
rs756234746 Chr.7: 157568939-157568939 on Build GRCh38
rs1038269798 Chr.7: 157569030-157569030 on Build GRCh38
rs767413265 Chr.7: 157569099-157569099 on Build GRCh38
rs754774155 Chr.7: 157568958-157568958 on Build GRCh38
rs904573239 Chr.7: 157569157-157569157 on Build GRCh38
rs964873331 Chr.7: 157569235-157569235 on Build GRCh38
rs747997930 Chr.7: 157568971-157568971 on Build GRCh38
rs769473869 Chr.7: 157569003-157569003 on Build GRCh38
rs556831993 Chr.7: 157569250-157569250 on Build GRCh38
rs548805667 Chr.7: 157568886-157568886 on Build GRCh38
rs764464048 Chr.7: 157568909-157568909 on Build GRCh38
rs113115279 Chr.7: 157569095-157569095 on Build GRCh38
rs940999516 Chr.7: 157569224-157569224 on Build GRCh38
rs55645555 Chr.7: 157569001-157569001 on Build GRCh38
rs1044458928 Chr.7: 157569285-157569285 on Build GRCh38
rs541664316 Chr.7: 157569293-157569293 on Build GRCh38
rs374336693 Chr.7: 157568881-157568881 on Build GRCh38
rs563011675 Chr.7: 157568947-157568947 on Build GRCh38

View Full Product Details