Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161189369-161189895 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS4
Entrez Gene ID
9507
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 4
Gene Aliases
ADAMTS-2, ADAMTS-4, ADMP-1
UniGene
Hs.211604
Gene Chromosome Location
Chr.1: 161189725-161199080 on Build GRCh38
Transcripts
NM_001320336.1 NM_005099.5

SNP Detail

SNP ID Chromosome Location
rs36123104 Chr.1: 161189645-161189645 on Build GRCh38
rs1048241100 Chr.1: 161189537-161189537 on Build GRCh38
rs1052499825 Chr.1: 161189672-161189672 on Build GRCh38
rs753673141 Chr.1: 161189709-161189709 on Build GRCh38
rs368885506 Chr.1: 161189709-161189712 on Build GRCh38
rs1051624437 Chr.1: 161189856-161189856 on Build GRCh38
rs773888260 Chr.1: 161189615-161189615 on Build GRCh38
rs909840945 Chr.1: 161189569-161189569 on Build GRCh38
rs969924085 Chr.1: 161189738-161189738 on Build GRCh38
rs752235990 Chr.1: 161189666-161189666 on Build GRCh38
rs763866279 Chr.1: 161189673-161189673 on Build GRCh38
rs756216475 Chr.1: 161189600-161189600 on Build GRCh38
rs963072074 Chr.1: 161189507-161189507 on Build GRCh38
rs866093490 Chr.1: 161189497-161189497 on Build GRCh38
rs892531279 Chr.1: 161189683-161189683 on Build GRCh38
rs567007345 Chr.1: 161189558-161189558 on Build GRCh38
rs1030102100 Chr.1: 161189459-161189459 on Build GRCh38
rs1002460307 Chr.1: 161189786-161189786 on Build GRCh38
rs778964965 Chr.1: 161189772-161189772 on Build GRCh38
rs144519251 Chr.1: 161189621-161189621 on Build GRCh38
rs757314087 Chr.1: 161189771-161189771 on Build GRCh38
rs147638828 Chr.1: 161189484-161189484 on Build GRCh38
rs929824850 Chr.1: 161189505-161189505 on Build GRCh38
rs938127124 Chr.1: 161189629-161189629 on Build GRCh38
rs997413869 Chr.1: 161189807-161189807 on Build GRCh38
rs970882342 Chr.1: 161189587-161189587 on Build GRCh38
rs185423435 Chr.1: 161189714-161189714 on Build GRCh38
rs755945929 Chr.1: 161189671-161189671 on Build GRCh38
rs760184071 Chr.1: 161189562-161189562 on Build GRCh38
rs953129274 Chr.1: 161189475-161189475 on Build GRCh38
rs765706620 Chr.1: 161189611-161189611 on Build GRCh38
rs1041634741 Chr.1: 161189430-161189430 on Build GRCh38
rs777049064 Chr.1: 161189462-161189462 on Build GRCh38
rs539013024 Chr.1: 161189586-161189586 on Build GRCh38
rs550710627 Chr.1: 161189441-161189441 on Build GRCh38
rs1035660021 Chr.1: 161189799-161189799 on Build GRCh38
rs762361159 Chr.1: 161189478-161189478 on Build GRCh38
rs1016376310 Chr.1: 161189501-161189501 on Build GRCh38
rs761150966 Chr.1: 161189422-161189422 on Build GRCh38
rs145543139 Chr.1: 161189862-161189862 on Build GRCh38
rs942625635 Chr.1: 161189588-161189588 on Build GRCh38
rs764922472 Chr.1: 161189719-161189719 on Build GRCh38
rs919482987 Chr.1: 161189582-161189582 on Build GRCh38
rs997318808 Chr.1: 161189438-161189440 on Build GRCh38
rs142313268 Chr.1: 161189498-161189498 on Build GRCh38
rs908306766 Chr.1: 161189432-161189444 on Build GRCh38
rs547132313 Chr.1: 161189524-161189524 on Build GRCh38

View Full Product Details