Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161191715-161192257 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS4
Entrez Gene ID
9507
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 4
Gene Aliases
ADAMTS-2, ADAMTS-4, ADMP-1
UniGene
Hs.211604
Gene Chromosome Location
Chr.1: 161189725-161199080 on Build GRCh38
Transcripts
NM_001320336.1 NM_005099.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1335643 c.2057G>T p.S686I Chr.1: 161192095-161192095 on Build GRCh38
4576482 c.2026A>T p.M676L Chr.1: 161192126-161192126 on Build GRCh38
5083944 c.1991G>A p.R664H Chr.1: 161192161-161192161 on Build GRCh38
6594231 c.2084T>G p.F695C Chr.1: 161192068-161192068 on Build GRCh38
2086897 c.1970G>A p.C657Y Chr.1: 161192182-161192182 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs987651324 Chr.1: 161191873-161191873 on Build GRCh38
rs3134667 Chr.1: 161192108-161192108 on Build GRCh38
rs370229251 Chr.1: 161192061-161192061 on Build GRCh38
rs370623177 Chr.1: 161192156-161192156 on Build GRCh38
rs774609955 Chr.1: 161192188-161192188 on Build GRCh38
rs749255932 Chr.1: 161192133-161192133 on Build GRCh38
rs1016379116 Chr.1: 161192053-161192053 on Build GRCh38
rs1024872529 Chr.1: 161191989-161191989 on Build GRCh38
rs550067062 Chr.1: 161192118-161192118 on Build GRCh38
rs761703641 Chr.1: 161191824-161191824 on Build GRCh38
rs1004871938 Chr.1: 161192030-161192030 on Build GRCh38
rs763301244 Chr.1: 161192085-161192085 on Build GRCh38
rs113252551 Chr.1: 161191738-161191738 on Build GRCh38
rs773785728 Chr.1: 161192040-161192040 on Build GRCh38
rs755296367 Chr.1: 161192112-161192112 on Build GRCh38
rs762843733 Chr.1: 161192103-161192103 on Build GRCh38
rs116361265 Chr.1: 161192143-161192143 on Build GRCh38
rs376630100 Chr.1: 161192126-161192126 on Build GRCh38
rs752980389 Chr.1: 161192209-161192209 on Build GRCh38
rs537518142 Chr.1: 161192223-161192223 on Build GRCh38
rs963493158 Chr.1: 161192078-161192078 on Build GRCh38
rs747949266 Chr.1: 161192019-161192019 on Build GRCh38
rs756226957 Chr.1: 161192229-161192229 on Build GRCh38
rs750229637 Chr.1: 161192043-161192043 on Build GRCh38
rs925057053 Chr.1: 161192215-161192215 on Build GRCh38
rs903590530 Chr.1: 161191912-161191912 on Build GRCh38
rs1026075974 Chr.1: 161191739-161191739 on Build GRCh38
rs936539466 Chr.1: 161191925-161191925 on Build GRCh38
rs561900481 Chr.1: 161191761-161191761 on Build GRCh38
rs770443663 Chr.1: 161192031-161192031 on Build GRCh38
rs1023445293 Chr.1: 161191886-161191887 on Build GRCh38
rs374646080 Chr.1: 161192214-161192214 on Build GRCh38
rs143096816 Chr.1: 161192109-161192109 on Build GRCh38
rs1013511392 Chr.1: 161191958-161191958 on Build GRCh38
rs778419465 Chr.1: 161192146-161192146 on Build GRCh38
rs977710745 Chr.1: 161192206-161192206 on Build GRCh38
rs987083853 Chr.1: 161192008-161192008 on Build GRCh38
rs898632745 Chr.1: 161192136-161192136 on Build GRCh38
rs750952231 Chr.1: 161192080-161192080 on Build GRCh38
rs1042133863 Chr.1: 161191908-161191908 on Build GRCh38
rs759219047 Chr.1: 161192055-161192055 on Build GRCh38
rs374106325 Chr.1: 161192068-161192068 on Build GRCh38
rs746584852 Chr.1: 161192160-161192160 on Build GRCh38
rs1034893655 Chr.1: 161191788-161191788 on Build GRCh38
rs1009380614 Chr.1: 161192057-161192057 on Build GRCh38
rs777765730 Chr.1: 161192027-161192027 on Build GRCh38
rs35395819 Chr.1: 161192227-161192228 on Build GRCh38
rs1004798443 Chr.1: 161191764-161191764 on Build GRCh38
rs765190124 Chr.1: 161191946-161191946 on Build GRCh38
rs748808947 Chr.1: 161192028-161192028 on Build GRCh38
rs912233767 Chr.1: 161191880-161191880 on Build GRCh38
rs527662733 Chr.1: 161191769-161191769 on Build GRCh38
rs751618429 Chr.1: 161192161-161192161 on Build GRCh38
rs936440440 Chr.1: 161192224-161192224 on Build GRCh38
rs745360883 Chr.1: 161192155-161192155 on Build GRCh38
rs759687491 Chr.1: 161192202-161192202 on Build GRCh38
rs945020622 Chr.1: 161191907-161191907 on Build GRCh38
rs757148503 Chr.1: 161192139-161192139 on Build GRCh38
rs933581135 Chr.1: 161191860-161191860 on Build GRCh38
rs771753545 Chr.1: 161192159-161192159 on Build GRCh38
rs370892564 Chr.1: 161192185-161192185 on Build GRCh38
rs887071655 Chr.1: 161191743-161191743 on Build GRCh38
rs570236275 Chr.1: 161192162-161192162 on Build GRCh38
rs751036482 Chr.1: 161192009-161192009 on Build GRCh38
rs777375359 Chr.1: 161192130-161192130 on Build GRCh38
rs767912629 Chr.1: 161192203-161192203 on Build GRCh38
rs766965749 Chr.1: 161192186-161192186 on Build GRCh38
rs767813779 Chr.1: 161192117-161192117 on Build GRCh38
rs116393628 Chr.1: 161191947-161191947 on Build GRCh38
rs113500169 Chr.1: 161192020-161192020 on Build GRCh38

View Full Product Details