Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161526670-161527215 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPA6
Entrez Gene ID
3310
Gene Name
heat shock protein family A (Hsp70) member 6
Gene Aliases
HSP70B'
UniGene
Hs.654614
Gene Chromosome Location
Chr.1: 161524540-161526897 on Build GRCh38
Transcripts
NM_002155.4

SNP Detail

SNP ID Chromosome Location
rs4656310 Chr.1: 161527110-161527110 on Build GRCh38
rs375069063 Chr.1: 161526801-161526801 on Build GRCh38
rs12128362 Chr.1: 161526852-161526852 on Build GRCh38
rs866607150 Chr.1: 161526791-161526791 on Build GRCh38
rs966347992 Chr.1: 161526835-161526835 on Build GRCh38
rs539324859 Chr.1: 161527146-161527146 on Build GRCh38
rs41299264 Chr.1: 161526741-161526741 on Build GRCh38
rs1028336827 Chr.1: 161527077-161527077 on Build GRCh38
rs984174726 Chr.1: 161526908-161526908 on Build GRCh38
rs950345076 Chr.1: 161527152-161527152 on Build GRCh38
rs984206087 Chr.1: 161527118-161527118 on Build GRCh38
rs1037250594 Chr.1: 161526705-161526705 on Build GRCh38
rs10919261 Chr.1: 161526840-161526840 on Build GRCh38
rs41299262 Chr.1: 161526739-161526739 on Build GRCh38
rs536805666 Chr.1: 161526948-161526948 on Build GRCh38
rs41299268 Chr.1: 161527054-161527054 on Build GRCh38
rs182070643 Chr.1: 161526983-161526983 on Build GRCh38
rs879944480 Chr.1: 161527108-161527108 on Build GRCh38
rs953992578 Chr.1: 161527087-161527087 on Build GRCh38
rs1042881 Chr.1: 161526740-161526740 on Build GRCh38
rs891545153 Chr.1: 161526996-161526996 on Build GRCh38
rs976808057 Chr.1: 161526886-161526886 on Build GRCh38
rs573118758 Chr.1: 161527123-161527123 on Build GRCh38
rs12120133 Chr.1: 161526898-161526898 on Build GRCh38
rs932740157 Chr.1: 161526899-161526899 on Build GRCh38
rs1037601343 Chr.1: 161526933-161526933 on Build GRCh38
rs41299266 Chr.1: 161527015-161527015 on Build GRCh38
rs1019180637 Chr.1: 161527016-161527016 on Build GRCh38
rs10919266 Chr.1: 161526953-161526953 on Build GRCh38
rs1018902637 Chr.1: 161526818-161526818 on Build GRCh38
rs59491199 Chr.1: 161527135-161527135 on Build GRCh38
rs41299270 Chr.1: 161527066-161527066 on Build GRCh38
rs186603139 Chr.1: 161527172-161527172 on Build GRCh38
rs955839684 Chr.1: 161526780-161526780 on Build GRCh38
rs2499431 Chr.1: 161527028-161527028 on Build GRCh38
rs12145551 Chr.1: 161526918-161526918 on Build GRCh38
rs10919265 Chr.1: 161526952-161526952 on Build GRCh38
rs896101711 Chr.1: 161526943-161526943 on Build GRCh38
rs961443364 Chr.1: 161527125-161527125 on Build GRCh38
rs565829922 Chr.1: 161526966-161526966 on Build GRCh38
rs910057814 Chr.1: 161526911-161526913 on Build GRCh38
rs940129261 Chr.1: 161526932-161526932 on Build GRCh38
rs1032822646 Chr.1: 161526750-161526750 on Build GRCh38
rs548995950 Chr.1: 161526703-161526703 on Build GRCh38
rs972952277 Chr.1: 161527131-161527131 on Build GRCh38
rs12073773 Chr.1: 161526709-161526709 on Build GRCh38
rs1013958839 Chr.1: 161526732-161526732 on Build GRCh38
rs917383445 Chr.1: 161527137-161527137 on Build GRCh38
rs999881047 Chr.1: 161526743-161526743 on Build GRCh38
rs779399128 Chr.1: 161526963-161526963 on Build GRCh38
rs10919264 Chr.1: 161526937-161526937 on Build GRCh38

View Full Product Details