Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 76681143-76681690 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFHX4
Entrez Gene ID
79776
Gene Name
zinc finger homeobox 4
Gene Aliases
ZFH4, ZHF4
UniGene
Hs.458973
Gene Chromosome Location
Chr.8: 76681219-76867285 on Build GRCh38
Transcripts
NM_024721.4 XM_011517596.2 XM_011517597.2 XM_017013845.1 NR_024360.1

Gene Symbol
ZFHX4-AS1
Entrez Gene ID
100192378
Gene Name
ZFHX4 antisense RNA 1
Gene Aliases
-
UniGene
Hs.596420
Gene Chromosome Location
Chr.8: 76610879-76683275 on Build GRCh38
Transcripts
NM_024721.4 XM_011517596.2 XM_011517597.2 XM_017013845.1 NR_024360.1

SNP Detail

SNP ID Chromosome Location
rs980512483 Chr.8: 76681594-76681594 on Build GRCh38
rs747366041 Chr.8: 76681180-76681181 on Build GRCh38
rs904814076 Chr.8: 76681483-76681483 on Build GRCh38
rs144397783 Chr.8: 76681527-76681527 on Build GRCh38
rs775686485 Chr.8: 76681302-76681302 on Build GRCh38
rs1031842557 Chr.8: 76681185-76681185 on Build GRCh38
rs934170250 Chr.8: 76681257-76681257 on Build GRCh38
rs975748118 Chr.8: 76681457-76681457 on Build GRCh38
rs1051969299 Chr.8: 76681416-76681416 on Build GRCh38
rs768490673 Chr.8: 76681385-76681386 on Build GRCh38
rs907116546 Chr.8: 76681609-76681609 on Build GRCh38
rs554205459 Chr.8: 76681530-76681530 on Build GRCh38
rs911387319 Chr.8: 76681507-76681507 on Build GRCh38
rs879781511 Chr.8: 76681538-76681538 on Build GRCh38
rs1045699909 Chr.8: 76681579-76681579 on Build GRCh38
rs970479874 Chr.8: 76681364-76681364 on Build GRCh38
rs569736041 Chr.8: 76681286-76681286 on Build GRCh38
rs866463404 Chr.8: 76681473-76681473 on Build GRCh38
rs774296490 Chr.8: 76681510-76681511 on Build GRCh38
rs187326595 Chr.8: 76681322-76681322 on Build GRCh38
rs1049962688 Chr.8: 76681502-76681502 on Build GRCh38
rs551594136 Chr.8: 76681193-76681193 on Build GRCh38
rs766491823 Chr.8: 76681390-76681390 on Build GRCh38
rs1018287753 Chr.8: 76681176-76681176 on Build GRCh38
rs889878974 Chr.8: 76681272-76681272 on Build GRCh38
rs1007647186 Chr.8: 76681425-76681425 on Build GRCh38
rs1031870293 Chr.8: 76681500-76681500 on Build GRCh38
rs917277948 Chr.8: 76681315-76681315 on Build GRCh38
rs1037674679 Chr.8: 76681233-76681233 on Build GRCh38
rs956172109 Chr.8: 76681501-76681501 on Build GRCh38
rs751529598 Chr.8: 76681487-76681487 on Build GRCh38
rs1049890923 Chr.8: 76681271-76681271 on Build GRCh38
rs963297902 Chr.8: 76681616-76681616 on Build GRCh38
rs750723411 Chr.8: 76681201-76681201 on Build GRCh38
rs929082361 Chr.8: 76681400-76681400 on Build GRCh38
rs909881400 Chr.8: 76681611-76681611 on Build GRCh38
rs1034180193 Chr.8: 76681651-76681651 on Build GRCh38
rs754939944 Chr.8: 76681519-76681519 on Build GRCh38
rs968964188 Chr.8: 76681183-76681183 on Build GRCh38
rs926540359 Chr.8: 76681376-76681376 on Build GRCh38
rs537381672 Chr.8: 76681325-76681325 on Build GRCh38
rs1000280016 Chr.8: 76681280-76681281 on Build GRCh38
rs970132215 Chr.8: 76681551-76681551 on Build GRCh38
rs532883858 Chr.8: 76681184-76681184 on Build GRCh38
rs980095938 Chr.8: 76681372-76681372 on Build GRCh38
rs761436960 Chr.8: 76681580-76681582 on Build GRCh38
rs946051890 Chr.8: 76681574-76681574 on Build GRCh38
rs143306746 Chr.8: 76681445-76681446 on Build GRCh38
rs1038660449 Chr.8: 76681445-76681445 on Build GRCh38
rs564713856 Chr.8: 76681501-76681502 on Build GRCh38
rs555665238 Chr.8: 76681350-76681350 on Build GRCh38
rs763026742 Chr.8: 76681374-76681374 on Build GRCh38
rs922816461 Chr.8: 76681249-76681249 on Build GRCh38
rs890185107 Chr.8: 76681420-76681420 on Build GRCh38
rs368102806 Chr.8: 76681391-76681391 on Build GRCh38
rs568472321 Chr.8: 76681381-76681381 on Build GRCh38
rs1010178165 Chr.8: 76681375-76681375 on Build GRCh38
rs973275733 Chr.8: 76681646-76681646 on Build GRCh38

View Full Product Details