Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 80050358-80050868 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TPD52
Entrez Gene ID
7163
Gene Name
tumor protein D52
Gene Aliases
D52, N8L, PC-1, PrLZ, hD52
UniGene
Hs.368433
Gene Chromosome Location
Chr.8: 80034868-80171659 on Build GRCh38
Transcripts
NM_001025252.2 NM_001025253.2 NM_001287140.1 NM_001287142.1 NM_001287143.1 NM_001287144.1 NM_005079.3 NR_105033.1 NR_105034.1 NR_105035.1 NR_105036.1 NR_105037.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4492987 c.407C>T p.S136F Chr.8: 80050451-80050451 on Build GRCh38
4450004 c.413+1G>A p.? Chr.8: 80050444-80050444 on Build GRCh38
5776962 c.387-1G>C p.? Chr.8: 80050472-80050472 on Build GRCh38
6432855 c.413G>T p.S138I Chr.8: 80050445-80050445 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs376245887 Chr.8: 80050500-80050500 on Build GRCh38
rs917683945 Chr.8: 80050579-80050579 on Build GRCh38
rs780144022 Chr.8: 80050501-80050501 on Build GRCh38
rs753996822 Chr.8: 80050462-80050462 on Build GRCh38
rs923106215 Chr.8: 80050777-80050777 on Build GRCh38
rs774113084 Chr.8: 80050395-80050395 on Build GRCh38
rs940483841 Chr.8: 80050700-80050700 on Build GRCh38
rs972568778 Chr.8: 80050609-80050609 on Build GRCh38
rs907624946 Chr.8: 80050659-80050659 on Build GRCh38
rs1010402429 Chr.8: 80050699-80050699 on Build GRCh38
rs917905819 Chr.8: 80050732-80050747 on Build GRCh38
rs534791978 Chr.8: 80050809-80050809 on Build GRCh38
rs774733381 Chr.8: 80050415-80050415 on Build GRCh38
rs1041134265 Chr.8: 80050682-80050682 on Build GRCh38
rs536378758 Chr.8: 80050400-80050400 on Build GRCh38
rs931889794 Chr.8: 80050778-80050778 on Build GRCh38
rs771297422 Chr.8: 80050690-80050690 on Build GRCh38
rs1050752536 Chr.8: 80050784-80050784 on Build GRCh38
rs984453460 Chr.8: 80050640-80050640 on Build GRCh38
rs951698771 Chr.8: 80050637-80050637 on Build GRCh38
rs764324039 Chr.8: 80050439-80050439 on Build GRCh38
rs944057880 Chr.8: 80050635-80050635 on Build GRCh38
rs931283011 Chr.8: 80050623-80050623 on Build GRCh38
rs548952496 Chr.8: 80050742-80050742 on Build GRCh38
rs1024665492 Chr.8: 80050822-80050822 on Build GRCh38
rs916227405 Chr.8: 80050584-80050584 on Build GRCh38
rs1035599789 Chr.8: 80050543-80050543 on Build GRCh38
rs959306218 Chr.8: 80050528-80050528 on Build GRCh38
rs961144702 Chr.8: 80050834-80050835 on Build GRCh38
rs751054048 Chr.8: 80050506-80050506 on Build GRCh38
rs893080590 Chr.8: 80050815-80050815 on Build GRCh38
rs991490121 Chr.8: 80050559-80050559 on Build GRCh38
rs991893537 Chr.8: 80050555-80050555 on Build GRCh38
rs780821596 Chr.8: 80050510-80050510 on Build GRCh38
rs771191997 Chr.8: 80050406-80050406 on Build GRCh38
rs760011806 Chr.8: 80050417-80050417 on Build GRCh38
rs960951464 Chr.8: 80050556-80050556 on Build GRCh38
rs919834078 Chr.8: 80050620-80050620 on Build GRCh38
rs766921839 Chr.8: 80050622-80050622 on Build GRCh38
rs764910671 Chr.8: 80050471-80050471 on Build GRCh38
rs891915539 Chr.8: 80050688-80050688 on Build GRCh38
rs959046062 Chr.8: 80050752-80050752 on Build GRCh38
rs969032725 Chr.8: 80050474-80050475 on Build GRCh38
rs1040169079 Chr.8: 80050732-80050732 on Build GRCh38
rs182667421 Chr.8: 80050751-80050751 on Build GRCh38
rs749571522 Chr.8: 80050600-80050600 on Build GRCh38
rs551416098 Chr.8: 80050843-80050843 on Build GRCh38
rs775510617 Chr.8: 80050426-80050426 on Build GRCh38
rs1035028406 Chr.8: 80050750-80050750 on Build GRCh38
rs777679253 Chr.8: 80050474-80050474 on Build GRCh38
rs999689762 Chr.8: 80050508-80050508 on Build GRCh38
rs772586616 Chr.8: 80050422-80050422 on Build GRCh38
rs529169613 Chr.8: 80050561-80050561 on Build GRCh38
rs570084651 Chr.8: 80050404-80050404 on Build GRCh38
rs1050108494 Chr.8: 80050625-80050625 on Build GRCh38
rs560192051 Chr.8: 80050835-80050835 on Build GRCh38
rs77756924 Chr.8: 80050428-80050428 on Build GRCh38
rs762162339 Chr.8: 80050467-80050467 on Build GRCh38
rs1010904294 Chr.8: 80050824-80050824 on Build GRCh38
rs1001760053 Chr.8: 80050734-80050734 on Build GRCh38
rs754432386 Chr.8: 80050509-80050509 on Build GRCh38

View Full Product Details