Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 169376425-169376950 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BLZF1
Entrez Gene ID
8548
Gene Name
basic leucine zipper nuclear factor 1
Gene Aliases
GOLGIN-45, JEM-1, JEM-1s, JEM1
UniGene
Hs.130746
Gene Chromosome Location
Chr.1: 169367879-169396544 on Build GRCh38
Transcripts
NM_001320972.1 NM_001320973.1 NM_003666.3 XM_005245561.4 XM_011510092.2 XM_017002645.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7130597 c.174G>C p.E58D Chr.1: 169376685-169376685 on Build GRCh38
6611820 c.112G>A p.G38R Chr.1: 169376623-169376623 on Build GRCh38
2073411 c.300G>T p.K100N Chr.1: 169376811-169376811 on Build GRCh38
6039927 c.58G>A p.A20T Chr.1: 169376569-169376569 on Build GRCh38
5543749 c.312T>C p.H104H Chr.1: 169376823-169376823 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768242599 Chr.1: 169376645-169376646 on Build GRCh38
rs772708018 Chr.1: 169376552-169376552 on Build GRCh38
rs766328635 Chr.1: 169376623-169376623 on Build GRCh38
rs781747448 Chr.1: 169376895-169376895 on Build GRCh38
rs376683357 Chr.1: 169376604-169376604 on Build GRCh38
rs369097632 Chr.1: 169376731-169376731 on Build GRCh38
rs373685508 Chr.1: 169376631-169376631 on Build GRCh38
rs200267284 Chr.1: 169376815-169376815 on Build GRCh38
rs746213830 Chr.1: 169376506-169376507 on Build GRCh38
rs762903045 Chr.1: 169376622-169376622 on Build GRCh38
rs753114990 Chr.1: 169376910-169376910 on Build GRCh38
rs760564299 Chr.1: 169376877-169376877 on Build GRCh38
rs748856934 Chr.1: 169376647-169376647 on Build GRCh38
rs139969512 Chr.1: 169376792-169376792 on Build GRCh38
rs771054710 Chr.1: 169376865-169376865 on Build GRCh38
rs755550842 Chr.1: 169376828-169376828 on Build GRCh38
rs141133669 Chr.1: 169376599-169376599 on Build GRCh38
rs779900034 Chr.1: 169376501-169376501 on Build GRCh38
rs969157009 Chr.1: 169376520-169376520 on Build GRCh38
rs763763600 Chr.1: 169376879-169376879 on Build GRCh38
rs901960632 Chr.1: 169376722-169376722 on Build GRCh38
rs764920388 Chr.1: 169376886-169376886 on Build GRCh38
rs773259204 Chr.1: 169376710-169376710 on Build GRCh38
rs115391337 Chr.1: 169376556-169376556 on Build GRCh38
rs12563090 Chr.1: 169376466-169376466 on Build GRCh38
rs757933902 Chr.1: 169376894-169376894 on Build GRCh38
rs761141553 Chr.1: 169376567-169376567 on Build GRCh38
rs112226207 Chr.1: 169376645-169376645 on Build GRCh38
rs769252535 Chr.1: 169376538-169376538 on Build GRCh38
rs755804345 Chr.1: 169376643-169376643 on Build GRCh38
rs762085138 Chr.1: 169376603-169376603 on Build GRCh38
rs776409088 Chr.1: 169376775-169376775 on Build GRCh38
rs988701432 Chr.1: 169376858-169376858 on Build GRCh38
rs565968264 Chr.1: 169376853-169376853 on Build GRCh38
rs564455805 Chr.1: 169376720-169376720 on Build GRCh38
rs751387736 Chr.1: 169376503-169376503 on Build GRCh38
rs767384089 Chr.1: 169376761-169376761 on Build GRCh38
rs777063096 Chr.1: 169376595-169376595 on Build GRCh38
rs981019862 Chr.1: 169376523-169376523 on Build GRCh38
rs756986387 Chr.1: 169376884-169376884 on Build GRCh38
rs747515594 Chr.1: 169376660-169376660 on Build GRCh38
rs753586765 Chr.1: 169376883-169376883 on Build GRCh38
rs750521552 Chr.1: 169376615-169376615 on Build GRCh38
rs752350260 Chr.1: 169376635-169376635 on Build GRCh38
rs756570009 Chr.1: 169376922-169376922 on Build GRCh38
rs761842054 Chr.1: 169376489-169376489 on Build GRCh38
rs754720051 Chr.1: 169376514-169376514 on Build GRCh38
rs761480949 Chr.1: 169376793-169376794 on Build GRCh38
rs201016056 Chr.1: 169376845-169376845 on Build GRCh38
rs765357521 Chr.1: 169376493-169376493 on Build GRCh38
rs564287842 Chr.1: 169376727-169376727 on Build GRCh38
rs200122718 Chr.1: 169376495-169376495 on Build GRCh38
rs769203821 Chr.1: 169376671-169376671 on Build GRCh38
rs145703017 Chr.1: 169376656-169376656 on Build GRCh38
rs759545677 Chr.1: 169376871-169376871 on Build GRCh38
rs777323039 Chr.1: 169376544-169376544 on Build GRCh38
rs748398107 Chr.1: 169376693-169376693 on Build GRCh38
rs747803518 Chr.1: 169376529-169376529 on Build GRCh38
rs530022938 Chr.1: 169376862-169376862 on Build GRCh38
rs1033223684 Chr.1: 169376834-169376834 on Build GRCh38
rs149479217 Chr.1: 169376554-169376554 on Build GRCh38
rs777071474 Chr.1: 169376672-169376672 on Build GRCh38
rs751479488 Chr.1: 169376626-169376626 on Build GRCh38
rs755821213 Chr.1: 169376793-169376793 on Build GRCh38
rs773525666 Chr.1: 169376861-169376861 on Build GRCh38
rs763826520 Chr.1: 169376798-169376798 on Build GRCh38
rs770034509 Chr.1: 169376856-169376856 on Build GRCh38
rs756775615 Chr.1: 169376653-169376653 on Build GRCh38
rs770086628 Chr.1: 169376705-169376705 on Build GRCh38
rs774427111 Chr.1: 169376869-169376869 on Build GRCh38
rs142785938 Chr.1: 169376746-169376746 on Build GRCh38
rs548356529 Chr.1: 169376872-169376872 on Build GRCh38
rs142091756 Chr.1: 169376843-169376843 on Build GRCh38
rs753407327 Chr.1: 169376801-169376801 on Build GRCh38
rs1028180 Chr.1: 169376630-169376630 on Build GRCh38
rs372377326 Chr.1: 169376545-169376545 on Build GRCh38
rs368115641 Chr.1: 169376739-169376739 on Build GRCh38
rs201682805 Chr.1: 169376570-169376570 on Build GRCh38
rs758374408 Chr.1: 169376498-169376498 on Build GRCh38
rs767517784 Chr.1: 169376875-169376875 on Build GRCh38
rs143882259 Chr.1: 169376813-169376813 on Build GRCh38
rs796786690 Chr.1: 169376481-169376481 on Build GRCh38
rs745319013 Chr.1: 169376822-169376822 on Build GRCh38
rs780873451 Chr.1: 169376528-169376528 on Build GRCh38

View Full Product Details