Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 94823211-94823716 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INTS8
Entrez Gene ID
55656
Gene Name
integrator complex subunit 8
Gene Aliases
C8orf52, INT8
UniGene
Hs.727669
Gene Chromosome Location
Chr.8: 94823290-94894664 on Build GRCh38
Transcripts
NM_017864.3 NR_073444.1 NR_073445.1 XM_017013616.1 XM_017013617.1 XM_017013618.1 XM_017013619.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6432857 c.65C>A p.T22N Chr.8: 94823496-94823496 on Build GRCh38
7254747 c.63G>A p.Q21Q Chr.8: 94823494-94823494 on Build GRCh38
404949 c.42delC p.R15fs*37 Chr.8: 94823473-94823473 on Build GRCh38
6549549 c.91G>T p.E31* Chr.8: 94823522-94823522 on Build GRCh38
5033714 c.52A>C p.T18P Chr.8: 94823483-94823483 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs982855710 Chr.8: 94823365-94823365 on Build GRCh38
rs116966031 Chr.8: 94823626-94823626 on Build GRCh38
rs898331397 Chr.8: 94823264-94823264 on Build GRCh38
rs961020811 Chr.8: 94823678-94823678 on Build GRCh38
rs942274548 Chr.8: 94823352-94823352 on Build GRCh38
rs953824940 Chr.8: 94823637-94823637 on Build GRCh38
rs570029094 Chr.8: 94823237-94823237 on Build GRCh38
rs939900446 Chr.8: 94823251-94823251 on Build GRCh38
rs868227331 Chr.8: 94823684-94823684 on Build GRCh38
rs558228803 Chr.8: 94823603-94823603 on Build GRCh38
rs535288532 Chr.8: 94823320-94823320 on Build GRCh38
rs1038023837 Chr.8: 94823377-94823377 on Build GRCh38
rs572324564 Chr.8: 94823619-94823619 on Build GRCh38
rs757947939 Chr.8: 94823311-94823311 on Build GRCh38
rs773053706 Chr.8: 94823548-94823548 on Build GRCh38
rs912266982 Chr.8: 94823677-94823677 on Build GRCh38
rs1016651527 Chr.8: 94823254-94823254 on Build GRCh38
rs766064483 Chr.8: 94823568-94823568 on Build GRCh38
rs538672061 Chr.8: 94823593-94823593 on Build GRCh38
rs898153343 Chr.8: 94823449-94823449 on Build GRCh38
rs757657197 Chr.8: 94823333-94823333 on Build GRCh38
rs202080432 Chr.8: 94823525-94823525 on Build GRCh38
rs775779799 Chr.8: 94823232-94823232 on Build GRCh38
rs768687518 Chr.8: 94823246-94823246 on Build GRCh38
rs1054935429 Chr.8: 94823452-94823452 on Build GRCh38
rs780258682 Chr.8: 94823453-94823453 on Build GRCh38
rs974318095 Chr.8: 94823483-94823483 on Build GRCh38
rs761696792 Chr.8: 94823281-94823281 on Build GRCh38
rs914547244 Chr.8: 94823317-94823317 on Build GRCh38
rs972366406 Chr.8: 94823688-94823688 on Build GRCh38
rs990122939 Chr.8: 94823315-94823315 on Build GRCh38
rs775086485 Chr.8: 94823349-94823349 on Build GRCh38
rs776258209 Chr.8: 94823271-94823271 on Build GRCh38
rs773814685 Chr.8: 94823580-94823580 on Build GRCh38
rs759111018 Chr.8: 94823581-94823581 on Build GRCh38
rs150893760 Chr.8: 94823242-94823244 on Build GRCh38
rs574318023 Chr.8: 94823685-94823685 on Build GRCh38
rs776651574 Chr.8: 94823513-94823513 on Build GRCh38
rs994958359 Chr.8: 94823275-94823275 on Build GRCh38
rs772481208 Chr.8: 94823423-94823423 on Build GRCh38
rs779413330 Chr.8: 94823325-94823325 on Build GRCh38
rs748463684 Chr.8: 94823543-94823543 on Build GRCh38
rs747125099 Chr.8: 94823458-94823458 on Build GRCh38
rs919779328 Chr.8: 94823413-94823413 on Build GRCh38
rs886776043 Chr.8: 94823244-94823244 on Build GRCh38
rs868819375 Chr.8: 94823528-94823528 on Build GRCh38
rs140625955 Chr.8: 94823329-94823329 on Build GRCh38
rs986492665 Chr.8: 94823343-94823343 on Build GRCh38
rs886663070 Chr.8: 94823350-94823350 on Build GRCh38
rs933450093 Chr.8: 94823342-94823342 on Build GRCh38
rs1031833998 Chr.8: 94823313-94823313 on Build GRCh38
rs1013377210 Chr.8: 94823472-94823472 on Build GRCh38
rs568636359 Chr.8: 94823332-94823332 on Build GRCh38
rs757201725 Chr.8: 94823263-94823263 on Build GRCh38
rs113908747 Chr.8: 94823634-94823634 on Build GRCh38
rs537627111 Chr.8: 94823395-94823395 on Build GRCh38
rs187896056 Chr.8: 94823303-94823303 on Build GRCh38
rs1013063465 Chr.8: 94823283-94823283 on Build GRCh38
rs962987968 Chr.8: 94823468-94823468 on Build GRCh38
rs965034844 Chr.8: 94823664-94823664 on Build GRCh38
rs756551758 Chr.8: 94823326-94823326 on Build GRCh38
rs564907558 Chr.8: 94823310-94823310 on Build GRCh38
rs1031866623 Chr.8: 94823622-94823622 on Build GRCh38
rs981365920 Chr.8: 94823306-94823306 on Build GRCh38
rs768436666 Chr.8: 94823479-94823479 on Build GRCh38
rs1015223841 Chr.8: 94823465-94823465 on Build GRCh38
rs532584053 Chr.8: 94823285-94823285 on Build GRCh38
rs955971451 Chr.8: 94823625-94823625 on Build GRCh38
rs746085669 Chr.8: 94823421-94823421 on Build GRCh38
rs779212015 Chr.8: 94823405-94823405 on Build GRCh38
rs576055249 Chr.8: 94823575-94823575 on Build GRCh38
rs768185430 Chr.8: 94823347-94823347 on Build GRCh38
rs1002685684 Chr.8: 94823576-94823576 on Build GRCh38
rs75566084 Chr.8: 94823314-94823314 on Build GRCh38
rs931150349 Chr.8: 94823424-94823424 on Build GRCh38
rs769633903 Chr.8: 94823547-94823547 on Build GRCh38
rs762576838 Chr.8: 94823560-94823560 on Build GRCh38

View Full Product Details