Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 98595954-98596466 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STK3
Entrez Gene ID
6788
Gene Name
serine/threonine kinase 3
Gene Aliases
KRS1, MST2
UniGene
Hs.492333
Gene Chromosome Location
Chr.8: 98429677-98944329 on Build GRCh38
Transcripts
NM_001256312.1 NM_001256313.1 NM_006281.3 XM_011517248.2 XM_011517251.1 XM_011517252.2 XM_011517253.2 XM_011517255.2 XM_011517258.1 XM_017013756.1 XM_017013757.1 XM_017013758.1 XM_017013759.1 XM_017013760.1 XM_017013761.1 XM_017013763.1 XM_017013764.1 XM_017013765.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4398614 c.774T>A p.C258* Chr.8: 98596080-98596080 on Build GRCh38
6583742 c.770A>G p.K257R Chr.8: 98596084-98596084 on Build GRCh38
5711060 c.735A>C p.E245D Chr.8: 98596119-98596119 on Build GRCh38
3699229 c.752T>C p.F251S Chr.8: 98596102-98596102 on Build GRCh38
6338845 c.793C>A p.Q265K Chr.8: 98596061-98596061 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756524585 Chr.8: 98596082-98596082 on Build GRCh38
rs567929051 Chr.8: 98596028-98596028 on Build GRCh38
rs766207319 Chr.8: 98596285-98596285 on Build GRCh38
rs775218846 Chr.8: 98596002-98596002 on Build GRCh38
rs1044970778 Chr.8: 98596099-98596099 on Build GRCh38
rs1007253796 Chr.8: 98596387-98596387 on Build GRCh38
rs983939069 Chr.8: 98596077-98596077 on Build GRCh38
rs763696181 Chr.8: 98596076-98596076 on Build GRCh38
rs753518740 Chr.8: 98596204-98596204 on Build GRCh38
rs200968112 Chr.8: 98595982-98595982 on Build GRCh38
rs1033455548 Chr.8: 98595996-98595996 on Build GRCh38
rs528356388 Chr.8: 98595993-98595993 on Build GRCh38
rs376072788 Chr.8: 98596385-98596385 on Build GRCh38
rs779880596 Chr.8: 98596110-98596110 on Build GRCh38
rs772923376 Chr.8: 98596205-98596205 on Build GRCh38
rs764183632 Chr.8: 98596086-98596086 on Build GRCh38
rs1047651017 Chr.8: 98596308-98596308 on Build GRCh38
rs754065361 Chr.8: 98596095-98596095 on Build GRCh38
rs757271491 Chr.8: 98596097-98596097 on Build GRCh38
rs539573906 Chr.8: 98596312-98596312 on Build GRCh38
rs768524020 Chr.8: 98596149-98596149 on Build GRCh38
rs545250221 Chr.8: 98596376-98596376 on Build GRCh38
rs373705485 Chr.8: 98596010-98596010 on Build GRCh38
rs753043269 Chr.8: 98596067-98596067 on Build GRCh38
rs536253432 Chr.8: 98596188-98596188 on Build GRCh38
rs146193062 Chr.8: 98595997-98595997 on Build GRCh38
rs993245976 Chr.8: 98596245-98596245 on Build GRCh38
rs377634083 Chr.8: 98596021-98596021 on Build GRCh38
rs746728067 Chr.8: 98596113-98596113 on Build GRCh38
rs767919046 Chr.8: 98596048-98596048 on Build GRCh38
rs550650334 Chr.8: 98596211-98596211 on Build GRCh38
rs928930708 Chr.8: 98596299-98596299 on Build GRCh38
rs911365020 Chr.8: 98596210-98596210 on Build GRCh38
rs758420963 Chr.8: 98596190-98596190 on Build GRCh38
rs1018502820 Chr.8: 98596417-98596417 on Build GRCh38
rs771891390 Chr.8: 98596001-98596001 on Build GRCh38
rs764842623 Chr.8: 98596027-98596027 on Build GRCh38
rs760017613 Chr.8: 98596043-98596043 on Build GRCh38
rs556478681 Chr.8: 98596427-98596427 on Build GRCh38
rs745804050 Chr.8: 98596107-98596107 on Build GRCh38
rs747813345 Chr.8: 98596180-98596180 on Build GRCh38
rs760347812 Chr.8: 98596003-98596003 on Build GRCh38
rs776353449 Chr.8: 98596151-98596151 on Build GRCh38
rs776441628 Chr.8: 98596014-98596014 on Build GRCh38
rs552472303 Chr.8: 98596337-98596337 on Build GRCh38
rs1005986692 Chr.8: 98595992-98595992 on Build GRCh38
rs1033507996 Chr.8: 98596431-98596431 on Build GRCh38
rs149198380 Chr.8: 98596098-98596098 on Build GRCh38
rs943165270 Chr.8: 98596225-98596225 on Build GRCh38
rs778757118 Chr.8: 98595989-98595989 on Build GRCh38
rs146713666 Chr.8: 98596253-98596253 on Build GRCh38

View Full Product Details