Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 98944311-98944817 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OSR2
Entrez Gene ID
116039
Gene Name
odd-skipped related transciption factor 2
Gene Aliases
-
UniGene
Hs.253247
Gene Chromosome Location
Chr.8: 98944403-98952104 on Build GRCh38
Transcripts
NM_001142462.2 NM_001286841.1 NM_053001.3 XM_005250778.4 XM_011516825.2 XM_011517248.2 XM_011517252.2 XM_011517257.2 XM_017013756.1 XM_017013757.1 XM_017013762.1

Gene Symbol
STK3
Entrez Gene ID
6788
Gene Name
serine/threonine kinase 3
Gene Aliases
KRS1, MST2
UniGene
Hs.492333
Gene Chromosome Location
Chr.8: 98429677-98944329 on Build GRCh38
Transcripts
NM_001142462.2 NM_001286841.1 NM_053001.3 XM_005250778.4 XM_011516825.2 XM_011517248.2 XM_011517252.2 XM_011517257.2 XM_017013756.1 XM_017013757.1 XM_017013762.1

SNP Detail

SNP ID Chromosome Location
rs1021137082 Chr.8: 98944492-98944492 on Build GRCh38
rs577711386 Chr.8: 98944417-98944417 on Build GRCh38
rs1040171105 Chr.8: 98944460-98944460 on Build GRCh38
rs1011178413 Chr.8: 98944482-98944482 on Build GRCh38
rs924837179 Chr.8: 98944795-98944795 on Build GRCh38
rs7829755 Chr.8: 98944384-98944384 on Build GRCh38
rs866145346 Chr.8: 98944377-98944377 on Build GRCh38
rs944561225 Chr.8: 98944694-98944694 on Build GRCh38
rs376653056 Chr.8: 98944444-98944444 on Build GRCh38
rs771195257 Chr.8: 98944392-98944392 on Build GRCh38
rs1033039215 Chr.8: 98944379-98944379 on Build GRCh38
rs1041940563 Chr.8: 98944479-98944479 on Build GRCh38
rs1041661110 Chr.8: 98944359-98944359 on Build GRCh38
rs538124956 Chr.8: 98944358-98944358 on Build GRCh38
rs991249365 Chr.8: 98944676-98944676 on Build GRCh38
rs186395496 Chr.8: 98944341-98944341 on Build GRCh38
rs959390938 Chr.8: 98944412-98944412 on Build GRCh38
rs536645557 Chr.8: 98944716-98944716 on Build GRCh38
rs545440286 Chr.8: 98944424-98944424 on Build GRCh38
rs1007109479 Chr.8: 98944393-98944393 on Build GRCh38
rs965634733 Chr.8: 98944397-98944397 on Build GRCh38
rs750902297 Chr.8: 98944437-98944437 on Build GRCh38
rs1027658816 Chr.8: 98944598-98944598 on Build GRCh38
rs998130054 Chr.8: 98944558-98944558 on Build GRCh38
rs945692831 Chr.8: 98944784-98944784 on Build GRCh38
rs1054483779 Chr.8: 98944491-98944491 on Build GRCh38
rs892818805 Chr.8: 98944770-98944770 on Build GRCh38
rs983758122 Chr.8: 98944647-98944647 on Build GRCh38
rs555511444 Chr.8: 98944333-98944333 on Build GRCh38
rs1035107557 Chr.8: 98944546-98944546 on Build GRCh38
rs950342177 Chr.8: 98944469-98944469 on Build GRCh38
rs542370359 Chr.8: 98944525-98944525 on Build GRCh38
rs902719911 Chr.8: 98944496-98944496 on Build GRCh38
rs889168434 Chr.8: 98944499-98944499 on Build GRCh38
rs1018566206 Chr.8: 98944532-98944532 on Build GRCh38
rs186424792 Chr.8: 98944613-98944613 on Build GRCh38
rs532855642 Chr.8: 98944343-98944343 on Build GRCh38
rs951044004 Chr.8: 98944634-98944634 on Build GRCh38
rs13277546 Chr.8: 98944411-98944411 on Build GRCh38
rs1011538780 Chr.8: 98944381-98944381 on Build GRCh38
rs903184639 Chr.8: 98944365-98944365 on Build GRCh38
rs573649728 Chr.8: 98944463-98944463 on Build GRCh38
rs931750660 Chr.8: 98944717-98944717 on Build GRCh38
rs917510055 Chr.8: 98944465-98944465 on Build GRCh38
rs930585206 Chr.8: 98944437-98944437 on Build GRCh38
rs983893537 Chr.8: 98944432-98944432 on Build GRCh38
rs977661951 Chr.8: 98944759-98944759 on Build GRCh38
rs1000129398 Chr.8: 98944489-98944489 on Build GRCh38
rs1056333685 Chr.8: 98944458-98944458 on Build GRCh38
rs966829261 Chr.8: 98944388-98944388 on Build GRCh38
rs1054588798 Chr.8: 98944762-98944762 on Build GRCh38
rs983417080 Chr.8: 98944632-98944632 on Build GRCh38
rs753214092 Chr.8: 98944371-98944372 on Build GRCh38
rs951012879 Chr.8: 98944428-98944428 on Build GRCh38
rs920543707 Chr.8: 98944427-98944427 on Build GRCh38
rs1014263260 Chr.8: 98944345-98944345 on Build GRCh38
rs1006729461 Chr.8: 98944527-98944527 on Build GRCh38
rs907773675 Chr.8: 98944668-98944668 on Build GRCh38
rs918288894 Chr.8: 98944423-98944423 on Build GRCh38
rs892753190 Chr.8: 98944473-98944473 on Build GRCh38
rs1054133360 Chr.8: 98944471-98944471 on Build GRCh38
rs950203075 Chr.8: 98944742-98944742 on Build GRCh38
rs1016691035 Chr.8: 98944650-98944650 on Build GRCh38
rs759968661 Chr.8: 98944545-98944545 on Build GRCh38
rs181731116 Chr.8: 98944456-98944456 on Build GRCh38
rs907721338 Chr.8: 98944448-98944448 on Build GRCh38
rs764224945 Chr.8: 98944580-98944580 on Build GRCh38
rs917435970 Chr.8: 98944737-98944737 on Build GRCh38
rs976664672 Chr.8: 98944413-98944413 on Build GRCh38
rs958659380 Chr.8: 98944673-98944673 on Build GRCh38
rs903217065 Chr.8: 98944483-98944483 on Build GRCh38
rs4735586 Chr.8: 98944533-98944533 on Build GRCh38
rs900196827 Chr.8: 98944336-98944336 on Build GRCh38
rs573090111 Chr.8: 98944699-98944699 on Build GRCh38
rs552568042 Chr.8: 98944625-98944625 on Build GRCh38
rs1018200733 Chr.8: 98944395-98944395 on Build GRCh38
rs959478073 Chr.8: 98944549-98944549 on Build GRCh38

View Full Product Details