Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 102205679-102206215 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RRM2B
Entrez Gene ID
50484
Gene Name
ribonucleotide reductase regulatory TP53 inducible subunit M2B
Gene Aliases
MTDPS8A, MTDPS8B, P53R2
UniGene
Hs.512592
Gene Chromosome Location
Chr.8: 102204501-102239118 on Build GRCh38
Transcripts
NM_001172477.1 NM_001172478.1 NM_015713.4

SNP Detail

SNP ID Chromosome Location
rs915101195 Chr.8: 102206154-102206154 on Build GRCh38
rs976924911 Chr.8: 102205769-102205769 on Build GRCh38
rs886062565 Chr.8: 102205805-102205805 on Build GRCh38
rs987923267 Chr.8: 102206150-102206150 on Build GRCh38
rs956518317 Chr.8: 102206148-102206148 on Build GRCh38
rs566194929 Chr.8: 102205897-102205897 on Build GRCh38
rs994070496 Chr.8: 102205912-102205912 on Build GRCh38
rs200678743 Chr.8: 102205917-102205936 on Build GRCh38
rs775418054 Chr.8: 102205790-102205790 on Build GRCh38
rs11987823 Chr.8: 102206180-102206180 on Build GRCh38
rs767476703 Chr.8: 102205723-102205723 on Build GRCh38
rs937253891 Chr.8: 102205865-102205865 on Build GRCh38
rs1055958 Chr.8: 102206098-102206098 on Build GRCh38
rs533746921 Chr.8: 102205917-102205917 on Build GRCh38
rs144351681 Chr.8: 102206082-102206082 on Build GRCh38
rs374142867 Chr.8: 102206126-102206127 on Build GRCh38
rs1036553376 Chr.8: 102205931-102205931 on Build GRCh38
rs926593167 Chr.8: 102206185-102206185 on Build GRCh38
rs562089263 Chr.8: 102205720-102205720 on Build GRCh38
rs927178780 Chr.8: 102205856-102205856 on Build GRCh38
rs1032303252 Chr.8: 102206155-102206155 on Build GRCh38
rs765908148 Chr.8: 102205834-102205834 on Build GRCh38
rs915153197 Chr.8: 102205805-102205805 on Build GRCh38
rs981299480 Chr.8: 102205835-102205835 on Build GRCh38
rs988034737 Chr.8: 102205787-102205787 on Build GRCh38
rs116394626 Chr.8: 102206036-102206036 on Build GRCh38
rs191550743 Chr.8: 102205882-102205882 on Build GRCh38
rs139212686 Chr.8: 102205997-102205997 on Build GRCh38
rs1045028065 Chr.8: 102205853-102205853 on Build GRCh38
rs1049292303 Chr.8: 102205963-102205963 on Build GRCh38
rs967917245 Chr.8: 102206172-102206172 on Build GRCh38
rs946666638 Chr.8: 102205839-102205839 on Build GRCh38
rs940417354 Chr.8: 102205911-102205911 on Build GRCh38
rs997909756 Chr.8: 102206100-102206100 on Build GRCh38
rs1027912262 Chr.8: 102205707-102205707 on Build GRCh38
rs1017981106 Chr.8: 102205724-102205724 on Build GRCh38
rs73698394 Chr.8: 102206146-102206146 on Build GRCh38
rs752269488 Chr.8: 102206114-102206114 on Build GRCh38
rs535466108 Chr.8: 102206121-102206121 on Build GRCh38
rs563908556 Chr.8: 102205981-102205982 on Build GRCh38
rs993731268 Chr.8: 102205704-102205705 on Build GRCh38
rs956751959 Chr.8: 102205780-102205780 on Build GRCh38
rs552389522 Chr.8: 102205982-102205982 on Build GRCh38
rs1007837281 Chr.8: 102206028-102206028 on Build GRCh38
rs887463345 Chr.8: 102205901-102205901 on Build GRCh38
rs529374843 Chr.8: 102205756-102205756 on Build GRCh38
rs1029177769 Chr.8: 102206043-102206043 on Build GRCh38

View Full Product Details