Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 102206541-102207050 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RRM2B
Entrez Gene ID
50484
Gene Name
ribonucleotide reductase regulatory TP53 inducible subunit M2B
Gene Aliases
MTDPS8A, MTDPS8B, P53R2
UniGene
Hs.512592
Gene Chromosome Location
Chr.8: 102204501-102239118 on Build GRCh38
Transcripts
NM_001172477.1 NM_001172478.1 NM_015713.4

SNP Detail

SNP ID Chromosome Location
rs577176343 Chr.8: 102206623-102206623 on Build GRCh38
rs1050220684 Chr.8: 102206860-102206860 on Build GRCh38
rs553768124 Chr.8: 102206887-102206887 on Build GRCh38
rs568039559 Chr.8: 102206861-102206861 on Build GRCh38
rs553439824 Chr.8: 102206608-102206608 on Build GRCh38
rs906573185 Chr.8: 102206983-102206983 on Build GRCh38
rs769066939 Chr.8: 102207008-102207008 on Build GRCh38
rs537712055 Chr.8: 102206794-102206794 on Build GRCh38
rs764193033 Chr.8: 102206786-102206786 on Build GRCh38
rs4102401 Chr.8: 102207012-102207012 on Build GRCh38
rs992107305 Chr.8: 102206851-102206851 on Build GRCh38
rs889321989 Chr.8: 102206611-102206611 on Build GRCh38
rs560789226 Chr.8: 102206886-102206886 on Build GRCh38
rs1009850064 Chr.8: 102206951-102206951 on Build GRCh38
rs962002084 Chr.8: 102206796-102206796 on Build GRCh38
rs936803811 Chr.8: 102206892-102206892 on Build GRCh38
rs114108597 Chr.8: 102206782-102206782 on Build GRCh38
rs1026704249 Chr.8: 102206852-102206852 on Build GRCh38
rs892146071 Chr.8: 102206769-102206769 on Build GRCh38
rs29000288 Chr.8: 102206879-102206879 on Build GRCh38
rs759674886 Chr.8: 102206785-102206785 on Build GRCh38
rs972128900 Chr.8: 102206797-102206797 on Build GRCh38
rs1051017101 Chr.8: 102206581-102206581 on Build GRCh38
rs11575864 Chr.8: 102206682-102206682 on Build GRCh38
rs1024987811 Chr.8: 102206684-102206684 on Build GRCh38
rs909291479 Chr.8: 102206930-102206930 on Build GRCh38
rs971206464 Chr.8: 102206730-102206730 on Build GRCh38
rs5005121 Chr.8: 102206856-102206856 on Build GRCh38
rs1265116 Chr.8: 102206645-102206645 on Build GRCh38
rs929351400 Chr.8: 102206569-102206569 on Build GRCh38
rs11575865 Chr.8: 102206675-102206675 on Build GRCh38
rs533476352 Chr.8: 102206738-102206738 on Build GRCh38
rs11540327 Chr.8: 102206962-102206962 on Build GRCh38
rs1046811088 Chr.8: 102206954-102206954 on Build GRCh38
rs370688634 Chr.8: 102206920-102206920 on Build GRCh38
rs1053898857 Chr.8: 102206919-102206919 on Build GRCh38
rs902081926 Chr.8: 102206663-102206663 on Build GRCh38
rs922988015 Chr.8: 102206844-102206844 on Build GRCh38
rs892723282 Chr.8: 102206640-102206640 on Build GRCh38
rs29000289 Chr.8: 102206839-102206839 on Build GRCh38
rs943519162 Chr.8: 102206633-102206633 on Build GRCh38
rs1001065537 Chr.8: 102206612-102206612 on Build GRCh38
rs553453635 Chr.8: 102206764-102206764 on Build GRCh38
rs547817608 Chr.8: 102206607-102206607 on Build GRCh38

View Full Product Details