Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 103399536-103400076 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A32
Entrez Gene ID
81034
Gene Name
solute carrier family 25 member 32
Gene Aliases
MFT, MFTC, RREI
UniGene
Hs.532265
Gene Chromosome Location
Chr.8: 103398638-103415335 on Build GRCh38
Transcripts
NM_030780.4 NR_102337.1 NR_102338.1 XM_017013877.1

SNP Detail

SNP ID Chromosome Location
rs926630412 Chr.8: 103399561-103399561 on Build GRCh38
rs891694700 Chr.8: 103400029-103400029 on Build GRCh38
rs908830249 Chr.8: 103399883-103399883 on Build GRCh38
rs141758872 Chr.8: 103399969-103399969 on Build GRCh38
rs1040749746 Chr.8: 103399703-103399703 on Build GRCh38
rs891921436 Chr.8: 103399983-103399983 on Build GRCh38
rs1048715891 Chr.8: 103399643-103399643 on Build GRCh38
rs919163113 Chr.8: 103399605-103399605 on Build GRCh38
rs1041165439 Chr.8: 103399900-103399900 on Build GRCh38
rs1031626748 Chr.8: 103399824-103399824 on Build GRCh38
rs1049368416 Chr.8: 103399763-103399763 on Build GRCh38
rs955844006 Chr.8: 103399829-103399829 on Build GRCh38
rs940293252 Chr.8: 103399884-103399884 on Build GRCh38
rs919204035 Chr.8: 103399857-103399857 on Build GRCh38
rs902025445 Chr.8: 103399671-103399671 on Build GRCh38
rs537947607 Chr.8: 103399655-103399655 on Build GRCh38
rs779369117 Chr.8: 103399836-103399836 on Build GRCh38
rs939161673 Chr.8: 103399568-103399568 on Build GRCh38
rs1049981187 Chr.8: 103399927-103399927 on Build GRCh38
rs969998956 Chr.8: 103399806-103399806 on Build GRCh38
rs1038768121 Chr.8: 103399665-103399665 on Build GRCh38
rs552235128 Chr.8: 103399747-103399747 on Build GRCh38
rs539289465 Chr.8: 103399861-103399861 on Build GRCh38
rs375001979 Chr.8: 103399719-103399719 on Build GRCh38
rs1007317620 Chr.8: 103399662-103399662 on Build GRCh38
rs749653197 Chr.8: 103399955-103399955 on Build GRCh38
rs952865558 Chr.8: 103399874-103399874 on Build GRCh38
rs940228503 Chr.8: 103399702-103399702 on Build GRCh38
rs997010802 Chr.8: 103399720-103399720 on Build GRCh38
rs878912062 Chr.8: 103399939-103399939 on Build GRCh38
rs970808930 Chr.8: 103399840-103399840 on Build GRCh38
rs184020953 Chr.8: 103399876-103399876 on Build GRCh38
rs928993932 Chr.8: 103399607-103399607 on Build GRCh38
rs565205394 Chr.8: 103400034-103400034 on Build GRCh38
rs148028640 Chr.8: 103399923-103399923 on Build GRCh38
rs552422295 Chr.8: 103399615-103399615 on Build GRCh38
rs188464949 Chr.8: 103399813-103399813 on Build GRCh38
rs980171784 Chr.8: 103399811-103399813 on Build GRCh38
rs900895687 Chr.8: 103399718-103399718 on Build GRCh38
rs1014121498 Chr.8: 103399830-103399830 on Build GRCh38
rs931550629 Chr.8: 103399593-103399593 on Build GRCh38
rs368254292 Chr.8: 103399766-103399766 on Build GRCh38
rs192237910 Chr.8: 103399869-103399869 on Build GRCh38
rs1053006434 Chr.8: 103400004-103400004 on Build GRCh38
rs778523785 Chr.8: 103399678-103399678 on Build GRCh38
rs904179989 Chr.8: 103399935-103399935 on Build GRCh38
rs1048632985 Chr.8: 103399640-103399640 on Build GRCh38
rs887564809 Chr.8: 103399661-103399661 on Build GRCh38
rs942984462 Chr.8: 103399901-103399901 on Build GRCh38
rs774556062 Chr.8: 103400009-103400009 on Build GRCh38
rs1053281720 Chr.8: 103399960-103399960 on Build GRCh38
rs779291965 Chr.8: 103399641-103399641 on Build GRCh38
rs748475839 Chr.8: 103399745-103399745 on Build GRCh38
rs997659348 Chr.8: 103399690-103399690 on Build GRCh38
rs1021631692 Chr.8: 103399770-103399770 on Build GRCh38
rs570626103 Chr.8: 103399646-103399646 on Build GRCh38
rs778325079 Chr.8: 103399943-103399943 on Build GRCh38
rs376307460 Chr.8: 103399813-103399815 on Build GRCh38
rs770163870 Chr.8: 103399915-103399915 on Build GRCh38

View Full Product Details