Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 103401409-103401955 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A32
Entrez Gene ID
81034
Gene Name
solute carrier family 25 member 32
Gene Aliases
MFT, MFTC, RREI
UniGene
Hs.532265
Gene Chromosome Location
Chr.8: 103398638-103415335 on Build GRCh38
Transcripts
NM_030780.4 NR_102337.1 NR_102338.1 XM_017013877.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6580323 c.666+8T>G p.? Chr.8: 103401933-103401933 on Build GRCh38
6179212 c.680A>T p.Y227F Chr.8: 103401648-103401648 on Build GRCh38
4846271 c.741C>T p.V247V Chr.8: 103401587-103401587 on Build GRCh38
4430788 c.685_686insAT p.S229fs*21 Chr.8: 103401642-103401643 on Build GRCh38
1094474 c.752G>A p.R251H Chr.8: 103401576-103401576 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs942746458 Chr.8: 103401818-103401818 on Build GRCh38
rs771751865 Chr.8: 103401903-103401903 on Build GRCh38
rs1024895231 Chr.8: 103401559-103401559 on Build GRCh38
rs753803892 Chr.8: 103401852-103401852 on Build GRCh38
rs376664535 Chr.8: 103401635-103401635 on Build GRCh38
rs773133326 Chr.8: 103401515-103401515 on Build GRCh38
rs146625518 Chr.8: 103401578-103401578 on Build GRCh38
rs568817869 Chr.8: 103401468-103401479 on Build GRCh38
rs934029747 Chr.8: 103401721-103401721 on Build GRCh38
rs1010508502 Chr.8: 103401895-103401895 on Build GRCh38
rs529126483 Chr.8: 103401860-103401860 on Build GRCh38
rs752845114 Chr.8: 103401677-103401677 on Build GRCh38
rs889796729 Chr.8: 103401468-103401468 on Build GRCh38
rs774089760 Chr.8: 103401550-103401550 on Build GRCh38
rs369582581 Chr.8: 103401500-103401500 on Build GRCh38
rs376243577 Chr.8: 103401913-103401913 on Build GRCh38
rs199724541 Chr.8: 103401477-103401477 on Build GRCh38
rs758926885 Chr.8: 103401596-103401596 on Build GRCh38
rs749320247 Chr.8: 103401712-103401712 on Build GRCh38
rs193210327 Chr.8: 103401850-103401850 on Build GRCh38
rs999850066 Chr.8: 103401759-103401763 on Build GRCh38
rs371592738 Chr.8: 103401605-103401605 on Build GRCh38
rs763585056 Chr.8: 103401646-103401646 on Build GRCh38
rs372532340 Chr.8: 103401611-103401611 on Build GRCh38
rs59283978 Chr.8: 103401496-103401496 on Build GRCh38
rs894106040 Chr.8: 103401798-103401798 on Build GRCh38
rs1033973938 Chr.8: 103401794-103401794 on Build GRCh38
rs757236671 Chr.8: 103401898-103401898 on Build GRCh38
rs907406017 Chr.8: 103401914-103401914 on Build GRCh38
rs141367018 Chr.8: 103401586-103401586 on Build GRCh38
rs3133808 Chr.8: 103401824-103401824 on Build GRCh38
rs781540686 Chr.8: 103401675-103401675 on Build GRCh38
rs889778132 Chr.8: 103401769-103401769 on Build GRCh38
rs767017678 Chr.8: 103401558-103401558 on Build GRCh38
rs759232426 Chr.8: 103401633-103401633 on Build GRCh38
rs766941239 Chr.8: 103401655-103401655 on Build GRCh38
rs904433258 Chr.8: 103401744-103401744 on Build GRCh38
rs765813564 Chr.8: 103401564-103401564 on Build GRCh38
rs776301527 Chr.8: 103401928-103401928 on Build GRCh38
rs758804462 Chr.8: 103401483-103401483 on Build GRCh38
rs752308059 Chr.8: 103401560-103401560 on Build GRCh38
rs748296598 Chr.8: 103401610-103401610 on Build GRCh38
rs763710113 Chr.8: 103401641-103401641 on Build GRCh38
rs756297739 Chr.8: 103401698-103401698 on Build GRCh38
rs1024027768 Chr.8: 103401843-103401843 on Build GRCh38
rs759228145 Chr.8: 103401556-103401556 on Build GRCh38
rs762781054 Chr.8: 103401526-103401526 on Build GRCh38
rs377027414 Chr.8: 103401912-103401912 on Build GRCh38
rs904214067 Chr.8: 103401442-103401442 on Build GRCh38
rs145785157 Chr.8: 103401630-103401630 on Build GRCh38
rs752016774 Chr.8: 103401665-103401665 on Build GRCh38
rs768382437 Chr.8: 103401919-103401919 on Build GRCh38
rs771797789 Chr.8: 103401634-103401634 on Build GRCh38
rs760097451 Chr.8: 103401561-103401561 on Build GRCh38
rs755289808 Chr.8: 103401913-103401913 on Build GRCh38
rs971518583 Chr.8: 103401600-103401600 on Build GRCh38
rs754096857 Chr.8: 103401669-103401669 on Build GRCh38
rs766450156 Chr.8: 103401558-103401559 on Build GRCh38
rs755263271 Chr.8: 103401668-103401668 on Build GRCh38
rs371908900 Chr.8: 103401472-103401472 on Build GRCh38
rs781261123 Chr.8: 103401609-103401609 on Build GRCh38
rs780598021 Chr.8: 103401603-103401603 on Build GRCh38
rs201952067 Chr.8: 103401527-103401527 on Build GRCh38
rs374270449 Chr.8: 103401505-103401505 on Build GRCh38
rs760290484 Chr.8: 103401638-103401638 on Build GRCh38
rs541213162 Chr.8: 103401872-103401872 on Build GRCh38
rs1051230061 Chr.8: 103401742-103401742 on Build GRCh38
rs1054696999 Chr.8: 103401821-103401821 on Build GRCh38
rs143775940 Chr.8: 103401619-103401619 on Build GRCh38
rs369641181 Chr.8: 103401701-103401701 on Build GRCh38
rs893371416 Chr.8: 103401882-103401882 on Build GRCh38
rs755243766 Chr.8: 103401607-103401607 on Build GRCh38
rs754143542 Chr.8: 103401680-103401681 on Build GRCh38
rs983458477 Chr.8: 103401594-103401594 on Build GRCh38
rs982957978 Chr.8: 103401870-103401870 on Build GRCh38
rs745743736 Chr.8: 103401902-103401902 on Build GRCh38
rs115968673 Chr.8: 103401506-103401506 on Build GRCh38
rs777776802 Chr.8: 103401613-103401613 on Build GRCh38
rs753330767 Chr.8: 103401642-103401642 on Build GRCh38
rs770819305 Chr.8: 103401628-103401628 on Build GRCh38
rs778793529 Chr.8: 103401900-103401900 on Build GRCh38
rs940630776 Chr.8: 103401685-103401685 on Build GRCh38

View Full Product Details