Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 115408344-115408880 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRPS1
Entrez Gene ID
7227
Gene Name
transcriptional repressor GATA binding 1
Gene Aliases
GC79, LGCR
UniGene
Hs.657018
Gene Chromosome Location
Chr.8: 115408496-115669633 on Build GRCh38
Transcripts
NM_001282902.2 NM_001282903.2 NM_014112.4 XM_005251049.2 XM_006716625.1 XM_011517264.1 XM_011517266.2 XM_011517267.1 XM_011517268.1 XM_017013801.1

SNP Detail

SNP ID Chromosome Location
rs570380170 Chr.8: 115408488-115408488 on Build GRCh38
rs1010075659 Chr.8: 115408817-115408817 on Build GRCh38
rs1661215 Chr.8: 115408575-115408575 on Build GRCh38
rs534635252 Chr.8: 115408491-115408491 on Build GRCh38
rs971721097 Chr.8: 115408628-115408628 on Build GRCh38
rs575669844 Chr.8: 115408704-115408704 on Build GRCh38
rs562132571 Chr.8: 115408824-115408824 on Build GRCh38
rs749593830 Chr.8: 115408700-115408700 on Build GRCh38
rs564851438 Chr.8: 115408720-115408723 on Build GRCh38
rs905013630 Chr.8: 115408732-115408732 on Build GRCh38
rs994914569 Chr.8: 115408806-115408806 on Build GRCh38
rs936422952 Chr.8: 115408605-115408605 on Build GRCh38
rs10694243 Chr.8: 115408425-115408426 on Build GRCh38
rs374819663 Chr.8: 115408643-115408643 on Build GRCh38
rs557387204 Chr.8: 115408671-115408671 on Build GRCh38
rs899161357 Chr.8: 115408800-115408800 on Build GRCh38
rs546182400 Chr.8: 115408759-115408759 on Build GRCh38
rs980861147 Chr.8: 115408557-115408557 on Build GRCh38
rs950875992 Chr.8: 115408498-115408498 on Build GRCh38
rs945180054 Chr.8: 115408676-115408676 on Build GRCh38
rs117166324 Chr.8: 115408785-115408785 on Build GRCh38
rs970310280 Chr.8: 115408552-115408552 on Build GRCh38
rs540973740 Chr.8: 115408789-115408789 on Build GRCh38
rs552730781 Chr.8: 115408544-115408544 on Build GRCh38
rs890386943 Chr.8: 115408750-115408750 on Build GRCh38
rs940351283 Chr.8: 115408617-115408617 on Build GRCh38
rs141975722 Chr.8: 115408492-115408492 on Build GRCh38
rs576161054 Chr.8: 115408371-115408412 on Build GRCh38
rs186559093 Chr.8: 115408637-115408637 on Build GRCh38
rs987171019 Chr.8: 115408604-115408604 on Build GRCh38
rs993067291 Chr.8: 115408543-115408543 on Build GRCh38
rs545147903 Chr.8: 115408597-115408597 on Build GRCh38
rs1026342461 Chr.8: 115408811-115408811 on Build GRCh38
rs564503364 Chr.8: 115408774-115408774 on Build GRCh38
rs903863914 Chr.8: 115408836-115408836 on Build GRCh38
rs879688990 Chr.8: 115408652-115408652 on Build GRCh38

View Full Product Details