Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 117146709-117147219 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC30A8
Entrez Gene ID
169026
Gene Name
solute carrier family 30 member 8
Gene Aliases
ZNT8, ZnT-8
UniGene
Hs.532270
Gene Chromosome Location
Chr.8: 116950273-117176714 on Build GRCh38
Transcripts
NM_001172811.1 NM_001172813.1 NM_001172814.1 NM_001172815.2 NM_173851.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5290882 c.187G>A p.G63R Chr.8: 117147069-117147069 on Build GRCh38
25387 c.246C>T p.F82F Chr.8: 117147128-117147128 on Build GRCh38
3164912 c.233C>T p.S78L Chr.8: 117147115-117147115 on Build GRCh38
3164906 c.87G>A p.Q29Q Chr.8: 117146969-117146969 on Build GRCh38
1313664 c.133C>A p.L45M Chr.8: 117147015-117147015 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200895741 Chr.8: 117147078-117147078 on Build GRCh38
rs1008559239 Chr.8: 117146902-117146902 on Build GRCh38
rs927398701 Chr.8: 117147004-117147004 on Build GRCh38
rs746582624 Chr.8: 117147032-117147032 on Build GRCh38
rs550022625 Chr.8: 117146742-117146747 on Build GRCh38
rs759603427 Chr.8: 117146961-117146961 on Build GRCh38
rs760025849 Chr.8: 117147194-117147194 on Build GRCh38
rs751667454 Chr.8: 117146975-117146975 on Build GRCh38
rs765261676 Chr.8: 117146962-117146962 on Build GRCh38
rs144023942 Chr.8: 117146954-117146954 on Build GRCh38
rs541885243 Chr.8: 117146877-117146877 on Build GRCh38
rs773379065 Chr.8: 117147180-117147180 on Build GRCh38
rs780969938 Chr.8: 117147138-117147138 on Build GRCh38
rs141730422 Chr.8: 117147043-117147043 on Build GRCh38
rs1000322119 Chr.8: 117146869-117146869 on Build GRCh38
rs141876609 Chr.8: 117146991-117146991 on Build GRCh38
rs757937209 Chr.8: 117147105-117147105 on Build GRCh38
rs764209881 Chr.8: 117147159-117147162 on Build GRCh38
rs904302130 Chr.8: 117146804-117146804 on Build GRCh38
rs267601738 Chr.8: 117147012-117147012 on Build GRCh38
rs35343819 Chr.8: 117147177-117147177 on Build GRCh38
rs769900402 Chr.8: 117147057-117147057 on Build GRCh38
rs142695311 Chr.8: 117146742-117146743 on Build GRCh38
rs1018906209 Chr.8: 117146919-117146919 on Build GRCh38
rs752162751 Chr.8: 117146780-117146780 on Build GRCh38
rs757655293 Chr.8: 117146976-117146976 on Build GRCh38
rs774478941 Chr.8: 117147073-117147073 on Build GRCh38
rs1041883539 Chr.8: 117147057-117147057 on Build GRCh38
rs966893825 Chr.8: 117146771-117146771 on Build GRCh38
rs377269953 Chr.8: 117147044-117147044 on Build GRCh38
rs192056904 Chr.8: 117146801-117146801 on Build GRCh38
rs200572112 Chr.8: 117147084-117147084 on Build GRCh38
rs184329608 Chr.8: 117146888-117146888 on Build GRCh38
rs896066165 Chr.8: 117146764-117146765 on Build GRCh38
rs768636602 Chr.8: 117147154-117147154 on Build GRCh38
rs531568902 Chr.8: 117146743-117146743 on Build GRCh38
rs920239523 Chr.8: 117146887-117146887 on Build GRCh38
rs772687155 Chr.8: 117146943-117146943 on Build GRCh38
rs762011807 Chr.8: 117147074-117147074 on Build GRCh38
rs749244005 Chr.8: 117147150-117147150 on Build GRCh38
rs756658613 Chr.8: 117147000-117147000 on Build GRCh38
rs965303232 Chr.8: 117146816-117146816 on Build GRCh38
rs149521249 Chr.8: 117146952-117146952 on Build GRCh38
rs767831564 Chr.8: 117146982-117146982 on Build GRCh38
rs777483638 Chr.8: 117147024-117147024 on Build GRCh38
rs751975620 Chr.8: 117146913-117146913 on Build GRCh38
rs188831705 Chr.8: 117146908-117146908 on Build GRCh38
rs777205285 Chr.8: 117147122-117147122 on Build GRCh38
rs766175372 Chr.8: 117146925-117146927 on Build GRCh38
rs193096607 Chr.8: 117146912-117146912 on Build GRCh38
rs781725588 Chr.8: 117146916-117146916 on Build GRCh38
rs746849548 Chr.8: 117146951-117146951 on Build GRCh38
rs747629035 Chr.8: 117146941-117146941 on Build GRCh38
rs369417167 Chr.8: 117147068-117147068 on Build GRCh38
rs773052424 Chr.8: 117147141-117147141 on Build GRCh38
rs587777582 Chr.8: 117146983-117146989 on Build GRCh38
rs775528421 Chr.8: 117147061-117147061 on Build GRCh38
rs917503960 Chr.8: 117147040-117147040 on Build GRCh38
rs750839622 Chr.8: 117147083-117147083 on Build GRCh38
rs761027904 Chr.8: 117147181-117147181 on Build GRCh38
rs777302856 Chr.8: 117146974-117146974 on Build GRCh38
rs1021106085 Chr.8: 117146763-117146763 on Build GRCh38
rs746522664 Chr.8: 117147130-117147130 on Build GRCh38
rs926149634 Chr.8: 117147058-117147059 on Build GRCh38
rs1027359754 Chr.8: 117147172-117147172 on Build GRCh38
rs753710592 Chr.8: 117146950-117146950 on Build GRCh38
rs770275610 Chr.8: 117146917-117146917 on Build GRCh38
rs780519971 Chr.8: 117147006-117147006 on Build GRCh38
rs189800066 Chr.8: 117147102-117147102 on Build GRCh38
rs1027555145 Chr.8: 117147172-117147172 on Build GRCh38
rs140404252 Chr.8: 117147103-117147103 on Build GRCh38
rs1010924650 Chr.8: 117146742-117146742 on Build GRCh38
rs772307654 Chr.8: 117147177-117147177 on Build GRCh38
rs143592691 Chr.8: 117147014-117147014 on Build GRCh38
rs114650878 Chr.8: 117147149-117147149 on Build GRCh38
rs774338536 Chr.8: 117147167-117147167 on Build GRCh38
rs200523051 Chr.8: 117146918-117146918 on Build GRCh38
rs766668829 Chr.8: 117147190-117147190 on Build GRCh38
rs1028726439 Chr.8: 117146878-117146878 on Build GRCh38
rs754186005 Chr.8: 117147193-117147193 on Build GRCh38
rs148043363 Chr.8: 117147015-117147015 on Build GRCh38
rs761933921 Chr.8: 117147168-117147168 on Build GRCh38
rs769647577 Chr.8: 117147146-117147146 on Build GRCh38
rs757523939 Chr.8: 117146914-117146914 on Build GRCh38
rs763078920 Chr.8: 117146966-117146966 on Build GRCh38
rs1009601626 Chr.8: 117146773-117146773 on Build GRCh38
rs778566684 Chr.8: 117147117-117147117 on Build GRCh38
rs756898981 Chr.8: 117147134-117147134 on Build GRCh38
rs150548337 Chr.8: 117147070-117147070 on Build GRCh38
rs574773610 Chr.8: 117146850-117146850 on Build GRCh38
rs951805138 Chr.8: 117146950-117146950 on Build GRCh38
rs745627696 Chr.8: 117147053-117147053 on Build GRCh38
rs547844916 Chr.8: 117147088-117147088 on Build GRCh38
rs771801861 Chr.8: 117146942-117146942 on Build GRCh38
rs775751286 Chr.8: 117146965-117146965 on Build GRCh38
rs984636211 Chr.8: 117146994-117146994 on Build GRCh38

View Full Product Details