Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 120419569-120420090 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRPL13
Entrez Gene ID
28998
Gene Name
mitochondrial ribosomal protein L13
Gene Aliases
L13, L13A, L13mt, RPL13, RPML13
UniGene
Hs.333823
Gene Chromosome Location
Chr.8: 120395844-120445407 on Build GRCh38
Transcripts
NM_014078.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4365105 c.330C>G p.G110G Chr.8: 120419915-120419915 on Build GRCh38
6536632 c.363G>C p.M121I Chr.8: 120419882-120419882 on Build GRCh38
453937 c.370A>G p.R124G Chr.8: 120419875-120419875 on Build GRCh38
133492 c.375_376GC>TT p.L125_H126>FY Chr.8: 120419869-120419870 on Build GRCh38
1454575 c.334C>T p.L112L Chr.8: 120419911-120419911 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375244395 Chr.8: 120419802-120419802 on Build GRCh38
rs756007295 Chr.8: 120419947-120419947 on Build GRCh38
rs965329281 Chr.8: 120419912-120419912 on Build GRCh38
rs986100519 Chr.8: 120419622-120419622 on Build GRCh38
rs369757173 Chr.8: 120419836-120419836 on Build GRCh38
rs966752911 Chr.8: 120419799-120419799 on Build GRCh38
rs759937576 Chr.8: 120419873-120419873 on Build GRCh38
rs372015707 Chr.8: 120419892-120419892 on Build GRCh38
rs894217120 Chr.8: 120419794-120419794 on Build GRCh38
rs938967047 Chr.8: 120420012-120420012 on Build GRCh38
rs1057200613 Chr.8: 120420026-120420026 on Build GRCh38
rs373065613 Chr.8: 120419975-120419975 on Build GRCh38
rs929706689 Chr.8: 120419955-120419955 on Build GRCh38
rs889931592 Chr.8: 120419612-120419612 on Build GRCh38
rs754896480 Chr.8: 120419957-120419957 on Build GRCh38
rs751147932 Chr.8: 120419883-120419883 on Build GRCh38
rs952700719 Chr.8: 120419662-120419662 on Build GRCh38
rs201992737 Chr.8: 120419949-120419949 on Build GRCh38
rs1041155689 Chr.8: 120419924-120419924 on Build GRCh38
rs948434329 Chr.8: 120420043-120420043 on Build GRCh38
rs766832666 Chr.8: 120419937-120419937 on Build GRCh38
rs777234904 Chr.8: 120419870-120419870 on Build GRCh38
rs904308289 Chr.8: 120420051-120420051 on Build GRCh38
rs1050416372 Chr.8: 120419847-120419847 on Build GRCh38
rs1014105423 Chr.8: 120420053-120420053 on Build GRCh38
rs752031271 Chr.8: 120419969-120419969 on Build GRCh38
rs199947630 Chr.8: 120419984-120419984 on Build GRCh38
rs1040783830 Chr.8: 120419618-120419618 on Build GRCh38
rs1036624273 Chr.8: 120419657-120419657 on Build GRCh38
rs963402588 Chr.8: 120419999-120419999 on Build GRCh38
rs761081068 Chr.8: 120419844-120419844 on Build GRCh38
rs972512420 Chr.8: 120420005-120420005 on Build GRCh38
rs999147181 Chr.8: 120419868-120419868 on Build GRCh38
rs998004251 Chr.8: 120419938-120419938 on Build GRCh38
rs139492414 Chr.8: 120419600-120419600 on Build GRCh38
rs771212058 Chr.8: 120419825-120419826 on Build GRCh38
rs1034287302 Chr.8: 120420014-120420014 on Build GRCh38
rs771195899 Chr.8: 120419863-120419863 on Build GRCh38
rs372934824 Chr.8: 120419963-120419963 on Build GRCh38
rs978153521 Chr.8: 120419827-120419827 on Build GRCh38
rs1007959362 Chr.8: 120419616-120419616 on Build GRCh38
rs191897491 Chr.8: 120419602-120419602 on Build GRCh38
rs1001839555 Chr.8: 120419689-120419689 on Build GRCh38
rs765724004 Chr.8: 120419881-120419881 on Build GRCh38
rs919497975 Chr.8: 120420007-120420007 on Build GRCh38
rs778099312 Chr.8: 120420009-120420009 on Build GRCh38
rs998531272 Chr.8: 120420060-120420060 on Build GRCh38
rs776880336 Chr.8: 120419983-120419983 on Build GRCh38
rs201007256 Chr.8: 120419982-120419982 on Build GRCh38
rs942235070 Chr.8: 120419643-120419643 on Build GRCh38
rs114318580 Chr.8: 120419640-120419640 on Build GRCh38
rs1027031881 Chr.8: 120419650-120419650 on Build GRCh38
rs772283027 Chr.8: 120419830-120419830 on Build GRCh38
rs776969978 Chr.8: 120419986-120419987 on Build GRCh38
rs752027874 Chr.8: 120419696-120419696 on Build GRCh38
rs111327261 Chr.8: 120419709-120419709 on Build GRCh38
rs778033108 Chr.8: 120419981-120419981 on Build GRCh38
rs925270884 Chr.8: 120419833-120419833 on Build GRCh38
rs1042821818 Chr.8: 120420044-120420044 on Build GRCh38
rs750088534 Chr.8: 120419941-120419941 on Build GRCh38
rs1057254237 Chr.8: 120419770-120419770 on Build GRCh38
rs992639001 Chr.8: 120419715-120419715 on Build GRCh38
rs182607308 Chr.8: 120419670-120419670 on Build GRCh38
rs748182927 Chr.8: 120419820-120419820 on Build GRCh38
rs187600078 Chr.8: 120419953-120419953 on Build GRCh38
rs916097588 Chr.8: 120420029-120420029 on Build GRCh38
rs549063619 Chr.8: 120419634-120419634 on Build GRCh38
rs768824412 Chr.8: 120419801-120419801 on Build GRCh38

View Full Product Details