Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 129752027-129752528 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSDMC
Entrez Gene ID
56169
Gene Name
gasdermin C
Gene Aliases
MLZE
UniGene
Hs.133244
Gene Chromosome Location
Chr.8: 129705903-129786888 on Build GRCh38
Transcripts
NM_031415.2 XM_011517158.1 XM_011517159.1 XM_011517161.1 XM_011517162.1 XM_011517163.2 XM_011517165.1 XM_011517166.2 XM_017013644.1 XM_017013645.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3317165 c.846G>A p.E282E Chr.8: 129752146-129752146 on Build GRCh38
4689068 c.848T>C p.L283P Chr.8: 129752144-129752144 on Build GRCh38
1253856 c.855G>T p.L285L Chr.8: 129752137-129752137 on Build GRCh38
1166299 c.866T>A p.F289Y Chr.8: 129752126-129752126 on Build GRCh38
5815355 c.845-2A>C p.? Chr.8: 129752149-129752149 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs569668811 Chr.8: 129752453-129752453 on Build GRCh38
rs192784577 Chr.8: 129752238-129752238 on Build GRCh38
rs1042665947 Chr.8: 129752418-129752418 on Build GRCh38
rs779955236 Chr.8: 129752087-129752088 on Build GRCh38
rs927244579 Chr.8: 129752424-129752424 on Build GRCh38
rs557813389 Chr.8: 129752055-129752055 on Build GRCh38
rs373573200 Chr.8: 129752118-129752118 on Build GRCh38
rs529616613 Chr.8: 129752121-129752121 on Build GRCh38
rs549585319 Chr.8: 129752282-129752282 on Build GRCh38
rs550836420 Chr.8: 129752285-129752286 on Build GRCh38
rs984067292 Chr.8: 129752337-129752337 on Build GRCh38
rs771009961 Chr.8: 129752104-129752104 on Build GRCh38
rs775375617 Chr.8: 129752090-129752090 on Build GRCh38
rs535368761 Chr.8: 129752332-129752332 on Build GRCh38
rs908387438 Chr.8: 129752101-129752101 on Build GRCh38
rs763784301 Chr.8: 129752093-129752093 on Build GRCh38
rs768876220 Chr.8: 129752060-129752060 on Build GRCh38
rs976593505 Chr.8: 129752430-129752430 on Build GRCh38
rs765798312 Chr.8: 129752222-129752222 on Build GRCh38
rs547278750 Chr.8: 129752355-129752355 on Build GRCh38
rs142205452 Chr.8: 129752173-129752173 on Build GRCh38
rs923350795 Chr.8: 129752435-129752435 on Build GRCh38
rs576596981 Chr.8: 129752067-129752067 on Build GRCh38
rs780326163 Chr.8: 129752132-129752132 on Build GRCh38
rs368850633 Chr.8: 129752138-129752138 on Build GRCh38
rs940742878 Chr.8: 129752103-129752103 on Build GRCh38
rs746634014 Chr.8: 129752064-129752064 on Build GRCh38
rs903366913 Chr.8: 129752256-129752256 on Build GRCh38
rs759786214 Chr.8: 129752197-129752197 on Build GRCh38
rs937230215 Chr.8: 129752431-129752431 on Build GRCh38
rs759002109 Chr.8: 129752073-129752073 on Build GRCh38
rs187538653 Chr.8: 129752165-129752165 on Build GRCh38
rs1035442991 Chr.8: 129752268-129752268 on Build GRCh38
rs533357635 Chr.8: 129752200-129752200 on Build GRCh38
rs115758556 Chr.8: 129752119-129752119 on Build GRCh38
rs961360595 Chr.8: 129752149-129752149 on Build GRCh38
rs567864726 Chr.8: 129752308-129752308 on Build GRCh38
rs769326665 Chr.8: 129752086-129752086 on Build GRCh38
rs117752007 Chr.8: 129752396-129752396 on Build GRCh38
rs762998683 Chr.8: 129752092-129752092 on Build GRCh38
rs1043596678 Chr.8: 129752243-129752243 on Build GRCh38
rs752297890 Chr.8: 129752085-129752085 on Build GRCh38
rs952711652 Chr.8: 129752333-129752333 on Build GRCh38
rs551610357 Chr.8: 129752219-129752219 on Build GRCh38
rs912705528 Chr.8: 129752342-129752342 on Build GRCh38
rs374320142 Chr.8: 129752061-129752061 on Build GRCh38
rs753187259 Chr.8: 129752223-129752223 on Build GRCh38
rs984340981 Chr.8: 129752080-129752080 on Build GRCh38
rs770506910 Chr.8: 129752065-129752065 on Build GRCh38
rs912941003 Chr.8: 129752373-129752373 on Build GRCh38
rs367589545 Chr.8: 129752265-129752265 on Build GRCh38
rs763389545 Chr.8: 129752306-129752306 on Build GRCh38
rs755571989 Chr.8: 129752150-129752150 on Build GRCh38
rs749154750 Chr.8: 129752154-129752155 on Build GRCh38
rs751300382 Chr.8: 129752110-129752110 on Build GRCh38
rs933437052 Chr.8: 129752436-129752436 on Build GRCh38

View Full Product Details