Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 132713868-132714270 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM71
Entrez Gene ID
137835
Gene Name
transmembrane protein 71
Gene Aliases
-
UniGene
Hs.293842
Gene Chromosome Location
Chr.8: 132709945-132760668 on Build GRCh38
Transcripts
NM_001145153.1 NM_144649.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7085467 c.775C>G p.L259V Chr.8: 132714035-132714035 on Build GRCh38
5026974 c.737C>G p.S246* Chr.8: 132714174-132714174 on Build GRCh38
7151612 c.586G>A p.A196T Chr.8: 132714193-132714193 on Build GRCh38
7151611 c.718G>A p.A240T Chr.8: 132714193-132714193 on Build GRCh38
5036439 c.815+7G>A p.? Chr.8: 132713988-132713988 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756359681 Chr.8: 132714225-132714225 on Build GRCh38
rs751493294 Chr.8: 132714215-132714215 on Build GRCh38
rs749383675 Chr.8: 132714103-132714103 on Build GRCh38
rs930177013 Chr.8: 132714200-132714200 on Build GRCh38
rs757061127 Chr.8: 132714068-132714068 on Build GRCh38
rs772388856 Chr.8: 132714142-132714142 on Build GRCh38
rs1007059905 Chr.8: 132713941-132713941 on Build GRCh38
rs758631491 Chr.8: 132713906-132713906 on Build GRCh38
rs751399728 Chr.8: 132714059-132714059 on Build GRCh38
rs376231775 Chr.8: 132714245-132714245 on Build GRCh38
rs956550728 Chr.8: 132713963-132713963 on Build GRCh38
rs757149034 Chr.8: 132714216-132714216 on Build GRCh38
rs774926325 Chr.8: 132714178-132714178 on Build GRCh38
rs756326628 Chr.8: 132714095-132714095 on Build GRCh38
rs776093690 Chr.8: 132713973-132713973 on Build GRCh38
rs979222110 Chr.8: 132714121-132714121 on Build GRCh38
rs141415713 Chr.8: 132714051-132714051 on Build GRCh38
rs774786408 Chr.8: 132714118-132714118 on Build GRCh38
rs969330103 Chr.8: 132714042-132714042 on Build GRCh38
rs577561745 Chr.8: 132713996-132713996 on Build GRCh38
rs541314217 Chr.8: 132713962-132713962 on Build GRCh38
rs773536706 Chr.8: 132714143-132714143 on Build GRCh38
rs530960868 Chr.8: 132713960-132713960 on Build GRCh38
rs762099327 Chr.8: 132713994-132713994 on Build GRCh38
rs774646425 Chr.8: 132713982-132713982 on Build GRCh38
rs891367453 Chr.8: 132713908-132713908 on Build GRCh38
rs371187265 Chr.8: 132713951-132713951 on Build GRCh38
rs758322988 Chr.8: 132714005-132714005 on Build GRCh38
rs764696283 Chr.8: 132713998-132713998 on Build GRCh38
rs1007004799 Chr.8: 132713916-132713916 on Build GRCh38
rs918656231 Chr.8: 132714199-132714199 on Build GRCh38
rs888577901 Chr.8: 132713922-132713922 on Build GRCh38
rs879069905 Chr.8: 132714030-132714030 on Build GRCh38
rs991981174 Chr.8: 132714002-132714002 on Build GRCh38
rs927329648 Chr.8: 132714129-132714129 on Build GRCh38
rs769712671 Chr.8: 132713966-132713966 on Build GRCh38
rs199598812 Chr.8: 132714238-132714238 on Build GRCh38
rs746046657 Chr.8: 132714082-132714082 on Build GRCh38
rs746427898 Chr.8: 132714220-132714222 on Build GRCh38
rs750349298 Chr.8: 132714153-132714153 on Build GRCh38
rs771342225 Chr.8: 132713957-132713957 on Build GRCh38
rs769194437 Chr.8: 132713981-132713981 on Build GRCh38
rs764925851 Chr.8: 132714161-132714161 on Build GRCh38
rs781681078 Chr.8: 132714161-132714161 on Build GRCh38
rs201486946 Chr.8: 132714043-132714043 on Build GRCh38
rs752540333 Chr.8: 132713944-132713944 on Build GRCh38
rs780257469 Chr.8: 132714233-132714233 on Build GRCh38
rs746783659 Chr.8: 132713955-132713955 on Build GRCh38
rs750231882 Chr.8: 132714222-132714222 on Build GRCh38
rs752226498 Chr.8: 132714001-132714001 on Build GRCh38
rs769047968 Chr.8: 132714107-132714107 on Build GRCh38
rs113178295 Chr.8: 132713995-132713995 on Build GRCh38
rs758323309 Chr.8: 132713949-132713949 on Build GRCh38
rs770170104 Chr.8: 132714236-132714236 on Build GRCh38
rs199912655 Chr.8: 132714147-132714147 on Build GRCh38
rs372197272 Chr.8: 132714206-132714206 on Build GRCh38
rs781587143 Chr.8: 132714069-132714069 on Build GRCh38
rs917861773 Chr.8: 132714025-132714025 on Build GRCh38
rs559111461 Chr.8: 132713979-132713979 on Build GRCh38
rs1017944293 Chr.8: 132713917-132713917 on Build GRCh38
rs767458247 Chr.8: 132714217-132714217 on Build GRCh38
rs762454398 Chr.8: 132714189-132714189 on Build GRCh38
rs191128883 Chr.8: 132714119-132714119 on Build GRCh38

View Full Product Details