Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 134477710-134478212 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFAT
Entrez Gene ID
57623
Gene Name
zinc finger and AT-hook domain containing
Gene Aliases
AITD3, ZFAT1, ZNF406
UniGene
Hs.446172
Gene Chromosome Location
Chr.8: 134477788-134713049 on Build GRCh38
Transcripts
NM_001029939.3 NM_001167583.2 NM_001174157.1 NM_001174158.1 NM_001289394.1 NM_020863.3 NR_110323.1 XM_011517203.1 XM_011517204.2

SNP Detail

SNP ID Chromosome Location
rs920856209 Chr.8: 134478113-134478113 on Build GRCh38
rs987232235 Chr.8: 134478087-134478087 on Build GRCh38
rs1033577163 Chr.8: 134478008-134478008 on Build GRCh38
rs1002160460 Chr.8: 134478004-134478004 on Build GRCh38
rs149312401 Chr.8: 134478185-134478185 on Build GRCh38
rs145343393 Chr.8: 134477872-134477874 on Build GRCh38
rs780513342 Chr.8: 134477996-134477996 on Build GRCh38
rs544343243 Chr.8: 134478148-134478148 on Build GRCh38
rs540627418 Chr.8: 134478189-134478189 on Build GRCh38
rs780278598 Chr.8: 134477842-134477842 on Build GRCh38
rs892104528 Chr.8: 134477769-134477769 on Build GRCh38
rs144769555 Chr.8: 134477939-134477939 on Build GRCh38
rs994972769 Chr.8: 134477741-134477741 on Build GRCh38
rs116877116 Chr.8: 134478098-134478098 on Build GRCh38
rs1039186744 Chr.8: 134478002-134478002 on Build GRCh38
rs538536188 Chr.8: 134477985-134477985 on Build GRCh38
rs899198305 Chr.8: 134477974-134477974 on Build GRCh38
rs571492563 Chr.8: 134477950-134477950 on Build GRCh38
rs933770383 Chr.8: 134478058-134478058 on Build GRCh38
rs868823218 Chr.8: 134477953-134477953 on Build GRCh38
rs903761476 Chr.8: 134478110-134478110 on Build GRCh38
rs558458951 Chr.8: 134478104-134478104 on Build GRCh38
rs761761082 Chr.8: 134477893-134477893 on Build GRCh38
rs188846777 Chr.8: 134477890-134477890 on Build GRCh38
rs550929407 Chr.8: 134478057-134478057 on Build GRCh38
rs768238275 Chr.8: 134477841-134477841 on Build GRCh38
rs765312527 Chr.8: 134478080-134478080 on Build GRCh38
rs950654387 Chr.8: 134477952-134477952 on Build GRCh38
rs974997465 Chr.8: 134478107-134478107 on Build GRCh38
rs921043346 Chr.8: 134478028-134478028 on Build GRCh38
rs550674444 Chr.8: 134477757-134477757 on Build GRCh38
rs762640501 Chr.8: 134477767-134477767 on Build GRCh38
rs995231154 Chr.8: 134477979-134477979 on Build GRCh38
rs909458371 Chr.8: 134477972-134477972 on Build GRCh38
rs1009457571 Chr.8: 134477877-134477877 on Build GRCh38
rs951272014 Chr.8: 134477762-134477762 on Build GRCh38
rs575416207 Chr.8: 134478108-134478108 on Build GRCh38
rs751768763 Chr.8: 134477853-134477853 on Build GRCh38
rs180881285 Chr.8: 134477913-134477913 on Build GRCh38
rs1033529884 Chr.8: 134478176-134478176 on Build GRCh38
rs913654942 Chr.8: 134477894-134477894 on Build GRCh38
rs138616859 Chr.8: 134478149-134478149 on Build GRCh38
rs1039260943 Chr.8: 134477971-134477971 on Build GRCh38
rs1024820381 Chr.8: 134477996-134477996 on Build GRCh38
rs767435834 Chr.8: 134477897-134477897 on Build GRCh38
rs992621755 Chr.8: 134477943-134477943 on Build GRCh38
rs970634745 Chr.8: 134477857-134477857 on Build GRCh38
rs369267280 Chr.8: 134478084-134478084 on Build GRCh38
rs980648871 Chr.8: 134477861-134477861 on Build GRCh38

View Full Product Details