Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 139730569-139731063 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRAPPC9
Entrez Gene ID
83696
Gene Name
trafficking protein particle complex 9
Gene Aliases
IBP, IKBKBBP, MRT13, NIBP, T1, TRS120
UniGene
Hs.654911
Gene Chromosome Location
Chr.8: 139727725-140458579 on Build GRCh38
Transcripts
NM_001160372.3 NM_001321646.1 NM_031466.7 XM_011517326.2 XM_011517330.2 XM_017013894.1

SNP Detail

SNP ID Chromosome Location
rs1018027902 Chr.8: 139730907-139730907 on Build GRCh38
rs184686731 Chr.8: 139730646-139730646 on Build GRCh38
rs886062721 Chr.8: 139730981-139730981 on Build GRCh38
rs897310917 Chr.8: 139730892-139730892 on Build GRCh38
rs201138637 Chr.8: 139730824-139730824 on Build GRCh38
rs759094680 Chr.8: 139731036-139731036 on Build GRCh38
rs577059363 Chr.8: 139730998-139730998 on Build GRCh38
rs752932738 Chr.8: 139730863-139730863 on Build GRCh38
rs771052277 Chr.8: 139731029-139731029 on Build GRCh38
rs982315273 Chr.8: 139730962-139730962 on Build GRCh38
rs903241993 Chr.8: 139730589-139730589 on Build GRCh38
rs934728624 Chr.8: 139730628-139730628 on Build GRCh38
rs928228874 Chr.8: 139731002-139731002 on Build GRCh38
rs773182942 Chr.8: 139731016-139731016 on Build GRCh38
rs554259530 Chr.8: 139730882-139730882 on Build GRCh38
rs117443048 Chr.8: 139730896-139730896 on Build GRCh38
rs369659784 Chr.8: 139730693-139730693 on Build GRCh38
rs1042842144 Chr.8: 139730878-139730878 on Build GRCh38
rs756337357 Chr.8: 139730865-139730865 on Build GRCh38
rs924587062 Chr.8: 139730981-139730981 on Build GRCh38
rs747820750 Chr.8: 139730922-139730951 on Build GRCh38
rs951660340 Chr.8: 139730791-139730791 on Build GRCh38
rs954824682 Chr.8: 139730891-139730891 on Build GRCh38
rs571129282 Chr.8: 139730809-139730809 on Build GRCh38
rs910631551 Chr.8: 139730710-139730710 on Build GRCh38
rs963474372 Chr.8: 139730924-139730924 on Build GRCh38
rs928227743 Chr.8: 139730833-139730833 on Build GRCh38
rs776226146 Chr.8: 139731031-139731031 on Build GRCh38
rs1008327592 Chr.8: 139730735-139730735 on Build GRCh38
rs192973260 Chr.8: 139730626-139730626 on Build GRCh38
rs574925920 Chr.8: 139730997-139730997 on Build GRCh38
rs559828056 Chr.8: 139730692-139730692 on Build GRCh38
rs149208078 Chr.8: 139730940-139730940 on Build GRCh38
rs1027369612 Chr.8: 139730900-139730900 on Build GRCh38
rs1002980133 Chr.8: 139730613-139730613 on Build GRCh38
rs548729054 Chr.8: 139730730-139730730 on Build GRCh38
rs199226 Chr.8: 139730957-139730957 on Build GRCh38
rs552629146 Chr.8: 139730879-139730879 on Build GRCh38
rs527976463 Chr.8: 139730829-139730829 on Build GRCh38
rs531153681 Chr.8: 139730793-139730793 on Build GRCh38
rs973530941 Chr.8: 139730733-139730733 on Build GRCh38
rs1016186923 Chr.8: 139730738-139730738 on Build GRCh38
rs991157935 Chr.8: 139730792-139730792 on Build GRCh38
rs949964528 Chr.8: 139730814-139730814 on Build GRCh38
rs771971831 Chr.8: 139731013-139731013 on Build GRCh38
rs577116244 Chr.8: 139730685-139730685 on Build GRCh38
rs894189907 Chr.8: 139730862-139730862 on Build GRCh38
rs552555971 Chr.8: 139730794-139730794 on Build GRCh38
rs1055901450 Chr.8: 139730857-139730857 on Build GRCh38
rs996838896 Chr.8: 139730674-139730674 on Build GRCh38
rs191976492 Chr.8: 139731003-139731003 on Build GRCh38
rs190303349 Chr.8: 139730776-139730776 on Build GRCh38
rs565362144 Chr.8: 139730880-139730880 on Build GRCh38
rs781519625 Chr.8: 139730670-139730670 on Build GRCh38
rs891380051 Chr.8: 139730697-139730697 on Build GRCh38
rs554930360 Chr.8: 139730669-139730669 on Build GRCh38
rs1012559616 Chr.8: 139730921-139730921 on Build GRCh38
rs893228404 Chr.8: 139730854-139730854 on Build GRCh38
rs754385217 Chr.8: 139730772-139730772 on Build GRCh38
rs373284152 Chr.8: 139730996-139730996 on Build GRCh38
rs530201993 Chr.8: 139730729-139730729 on Build GRCh38
rs1021421830 Chr.8: 139730679-139730679 on Build GRCh38
rs1015868475 Chr.8: 139730886-139730886 on Build GRCh38
rs1030235716 Chr.8: 139730688-139730688 on Build GRCh38
rs919441234 Chr.8: 139730750-139730750 on Build GRCh38
rs971077657 Chr.8: 139730936-139730936 on Build GRCh38
rs746941093 Chr.8: 139731028-139731028 on Build GRCh38
rs1030204609 Chr.8: 139730883-139730883 on Build GRCh38
rs546972114 Chr.8: 139730853-139730853 on Build GRCh38
rs535912662 Chr.8: 139730881-139730881 on Build GRCh38
rs973495008 Chr.8: 139730929-139730929 on Build GRCh38
rs79408026 Chr.8: 139730686-139730686 on Build GRCh38
rs576206788 Chr.8: 139730641-139730641 on Build GRCh38

View Full Product Details