Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 143021810-143022313 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LY6E
Entrez Gene ID
4061
Gene Name
lymphocyte antigen 6 complex, locus E
Gene Aliases
RIG-E, RIGE, SCA-2, SCA2, TSA-1
UniGene
Hs.521903
Gene Chromosome Location
Chr.8: 143018485-143022410 on Build GRCh38
Transcripts
NM_001127213.1 NM_002346.2

SNP Detail

SNP ID Chromosome Location
rs770998009 Chr.8: 143022165-143022165 on Build GRCh38
rs576592915 Chr.8: 143022232-143022232 on Build GRCh38
rs909642958 Chr.8: 143022229-143022229 on Build GRCh38
rs11547124 Chr.8: 143022050-143022050 on Build GRCh38
rs886455159 Chr.8: 143021907-143021907 on Build GRCh38
rs963394544 Chr.8: 143021948-143021948 on Build GRCh38
rs192838481 Chr.8: 143021883-143021883 on Build GRCh38
rs966372074 Chr.8: 143022106-143022106 on Build GRCh38
rs139919223 Chr.8: 143022107-143022107 on Build GRCh38
rs376994588 Chr.8: 143022170-143022170 on Build GRCh38
rs978536398 Chr.8: 143022061-143022061 on Build GRCh38
rs113719829 Chr.8: 143021945-143021945 on Build GRCh38
rs879270452 Chr.8: 143022283-143022283 on Build GRCh38
rs745674413 Chr.8: 143021837-143021837 on Build GRCh38
rs930814511 Chr.8: 143022187-143022187 on Build GRCh38
rs185330105 Chr.8: 143021967-143021967 on Build GRCh38
rs1049217540 Chr.8: 143022212-143022212 on Build GRCh38
rs1802886 Chr.8: 143022220-143022220 on Build GRCh38
rs370125043 Chr.8: 143022268-143022268 on Build GRCh38
rs543973681 Chr.8: 143022273-143022273 on Build GRCh38
rs555035621 Chr.8: 143022155-143022155 on Build GRCh38
rs1048185054 Chr.8: 143021931-143021931 on Build GRCh38
rs917820304 Chr.8: 143022051-143022051 on Build GRCh38
rs933474274 Chr.8: 143022176-143022176 on Build GRCh38
rs886653005 Chr.8: 143022191-143022191 on Build GRCh38
rs1014616365 Chr.8: 143022291-143022291 on Build GRCh38
rs1052464322 Chr.8: 143022178-143022178 on Build GRCh38
rs554215687 Chr.8: 143022021-143022021 on Build GRCh38
rs974977345 Chr.8: 143022166-143022166 on Build GRCh38
rs922121784 Chr.8: 143022173-143022173 on Build GRCh38
rs888170980 Chr.8: 143021973-143021973 on Build GRCh38
rs1014171566 Chr.8: 143022030-143022030 on Build GRCh38
rs4464948 Chr.8: 143022018-143022018 on Build GRCh38
rs932416342 Chr.8: 143021917-143021917 on Build GRCh38
rs1039648872 Chr.8: 143021892-143021892 on Build GRCh38
rs900653219 Chr.8: 143021910-143021910 on Build GRCh38
rs988805298 Chr.8: 143021858-143021858 on Build GRCh38
rs998200100 Chr.8: 143021867-143021875 on Build GRCh38
rs969853093 Chr.8: 143022052-143022052 on Build GRCh38
rs4478552 Chr.8: 143022053-143022053 on Build GRCh38
rs1004845844 Chr.8: 143022257-143022257 on Build GRCh38
rs922148579 Chr.8: 143022171-143022171 on Build GRCh38
rs939780698 Chr.8: 143022233-143022233 on Build GRCh38
rs749528517 Chr.8: 143022156-143022156 on Build GRCh38
rs188373137 Chr.8: 143021987-143021987 on Build GRCh38
rs537237172 Chr.8: 143021889-143021889 on Build GRCh38
rs956114177 Chr.8: 143022114-143022114 on Build GRCh38
rs777709631 Chr.8: 143022138-143022138 on Build GRCh38

View Full Product Details