Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 143252989-143253487 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFP41
Entrez Gene ID
286128
Gene Name
ZFP41 zinc finger protein
Gene Aliases
ZNF753, zfp-41
UniGene
Hs.668016
Gene Chromosome Location
Chr.8: 143246821-143262705 on Build GRCh38
Transcripts
NM_001271156.2 NM_173832.5

SNP Detail

SNP ID Chromosome Location
rs368403343 Chr.8: 143253295-143253295 on Build GRCh38
rs909270799 Chr.8: 143253234-143253234 on Build GRCh38
rs976763017 Chr.8: 143253363-143253363 on Build GRCh38
rs78411586 Chr.8: 143253199-143253199 on Build GRCh38
rs909303423 Chr.8: 143253409-143253409 on Build GRCh38
rs559803070 Chr.8: 143253338-143253338 on Build GRCh38
rs528306565 Chr.8: 143253220-143253220 on Build GRCh38
rs1020496379 Chr.8: 143253356-143253356 on Build GRCh38
rs994097259 Chr.8: 143253101-143253101 on Build GRCh38
rs967557725 Chr.8: 143253361-143253361 on Build GRCh38
rs772924082 Chr.8: 143253164-143253175 on Build GRCh38
rs932091465 Chr.8: 143253048-143253048 on Build GRCh38
rs145787498 Chr.8: 143253063-143253063 on Build GRCh38
rs906882692 Chr.8: 143253288-143253288 on Build GRCh38
rs1050518302 Chr.8: 143253062-143253062 on Build GRCh38
rs995885660 Chr.8: 143253089-143253089 on Build GRCh38
rs1024425892 Chr.8: 143253110-143253110 on Build GRCh38
rs1050274539 Chr.8: 143253302-143253302 on Build GRCh38
rs923292200 Chr.8: 143253019-143253019 on Build GRCh38
rs774547815 Chr.8: 143253266-143253266 on Build GRCh38
rs890188980 Chr.8: 143253320-143253320 on Build GRCh38
rs773402408 Chr.8: 143253222-143253222 on Build GRCh38
rs56330502 Chr.8: 143253158-143253160 on Build GRCh38
rs370315201 Chr.8: 143253114-143253114 on Build GRCh38
rs76074354 Chr.8: 143253151-143253151 on Build GRCh38
rs535222332 Chr.8: 143253456-143253456 on Build GRCh38
rs994521044 Chr.8: 143253280-143253280 on Build GRCh38
rs957072933 Chr.8: 143253301-143253301 on Build GRCh38
rs192917498 Chr.8: 143253428-143253428 on Build GRCh38
rs188469060 Chr.8: 143253254-143253254 on Build GRCh38
rs777140483 Chr.8: 143253158-143253172 on Build GRCh38
rs55900712 Chr.8: 143253162-143253174 on Build GRCh38
rs72695734 Chr.8: 143253174-143253174 on Build GRCh38
rs747108446 Chr.8: 143253162-143253176 on Build GRCh38
rs1034109263 Chr.8: 143253296-143253296 on Build GRCh38
rs570090534 Chr.8: 143253400-143253400 on Build GRCh38
rs976351698 Chr.8: 143253202-143253202 on Build GRCh38
rs1008682127 Chr.8: 143253323-143253323 on Build GRCh38
rs767362725 Chr.8: 143253158-143253174 on Build GRCh38
rs116826422 Chr.8: 143253438-143253438 on Build GRCh38
rs67048485 Chr.8: 143253136-143253151 on Build GRCh38
rs1038264357 Chr.8: 143253091-143253091 on Build GRCh38
rs761900256 Chr.8: 143253158-143253165 on Build GRCh38
rs1053674751 Chr.8: 143253435-143253435 on Build GRCh38
rs28693781 Chr.8: 143253365-143253365 on Build GRCh38
rs917451205 Chr.8: 143253385-143253385 on Build GRCh38
rs958843284 Chr.8: 143253373-143253373 on Build GRCh38
rs1011330008 Chr.8: 143253306-143253306 on Build GRCh38
rs759607190 Chr.8: 143253387-143253387 on Build GRCh38
rs767764022 Chr.8: 143253405-143253405 on Build GRCh38
rs531213903 Chr.8: 143253422-143253422 on Build GRCh38
rs991597269 Chr.8: 143253378-143253378 on Build GRCh38
rs377590236 Chr.8: 143253190-143253190 on Build GRCh38
rs771283780 Chr.8: 143253265-143253265 on Build GRCh38
rs973667066 Chr.8: 143253176-143253176 on Build GRCh38
rs185635790 Chr.8: 143253198-143253198 on Build GRCh38
rs148980166 Chr.8: 143253204-143253204 on Build GRCh38
rs1040735815 Chr.8: 143253017-143253017 on Build GRCh38
rs977571313 Chr.8: 143253413-143253413 on Build GRCh38
rs1005317555 Chr.8: 143253078-143253078 on Build GRCh38
rs534039264 Chr.8: 143253441-143253441 on Build GRCh38
rs531293363 Chr.8: 143253118-143253118 on Build GRCh38
rs541818631 Chr.8: 143253065-143253065 on Build GRCh38
rs897043359 Chr.8: 143253094-143253094 on Build GRCh38
rs775098686 Chr.8: 143253151-143253152 on Build GRCh38
rs548597190 Chr.8: 143253396-143253396 on Build GRCh38
rs58935399 Chr.8: 143253164-143253179 on Build GRCh38
rs1042096680 Chr.8: 143253231-143253231 on Build GRCh38
rs971074886 Chr.8: 143253116-143253116 on Build GRCh38
rs942153792 Chr.8: 143253416-143253416 on Build GRCh38
rs903216022 Chr.8: 143253238-143253238 on Build GRCh38
rs748461665 Chr.8: 143253203-143253203 on Build GRCh38
rs990285967 Chr.8: 143253188-143253188 on Build GRCh38
rs896986091 Chr.8: 143253269-143253269 on Build GRCh38
rs2272632 Chr.8: 143253158-143253158 on Build GRCh38
rs1045634181 Chr.8: 143253285-143253285 on Build GRCh38
rs2272631 Chr.8: 143253125-143253125 on Build GRCh38

View Full Product Details