Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 143325242-143325738 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TOP1MT
Entrez Gene ID
116447
Gene Name
topoisomerase (DNA) I, mitochondrial
Gene Aliases
-
UniGene
Hs.528574
Gene Chromosome Location
Chr.8: 143309324-143359977 on Build GRCh38
Transcripts
NM_001258446.1 NM_001258447.1 NM_052963.2 XM_005250781.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3951398 c.567C>G p.F189L Chr.8: 143325450-143325450 on Build GRCh38
1455316 c.538G>C p.D180H Chr.8: 143325479-143325479 on Build GRCh38
5472513 c.641C>T p.T214M Chr.8: 143325376-143325376 on Build GRCh38
1096929 c.550G>T p.E184* Chr.8: 143325467-143325467 on Build GRCh38
1319751 c.512A>G p.Q171R Chr.8: 143325505-143325505 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1005462155 Chr.8: 143325542-143325542 on Build GRCh38
rs774091536 Chr.8: 143325544-143325544 on Build GRCh38
rs747614398 Chr.8: 143325297-143325297 on Build GRCh38
rs775455546 Chr.8: 143325471-143325471 on Build GRCh38
rs372077707 Chr.8: 143325679-143325679 on Build GRCh38
rs753758457 Chr.8: 143325330-143325330 on Build GRCh38
rs371662022 Chr.8: 143325376-143325376 on Build GRCh38
rs765109086 Chr.8: 143325332-143325332 on Build GRCh38
rs558706090 Chr.8: 143325718-143325718 on Build GRCh38
rs56021206 Chr.8: 143325691-143325691 on Build GRCh38
rs73382105 Chr.8: 143325309-143325309 on Build GRCh38
rs769617980 Chr.8: 143325520-143325520 on Build GRCh38
rs747381119 Chr.8: 143325305-143325305 on Build GRCh38
rs760430103 Chr.8: 143325558-143325558 on Build GRCh38
rs139852014 Chr.8: 143325609-143325609 on Build GRCh38
rs746182073 Chr.8: 143325344-143325344 on Build GRCh38
rs780652970 Chr.8: 143325360-143325360 on Build GRCh38
rs143457706 Chr.8: 143325414-143325414 on Build GRCh38
rs201148475 Chr.8: 143325450-143325450 on Build GRCh38
rs759444586 Chr.8: 143325469-143325469 on Build GRCh38
rs754267323 Chr.8: 143325568-143325568 on Build GRCh38
rs148068470 Chr.8: 143325425-143325425 on Build GRCh38
rs566350770 Chr.8: 143325625-143325625 on Build GRCh38
rs954616171 Chr.8: 143325717-143325717 on Build GRCh38
rs754214288 Chr.8: 143325494-143325494 on Build GRCh38
rs369984830 Chr.8: 143325495-143325495 on Build GRCh38
rs889732323 Chr.8: 143325528-143325528 on Build GRCh38
rs146761712 Chr.8: 143325419-143325419 on Build GRCh38
rs773236034 Chr.8: 143325324-143325325 on Build GRCh38
rs1028734202 Chr.8: 143325710-143325710 on Build GRCh38
rs988122036 Chr.8: 143325448-143325448 on Build GRCh38
rs149312505 Chr.8: 143325301-143325301 on Build GRCh38
rs754973863 Chr.8: 143325331-143325331 on Build GRCh38
rs368776212 Chr.8: 143325635-143325635 on Build GRCh38
rs750815514 Chr.8: 143325437-143325437 on Build GRCh38
rs768174377 Chr.8: 143325403-143325403 on Build GRCh38
rs773238695 Chr.8: 143325468-143325468 on Build GRCh38
rs368971997 Chr.8: 143325541-143325541 on Build GRCh38
rs770449162 Chr.8: 143325348-143325348 on Build GRCh38
rs971506466 Chr.8: 143325404-143325404 on Build GRCh38
rs199605994 Chr.8: 143325401-143325401 on Build GRCh38
rs111874912 Chr.8: 143325320-143325320 on Build GRCh38
rs762846413 Chr.8: 143325478-143325478 on Build GRCh38
rs774708072 Chr.8: 143325321-143325321 on Build GRCh38
rs564823146 Chr.8: 143325334-143325334 on Build GRCh38
rs368980736 Chr.8: 143325375-143325375 on Build GRCh38
rs758605036 Chr.8: 143325576-143325577 on Build GRCh38
rs780530783 Chr.8: 143325493-143325493 on Build GRCh38
rs765162909 Chr.8: 143325480-143325481 on Build GRCh38
rs749786463 Chr.8: 143325442-143325442 on Build GRCh38
rs199525522 Chr.8: 143325299-143325299 on Build GRCh38
rs200673353 Chr.8: 143325514-143325514 on Build GRCh38
rs746507403 Chr.8: 143325382-143325382 on Build GRCh38
rs771325540 Chr.8: 143325306-143325306 on Build GRCh38
rs995879654 Chr.8: 143325695-143325695 on Build GRCh38
rs777676495 Chr.8: 143325595-143325595 on Build GRCh38
rs750028820 Chr.8: 143325488-143325488 on Build GRCh38
rs775299797 Chr.8: 143325527-143325527 on Build GRCh38
rs368057294 Chr.8: 143325438-143325438 on Build GRCh38
rs144026151 Chr.8: 143325356-143325356 on Build GRCh38
rs780663716 Chr.8: 143325429-143325429 on Build GRCh38
rs766079725 Chr.8: 143325560-143325560 on Build GRCh38
rs763983351 Chr.8: 143325481-143325481 on Build GRCh38
rs752976515 Chr.8: 143325520-143325522 on Build GRCh38
rs79138102 Chr.8: 143325505-143325505 on Build GRCh38
rs921617030 Chr.8: 143325298-143325298 on Build GRCh38
rs547418113 Chr.8: 143325623-143325623 on Build GRCh38
rs761441238 Chr.8: 143325550-143325550 on Build GRCh38
rs772351362 Chr.8: 143325462-143325462 on Build GRCh38
rs777470270 Chr.8: 143325336-143325336 on Build GRCh38
rs766170787 Chr.8: 143325329-143325329 on Build GRCh38
rs373786310 Chr.8: 143325342-143325342 on Build GRCh38
rs765838104 Chr.8: 143325583-143325583 on Build GRCh38
rs777066572 Chr.8: 143325307-143325307 on Build GRCh38
rs139942348 Chr.8: 143325365-143325365 on Build GRCh38
rs1040448132 Chr.8: 143325459-143325459 on Build GRCh38
rs759192844 Chr.8: 143325413-143325413 on Build GRCh38
rs763080855 Chr.8: 143325316-143325316 on Build GRCh38
rs576566148 Chr.8: 143325699-143325699 on Build GRCh38
rs372600591 Chr.8: 143325552-143325552 on Build GRCh38
rs367544690 Chr.8: 143325368-143325368 on Build GRCh38
rs752627609 Chr.8: 143325416-143325416 on Build GRCh38
rs199593820 Chr.8: 143325579-143325579 on Build GRCh38
rs189443826 Chr.8: 143325556-143325556 on Build GRCh38
rs747779526 Chr.8: 143325439-143325439 on Build GRCh38
rs990377294 Chr.8: 143325707-143325707 on Build GRCh38
rs758405400 Chr.8: 143325504-143325504 on Build GRCh38
rs113458638 Chr.8: 143325603-143325603 on Build GRCh38
rs763505417 Chr.8: 143325673-143325673 on Build GRCh38
rs375507955 Chr.8: 143325489-143325489 on Build GRCh38
rs112832992 Chr.8: 143325412-143325412 on Build GRCh38
rs376583579 Chr.8: 143325503-143325503 on Build GRCh38
rs372542090 Chr.8: 143325445-143325445 on Build GRCh38
rs759693841 Chr.8: 143325347-143325348 on Build GRCh38
rs143596489 Chr.8: 143325341-143325341 on Build GRCh38
rs762508243 Chr.8: 143325418-143325418 on Build GRCh38
rs772144521 Chr.8: 143325394-143325394 on Build GRCh38
rs751106254 Chr.8: 143325482-143325482 on Build GRCh38
rs760440859 Chr.8: 143325325-143325325 on Build GRCh38
rs758398521 Chr.8: 143325335-143325335 on Build GRCh38
rs957896411 Chr.8: 143325703-143325703 on Build GRCh38
rs946116286 Chr.8: 143325453-143325453 on Build GRCh38
rs75866457 Chr.8: 143325711-143325711 on Build GRCh38
rs749037123 Chr.8: 143325538-143325538 on Build GRCh38
rs761173780 Chr.8: 143325484-143325484 on Build GRCh38
rs767207040 Chr.8: 143325487-143325487 on Build GRCh38
rs559664578 Chr.8: 143325470-143325470 on Build GRCh38
rs977914581 Chr.8: 143325378-143325378 on Build GRCh38
rs993877718 Chr.8: 143325379-143325379 on Build GRCh38
rs180984779 Chr.8: 143325594-143325594 on Build GRCh38
rs372639416 Chr.8: 143325498-143325498 on Build GRCh38
rs751215094 Chr.8: 143325424-143325424 on Build GRCh38
rs779080856 Chr.8: 143325449-143325449 on Build GRCh38
rs769805238 Chr.8: 143325310-143325310 on Build GRCh38

View Full Product Details