Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 14085972-14086472 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NFIB
Entrez Gene ID
4781
Gene Name
nuclear factor I B
Gene Aliases
CTF, HMGIC/NFIB, NF-I/B, NF1-B, NFI-B, NFI-RED, NFIB2, NFIB3
UniGene
Hs.644095
Gene Chromosome Location
Chr.9: 14081843-14398983 on Build GRCh38
Transcripts
NM_001190737.1 NM_001190738.1 NM_001282787.1 NM_005596.3 XM_005251467.2 XM_005251468.4 XM_005251469.2 XM_005251470.2 XM_005251471.3 XM_006716773.2 XM_006716774.2 XM_006716775.2 XM_017014739.1 XM_017014740.1 XM_017014741.1 XM_017014742.1

SNP Detail

SNP ID Chromosome Location
rs553000748 Chr.9: 14086135-14086135 on Build GRCh38
rs963920768 Chr.9: 14086423-14086423 on Build GRCh38
rs1015521955 Chr.9: 14086144-14086144 on Build GRCh38
rs890100127 Chr.9: 14086380-14086384 on Build GRCh38
rs898025144 Chr.9: 14086238-14086238 on Build GRCh38
rs982064840 Chr.9: 14086248-14086249 on Build GRCh38
rs1021515887 Chr.9: 14086019-14086019 on Build GRCh38
rs889164564 Chr.9: 14086326-14086326 on Build GRCh38
rs35934119 Chr.9: 14086383-14086384 on Build GRCh38
rs763493412 Chr.9: 14086041-14086041 on Build GRCh38
rs559306422 Chr.9: 14086258-14086258 on Build GRCh38
rs754610381 Chr.9: 14086166-14086166 on Build GRCh38
rs1019279864 Chr.9: 14086035-14086035 on Build GRCh38
rs773708024 Chr.9: 14086013-14086013 on Build GRCh38
rs916828464 Chr.9: 14086162-14086162 on Build GRCh38
rs891628016 Chr.9: 14086280-14086281 on Build GRCh38
rs966030996 Chr.9: 14086411-14086411 on Build GRCh38
rs947162253 Chr.9: 14086288-14086288 on Build GRCh38
rs554908634 Chr.9: 14086362-14086362 on Build GRCh38
rs763816342 Chr.9: 14086374-14086374 on Build GRCh38
rs397800646 Chr.9: 14086392-14086393 on Build GRCh38
rs995867474 Chr.9: 14086361-14086361 on Build GRCh38
rs758063476 Chr.9: 14086382-14086382 on Build GRCh38
rs570489497 Chr.9: 14086069-14086069 on Build GRCh38
rs775093506 Chr.9: 14086150-14086150 on Build GRCh38
rs935431452 Chr.9: 14086268-14086268 on Build GRCh38
rs1014638600 Chr.9: 14086418-14086418 on Build GRCh38
rs80003943 Chr.9: 14085998-14085998 on Build GRCh38
rs1036556133 Chr.9: 14086229-14086229 on Build GRCh38
rs140046069 Chr.9: 14086351-14086354 on Build GRCh38
rs1049627920 Chr.9: 14086316-14086316 on Build GRCh38
rs979605565 Chr.9: 14086078-14086078 on Build GRCh38
rs760174937 Chr.9: 14086257-14086257 on Build GRCh38
rs1043030356 Chr.9: 14086329-14086329 on Build GRCh38
rs926369923 Chr.9: 14086084-14086084 on Build GRCh38
rs988047230 Chr.9: 14086059-14086059 on Build GRCh38
rs950344043 Chr.9: 14086212-14086212 on Build GRCh38
rs1005048680 Chr.9: 14086385-14086385 on Build GRCh38
rs1009509420 Chr.9: 14086435-14086435 on Build GRCh38
rs1018989319 Chr.9: 14086393-14086393 on Build GRCh38
rs568144928 Chr.9: 14086011-14086011 on Build GRCh38
rs533691695 Chr.9: 14086061-14086061 on Build GRCh38
rs186908673 Chr.9: 14086268-14086268 on Build GRCh38
rs149622629 Chr.9: 14086067-14086067 on Build GRCh38
rs928093441 Chr.9: 14086266-14086266 on Build GRCh38
rs541769690 Chr.9: 14086197-14086197 on Build GRCh38
rs963545418 Chr.9: 14086146-14086146 on Build GRCh38
rs375025396 Chr.9: 14086094-14086094 on Build GRCh38
rs967861435 Chr.9: 14086072-14086072 on Build GRCh38
rs954595129 Chr.9: 14086080-14086080 on Build GRCh38
rs917577469 Chr.9: 14086211-14086211 on Build GRCh38
rs538433665 Chr.9: 14086319-14086319 on Build GRCh38
rs948089789 Chr.9: 14086245-14086245 on Build GRCh38
rs199995513 Chr.9: 14086121-14086121 on Build GRCh38
rs555245077 Chr.9: 14086382-14086383 on Build GRCh38
rs75386916 Chr.9: 14086063-14086063 on Build GRCh38

View Full Product Details