Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 14087535-14088040 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NFIB
Entrez Gene ID
4781
Gene Name
nuclear factor I B
Gene Aliases
CTF, HMGIC/NFIB, NF-I/B, NF1-B, NFI-B, NFI-RED, NFIB2, NFIB3
UniGene
Hs.644095
Gene Chromosome Location
Chr.9: 14081843-14398983 on Build GRCh38
Transcripts
NM_001190737.1 NM_001190738.1 NM_001282787.1 NM_005596.3 XM_005251467.2 XM_005251468.4 XM_005251469.2 XM_005251470.2 XM_005251471.3 XM_006716773.2 XM_006716774.2 XM_006716775.2 XM_017014739.1 XM_017014740.1 XM_017014741.1 XM_017014742.1

SNP Detail

SNP ID Chromosome Location
rs7858 Chr.9: 14087770-14087770 on Build GRCh38
rs778162198 Chr.9: 14087652-14087652 on Build GRCh38
rs996874302 Chr.9: 14087599-14087604 on Build GRCh38
rs1006634390 Chr.9: 14087932-14087932 on Build GRCh38
rs111842561 Chr.9: 14087980-14087980 on Build GRCh38
rs1804815 Chr.9: 14087818-14087818 on Build GRCh38
rs189045742 Chr.9: 14087596-14087596 on Build GRCh38
rs554297054 Chr.9: 14087996-14087996 on Build GRCh38
rs991962058 Chr.9: 14087793-14087793 on Build GRCh38
rs749753017 Chr.9: 14087737-14087737 on Build GRCh38
rs897021957 Chr.9: 14087817-14087817 on Build GRCh38
rs993900876 Chr.9: 14087601-14087601 on Build GRCh38
rs768117841 Chr.9: 14087964-14087964 on Build GRCh38
rs980629332 Chr.9: 14087755-14087755 on Build GRCh38
rs891130225 Chr.9: 14087969-14087969 on Build GRCh38
rs914078549 Chr.9: 14087580-14087581 on Build GRCh38
rs1049775645 Chr.9: 14087637-14087637 on Build GRCh38
rs556179175 Chr.9: 14087655-14087655 on Build GRCh38
rs193129743 Chr.9: 14087761-14087761 on Build GRCh38
rs904093587 Chr.9: 14087599-14087599 on Build GRCh38
rs545790805 Chr.9: 14087653-14087653 on Build GRCh38
rs1027136488 Chr.9: 14087605-14087605 on Build GRCh38
rs966887662 Chr.9: 14088003-14088003 on Build GRCh38
rs891091544 Chr.9: 14087682-14087682 on Build GRCh38
rs201574652 Chr.9: 14087581-14087581 on Build GRCh38
rs993931848 Chr.9: 14087822-14087822 on Build GRCh38
rs774801754 Chr.9: 14087777-14087777 on Build GRCh38
rs3739759 Chr.9: 14087767-14087767 on Build GRCh38
rs1046280794 Chr.9: 14087808-14087808 on Build GRCh38
rs961941822 Chr.9: 14087703-14087703 on Build GRCh38
rs952424812 Chr.9: 14087641-14087641 on Build GRCh38
rs78215495 Chr.9: 14087580-14087580 on Build GRCh38
rs760214887 Chr.9: 14087828-14087828 on Build GRCh38
rs939129819 Chr.9: 14087572-14087572 on Build GRCh38
rs978624807 Chr.9: 14088014-14088014 on Build GRCh38
rs368484328 Chr.9: 14087781-14087781 on Build GRCh38
rs951983627 Chr.9: 14087736-14087736 on Build GRCh38
rs993280619 Chr.9: 14087703-14087704 on Build GRCh38
rs11543133 Chr.9: 14087563-14087563 on Build GRCh38
rs527508407 Chr.9: 14087795-14087795 on Build GRCh38
rs945557348 Chr.9: 14087914-14087915 on Build GRCh38
rs1043900961 Chr.9: 14087917-14087917 on Build GRCh38
rs1056584963 Chr.9: 14087577-14087577 on Build GRCh38
rs896990943 Chr.9: 14087597-14087597 on Build GRCh38
rs1018397323 Chr.9: 14087938-14087938 on Build GRCh38
rs1017864513 Chr.9: 14087657-14087657 on Build GRCh38
rs926269794 Chr.9: 14087571-14087571 on Build GRCh38
rs542351570 Chr.9: 14087941-14087941 on Build GRCh38
rs888160432 Chr.9: 14087876-14087876 on Build GRCh38
rs772133620 Chr.9: 14087616-14087616 on Build GRCh38
rs541085958 Chr.9: 14087829-14087829 on Build GRCh38
rs913633750 Chr.9: 14087859-14087859 on Build GRCh38
rs980390473 Chr.9: 14087564-14087570 on Build GRCh38
rs897217458 Chr.9: 14088007-14088007 on Build GRCh38
rs746420804 Chr.9: 14087701-14087701 on Build GRCh38
rs964994050 Chr.9: 14087685-14087685 on Build GRCh38
rs978215719 Chr.9: 14087754-14087754 on Build GRCh38
rs1049970892 Chr.9: 14087959-14087959 on Build GRCh38
rs932901965 Chr.9: 14087945-14087945 on Build GRCh38
rs923459935 Chr.9: 14087726-14087726 on Build GRCh38
rs976329621 Chr.9: 14087703-14087703 on Build GRCh38

View Full Product Details