Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 15171861-15172406 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTC39B
Entrez Gene ID
158219
Gene Name
tetratricopeptide repeat domain 39B
Gene Aliases
C9orf52
UniGene
Hs.563630
Gene Chromosome Location
Chr.9: 15167204-15307360 on Build GRCh38
Transcripts
NM_001168339.1 NM_001168340.1 NM_001168341.1 NM_001168342.1 NM_152574.2 XM_011517732.2 XM_017014310.1 XM_017014311.1 XM_017014312.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3090421 c.1825C>A p.L609I Chr.9: 15172045-15172045 on Build GRCh38
5483130 c.1761-6C>T p.? Chr.9: 15172115-15172115 on Build GRCh38
6879634 c.1830G>A p.W610* Chr.9: 15172040-15172040 on Build GRCh38
3926558 c.1979C>T p.S660F Chr.9: 15172089-15172089 on Build GRCh38
6321448 c.2022C>A p.H674Q Chr.9: 15172046-15172046 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369412614 Chr.9: 15171988-15171988 on Build GRCh38
rs549585549 Chr.9: 15171923-15171923 on Build GRCh38
rs1008885059 Chr.9: 15172161-15172161 on Build GRCh38
rs992380 Chr.9: 15172193-15172193 on Build GRCh38
rs781185978 Chr.9: 15172151-15172151 on Build GRCh38
rs113729233 Chr.9: 15172228-15172228 on Build GRCh38
rs754365052 Chr.9: 15172117-15172117 on Build GRCh38
rs1007707843 Chr.9: 15172362-15172362 on Build GRCh38
rs936642804 Chr.9: 15172227-15172227 on Build GRCh38
rs781657670 Chr.9: 15171974-15171974 on Build GRCh38
rs776309910 Chr.9: 15171972-15171972 on Build GRCh38
rs4401972 Chr.9: 15172222-15172222 on Build GRCh38
rs373714994 Chr.9: 15172046-15172046 on Build GRCh38
rs376814891 Chr.9: 15172300-15172300 on Build GRCh38
rs183780996 Chr.9: 15172264-15172264 on Build GRCh38
rs762225436 Chr.9: 15171982-15171982 on Build GRCh38
rs750861705 Chr.9: 15171985-15171985 on Build GRCh38
rs560908243 Chr.9: 15172274-15172274 on Build GRCh38
rs112377469 Chr.9: 15172256-15172256 on Build GRCh38
rs773587394 Chr.9: 15172095-15172095 on Build GRCh38
rs960345619 Chr.9: 15172238-15172238 on Build GRCh38
rs758212358 Chr.9: 15172023-15172023 on Build GRCh38
rs766836454 Chr.9: 15172106-15172106 on Build GRCh38
rs775148961 Chr.9: 15172073-15172073 on Build GRCh38
rs980810337 Chr.9: 15171943-15171943 on Build GRCh38
rs752900795 Chr.9: 15172135-15172135 on Build GRCh38
rs576476541 Chr.9: 15172170-15172170 on Build GRCh38
rs762515009 Chr.9: 15172078-15172078 on Build GRCh38
rs1009148371 Chr.9: 15172272-15172272 on Build GRCh38
rs764919705 Chr.9: 15171979-15171979 on Build GRCh38
rs756726329 Chr.9: 15172099-15172099 on Build GRCh38
rs748381271 Chr.9: 15172021-15172021 on Build GRCh38
rs190313241 Chr.9: 15172379-15172379 on Build GRCh38
rs1045408507 Chr.9: 15171933-15171933 on Build GRCh38
rs771017079 Chr.9: 15172041-15172041 on Build GRCh38
rs115979142 Chr.9: 15171901-15171901 on Build GRCh38
rs777664011 Chr.9: 15172142-15172142 on Build GRCh38
rs186036947 Chr.9: 15172269-15172269 on Build GRCh38
rs929281096 Chr.9: 15172167-15172167 on Build GRCh38
rs1030056062 Chr.9: 15172309-15172309 on Build GRCh38
rs1017795121 Chr.9: 15172372-15172372 on Build GRCh38
rs778431101 Chr.9: 15172119-15172119 on Build GRCh38
rs890183567 Chr.9: 15172336-15172336 on Build GRCh38
rs4335225 Chr.9: 15172187-15172187 on Build GRCh38
rs971907057 Chr.9: 15171902-15171902 on Build GRCh38
rs766675368 Chr.9: 15171887-15171887 on Build GRCh38
rs757159390 Chr.9: 15172150-15172150 on Build GRCh38
rs191955878 Chr.9: 15172260-15172260 on Build GRCh38
rs756642508 Chr.9: 15171987-15171987 on Build GRCh38
rs537909451 Chr.9: 15171908-15171908 on Build GRCh38
rs746779580 Chr.9: 15172145-15172145 on Build GRCh38
rs73646009 Chr.9: 15172223-15172223 on Build GRCh38
rs926556769 Chr.9: 15172235-15172235 on Build GRCh38
rs1001181251 Chr.9: 15171935-15171935 on Build GRCh38
rs758728517 Chr.9: 15172139-15172139 on Build GRCh38
rs1009298904 Chr.9: 15172022-15172022 on Build GRCh38
rs1042250799 Chr.9: 15172268-15172268 on Build GRCh38
rs746906015 Chr.9: 15172035-15172035 on Build GRCh38
rs552060922 Chr.9: 15172009-15172009 on Build GRCh38
rs571528304 Chr.9: 15171976-15171976 on Build GRCh38
rs779196250 Chr.9: 15172012-15172012 on Build GRCh38
rs543871268 Chr.9: 15172202-15172202 on Build GRCh38
rs187973471 Chr.9: 15172024-15172024 on Build GRCh38
rs565482860 Chr.9: 15172221-15172221 on Build GRCh38
rs753928128 Chr.9: 15171994-15171994 on Build GRCh38
rs374874462 Chr.9: 15171980-15171980 on Build GRCh38
rs185168790 Chr.9: 15171965-15171965 on Build GRCh38
rs949258334 Chr.9: 15172220-15172220 on Build GRCh38
rs769245921 Chr.9: 15172058-15172058 on Build GRCh38
rs761074525 Chr.9: 15172100-15172100 on Build GRCh38
rs980731389 Chr.9: 15172234-15172234 on Build GRCh38
rs372754350 Chr.9: 15172131-15172131 on Build GRCh38
rs531307781 Chr.9: 15172281-15172281 on Build GRCh38
rs748813604 Chr.9: 15171911-15171912 on Build GRCh38
rs917297171 Chr.9: 15172197-15172197 on Build GRCh38
rs1057221285 Chr.9: 15171895-15171895 on Build GRCh38
rs773915753 Chr.9: 15172055-15172055 on Build GRCh38
rs776759759 Chr.9: 15172045-15172045 on Build GRCh38
rs140541523 Chr.9: 15172094-15172094 on Build GRCh38
rs181067791 Chr.9: 15171949-15171949 on Build GRCh38
rs1050835998 Chr.9: 15172146-15172146 on Build GRCh38
rs537100096 Chr.9: 15172189-15172189 on Build GRCh38
rs150728160 Chr.9: 15171984-15171984 on Build GRCh38
rs751891156 Chr.9: 15171905-15171905 on Build GRCh38
rs551648044 Chr.9: 15171934-15171934 on Build GRCh38
rs926322275 Chr.9: 15171953-15171953 on Build GRCh38

View Full Product Details