Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 19512367-19512882 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC24A2
Entrez Gene ID
25769
Gene Name
solute carrier family 24 member 2
Gene Aliases
NCKX2
UniGene
Hs.283014
Gene Chromosome Location
Chr.9: 19507452-20307888 on Build GRCh38
Transcripts
NM_001193288.2 NM_020344.3 XM_005251425.2 XM_005251426.4 XM_006716750.3 XM_017014592.1 XM_017014593.1

SNP Detail

SNP ID Chromosome Location
rs1042808221 Chr.9: 19512667-19512667 on Build GRCh38
rs975153727 Chr.9: 19512854-19512854 on Build GRCh38
rs951656569 Chr.9: 19512807-19512808 on Build GRCh38
rs1039563653 Chr.9: 19512425-19512425 on Build GRCh38
rs546950005 Chr.9: 19512660-19512660 on Build GRCh38
rs534220574 Chr.9: 19512715-19512715 on Build GRCh38
rs576072296 Chr.9: 19512825-19512825 on Build GRCh38
rs542982460 Chr.9: 19512419-19512419 on Build GRCh38
rs942535004 Chr.9: 19512809-19512809 on Build GRCh38
rs975645850 Chr.9: 19512667-19512668 on Build GRCh38
rs4977545 Chr.9: 19512748-19512748 on Build GRCh38
rs761291129 Chr.9: 19512668-19512668 on Build GRCh38
rs571229101 Chr.9: 19512607-19512607 on Build GRCh38
rs759987799 Chr.9: 19512568-19512568 on Build GRCh38
rs1011528338 Chr.9: 19512477-19512477 on Build GRCh38
rs753416305 Chr.9: 19512680-19512680 on Build GRCh38
rs903117154 Chr.9: 19512709-19512709 on Build GRCh38
rs969563033 Chr.9: 19512566-19512566 on Build GRCh38
rs532928154 Chr.9: 19512773-19512773 on Build GRCh38
rs532239729 Chr.9: 19512610-19512610 on Build GRCh38
rs1026093789 Chr.9: 19512728-19512728 on Build GRCh38
rs1031066473 Chr.9: 19512492-19512492 on Build GRCh38
rs984276337 Chr.9: 19512515-19512515 on Build GRCh38
rs982318802 Chr.9: 19512582-19512582 on Build GRCh38
rs916720903 Chr.9: 19512554-19512554 on Build GRCh38
rs188506762 Chr.9: 19512766-19512766 on Build GRCh38
rs1027200827 Chr.9: 19512608-19512608 on Build GRCh38
rs947059461 Chr.9: 19512791-19512791 on Build GRCh38
rs965374912 Chr.9: 19512461-19512461 on Build GRCh38
rs951022535 Chr.9: 19512622-19512622 on Build GRCh38
rs779441083 Chr.9: 19512476-19512476 on Build GRCh38
rs372341797 Chr.9: 19512717-19512717 on Build GRCh38
rs549295726 Chr.9: 19512721-19512721 on Build GRCh38
rs978447900 Chr.9: 19512792-19512792 on Build GRCh38
rs544283960 Chr.9: 19512584-19512584 on Build GRCh38
rs943832925 Chr.9: 19512663-19512663 on Build GRCh38
rs928211281 Chr.9: 19512604-19512604 on Build GRCh38
rs1040880618 Chr.9: 19512815-19512815 on Build GRCh38
rs74677410 Chr.9: 19512770-19512770 on Build GRCh38
rs1019493886 Chr.9: 19512457-19512457 on Build GRCh38
rs185654581 Chr.9: 19512691-19512691 on Build GRCh38
rs531432183 Chr.9: 19512513-19512513 on Build GRCh38
rs549830094 Chr.9: 19512595-19512595 on Build GRCh38
rs942544831 Chr.9: 19512710-19512710 on Build GRCh38
rs763636208 Chr.9: 19512679-19512679 on Build GRCh38
rs1025725425 Chr.9: 19512502-19512502 on Build GRCh38
rs912868869 Chr.9: 19512788-19512788 on Build GRCh38
rs999537531 Chr.9: 19512491-19512491 on Build GRCh38
rs577999164 Chr.9: 19512776-19512776 on Build GRCh38
rs1005615636 Chr.9: 19512750-19512750 on Build GRCh38
rs899736188 Chr.9: 19512470-19512470 on Build GRCh38
rs943846727 Chr.9: 19512418-19512418 on Build GRCh38
rs1043746252 Chr.9: 19512483-19512483 on Build GRCh38
rs911005036 Chr.9: 19512795-19512795 on Build GRCh38
rs994162937 Chr.9: 19512722-19512722 on Build GRCh38
rs1013709724 Chr.9: 19512743-19512743 on Build GRCh38
rs947099493 Chr.9: 19512662-19512662 on Build GRCh38
rs7872265 Chr.9: 19512413-19512413 on Build GRCh38
rs1023837976 Chr.9: 19512585-19512585 on Build GRCh38
rs774576803 Chr.9: 19512484-19512484 on Build GRCh38
rs1023725577 Chr.9: 19512755-19512755 on Build GRCh38
rs758264231 Chr.9: 19512768-19512768 on Build GRCh38
rs772438519 Chr.9: 19512615-19512615 on Build GRCh38
rs556112958 Chr.9: 19512467-19512467 on Build GRCh38
rs771201762 Chr.9: 19512415-19512415 on Build GRCh38
rs982401035 Chr.9: 19512810-19512810 on Build GRCh38
rs929853185 Chr.9: 19512829-19512829 on Build GRCh38
rs538521533 Chr.9: 19512627-19512627 on Build GRCh38

View Full Product Details