Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 27180115-27180627 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TEK
Entrez Gene ID
7010
Gene Name
TEK receptor tyrosine kinase
Gene Aliases
CD202B, TIE-2, TIE2, VMCM, VMCM1
UniGene
Hs.89640
Gene Chromosome Location
Chr.9: 27109141-27230178 on Build GRCh38
Transcripts
NM_000459.4 NM_001290077.1 NM_001290078.1 XM_005251561.1 XM_005251563.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6495287 c.902-10C>T p.? Chr.9: 27180230-27180230 on Build GRCh38
7232641 c.981A>C p.Q327H Chr.9: 27180319-27180319 on Build GRCh38
5998428 c.1030+5A>C p.? Chr.9: 27180373-27180373 on Build GRCh38
7226704 c.973C>G p.R325G Chr.9: 27180311-27180311 on Build GRCh38
389697 c.933T>A p.C311* Chr.9: 27180271-27180271 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770036661 Chr.9: 27180213-27180213 on Build GRCh38
rs559473908 Chr.9: 27180291-27180291 on Build GRCh38
rs780271276 Chr.9: 27180298-27180298 on Build GRCh38
rs771862350 Chr.9: 27180484-27180484 on Build GRCh38
rs185470630 Chr.9: 27180430-27180430 on Build GRCh38
rs111813503 Chr.9: 27180536-27180536 on Build GRCh38
rs755358740 Chr.9: 27180425-27180425 on Build GRCh38
rs982722071 Chr.9: 27180445-27180445 on Build GRCh38
rs375366420 Chr.9: 27180447-27180447 on Build GRCh38
rs755127644 Chr.9: 27180311-27180311 on Build GRCh38
rs754111503 Chr.9: 27180408-27180408 on Build GRCh38
rs180827182 Chr.9: 27180319-27180319 on Build GRCh38
rs991212252 Chr.9: 27180587-27180587 on Build GRCh38
rs749678742 Chr.9: 27180302-27180302 on Build GRCh38
rs541217363 Chr.9: 27180259-27180259 on Build GRCh38
rs139143068 Chr.9: 27180268-27180268 on Build GRCh38
rs768401874 Chr.9: 27180216-27180217 on Build GRCh38
rs376241578 Chr.9: 27180400-27180400 on Build GRCh38
rs566949886 Chr.9: 27180483-27180483 on Build GRCh38
rs536961772 Chr.9: 27180429-27180429 on Build GRCh38
rs1035096736 Chr.9: 27180522-27180522 on Build GRCh38
rs989605542 Chr.9: 27180216-27180216 on Build GRCh38
rs774357901 Chr.9: 27180221-27180221 on Build GRCh38
rs200827916 Chr.9: 27180386-27180386 on Build GRCh38
rs372703259 Chr.9: 27180219-27180219 on Build GRCh38
rs944559197 Chr.9: 27180293-27180293 on Build GRCh38
rs760714202 Chr.9: 27180248-27180248 on Build GRCh38
rs528575879 Chr.9: 27180161-27180161 on Build GRCh38
rs990871408 Chr.9: 27180146-27180146 on Build GRCh38
rs368019588 Chr.9: 27180432-27180432 on Build GRCh38
rs746083594 Chr.9: 27180140-27180140 on Build GRCh38
rs763749051 Chr.9: 27180280-27180280 on Build GRCh38
rs952152168 Chr.9: 27180590-27180590 on Build GRCh38
rs781764670 Chr.9: 27180204-27180204 on Build GRCh38
rs533028014 Chr.9: 27180249-27180249 on Build GRCh38
rs756713430 Chr.9: 27180294-27180294 on Build GRCh38
rs534395703 Chr.9: 27180511-27180511 on Build GRCh38
rs41272241 Chr.9: 27180230-27180230 on Build GRCh38
rs373027386 Chr.9: 27180363-27180363 on Build GRCh38
rs983361680 Chr.9: 27180600-27180600 on Build GRCh38
rs772115655 Chr.9: 27180345-27180345 on Build GRCh38
rs371104063 Chr.9: 27180253-27180253 on Build GRCh38
rs749469926 Chr.9: 27180218-27180218 on Build GRCh38
rs1009138590 Chr.9: 27180163-27180163 on Build GRCh38
rs773380552 Chr.9: 27180352-27180352 on Build GRCh38
rs931631791 Chr.9: 27180424-27180424 on Build GRCh38
rs541922300 Chr.9: 27180545-27180545 on Build GRCh38
rs894031385 Chr.9: 27180446-27180446 on Build GRCh38
rs767486829 Chr.9: 27180589-27180589 on Build GRCh38
rs776229235 Chr.9: 27180361-27180361 on Build GRCh38
rs41272243 Chr.9: 27180493-27180493 on Build GRCh38
rs761886420 Chr.9: 27180226-27180226 on Build GRCh38
rs1031108466 Chr.9: 27180181-27180181 on Build GRCh38
rs770197818 Chr.9: 27180200-27180200 on Build GRCh38
rs770164895 Chr.9: 27180385-27180386 on Build GRCh38
rs144863798 Chr.9: 27180394-27180394 on Build GRCh38
rs900266840 Chr.9: 27180397-27180397 on Build GRCh38
rs748176673 Chr.9: 27180336-27180336 on Build GRCh38
rs190492481 Chr.9: 27180211-27180211 on Build GRCh38
rs190231761 Chr.9: 27180599-27180599 on Build GRCh38
rs752184169 Chr.9: 27180260-27180260 on Build GRCh38
rs111637971 Chr.9: 27180337-27180337 on Build GRCh38
rs375476750 Chr.9: 27180391-27180391 on Build GRCh38
rs766915144 Chr.9: 27180405-27180405 on Build GRCh38
rs759247392 Chr.9: 27180256-27180256 on Build GRCh38
rs941219209 Chr.9: 27180455-27180455 on Build GRCh38
rs775264500 Chr.9: 27180374-27180374 on Build GRCh38
rs773044536 Chr.9: 27180244-27180244 on Build GRCh38
rs780511049 Chr.9: 27180215-27180215 on Build GRCh38
rs757777004 Chr.9: 27180192-27180192 on Build GRCh38
rs762737786 Chr.9: 27180384-27180384 on Build GRCh38

View Full Product Details