Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 32570751-32571270 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDUFB6
Entrez Gene ID
4712
Gene Name
NADH:ubiquinone oxidoreductase subunit B6
Gene Aliases
B17, CI
UniGene
Hs.493668
Gene Chromosome Location
Chr.9: 32553526-32573184 on Build GRCh38
Transcripts
NM_001199987.1 NM_002493.4 NM_182739.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6115529 c.239_240GG>TT p.W80F Chr.9: 32570993-32570994 on Build GRCh38
3716174 c.243T>C p.I81I Chr.9: 32570990-32570990 on Build GRCh38
4908321 c.183C>A p.V61V Chr.9: 32571050-32571050 on Build GRCh38
1169165 c.261G>A p.K87K Chr.9: 32570972-32570972 on Build GRCh38
487365 c.223G>T p.V75L Chr.9: 32571010-32571010 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780887901 Chr.9: 32570948-32570948 on Build GRCh38
rs776835068 Chr.9: 32570909-32570909 on Build GRCh38
rs544185072 Chr.9: 32571145-32571145 on Build GRCh38
rs750044120 Chr.9: 32571060-32571060 on Build GRCh38
rs775513868 Chr.9: 32571055-32571055 on Build GRCh38
rs753296996 Chr.9: 32571094-32571094 on Build GRCh38
rs10971029 Chr.9: 32571099-32571099 on Build GRCh38
rs372259032 Chr.9: 32570998-32570998 on Build GRCh38
rs539902490 Chr.9: 32570862-32570862 on Build GRCh38
rs978324562 Chr.9: 32570888-32570888 on Build GRCh38
rs761634938 Chr.9: 32571055-32571055 on Build GRCh38
rs569113741 Chr.9: 32570822-32570822 on Build GRCh38
rs935126719 Chr.9: 32570968-32570968 on Build GRCh38
rs887713874 Chr.9: 32571082-32571082 on Build GRCh38
rs765844487 Chr.9: 32571068-32571068 on Build GRCh38
rs75143049 Chr.9: 32571154-32571154 on Build GRCh38
rs555004436 Chr.9: 32570898-32570898 on Build GRCh38
rs367629179 Chr.9: 32571044-32571044 on Build GRCh38
rs762045301 Chr.9: 32571061-32571061 on Build GRCh38
rs190741792 Chr.9: 32570928-32570928 on Build GRCh38
rs538539945 Chr.9: 32570868-32570868 on Build GRCh38
rs1050307164 Chr.9: 32571048-32571048 on Build GRCh38
rs771250384 Chr.9: 32571026-32571026 on Build GRCh38
rs200723755 Chr.9: 32571015-32571015 on Build GRCh38
rs1026670300 Chr.9: 32571182-32571182 on Build GRCh38
rs1038402787 Chr.9: 32571108-32571108 on Build GRCh38
rs749867616 Chr.9: 32570957-32570957 on Build GRCh38
rs1005538706 Chr.9: 32571098-32571098 on Build GRCh38
rs781361290 Chr.9: 32570914-32570914 on Build GRCh38
rs376279037 Chr.9: 32571031-32571031 on Build GRCh38
rs776963376 Chr.9: 32571028-32571028 on Build GRCh38
rs767705334 Chr.9: 32571102-32571104 on Build GRCh38
rs555115037 Chr.9: 32570967-32570967 on Build GRCh38
rs544749356 Chr.9: 32571030-32571030 on Build GRCh38
rs762833501 Chr.9: 32570922-32570922 on Build GRCh38
rs953739576 Chr.9: 32571199-32571199 on Build GRCh38
rs765272206 Chr.9: 32570933-32570936 on Build GRCh38
rs995222416 Chr.9: 32571167-32571167 on Build GRCh38
rs769852730 Chr.9: 32571013-32571013 on Build GRCh38
rs759704339 Chr.9: 32570913-32570913 on Build GRCh38
rs574789301 Chr.9: 32570921-32570921 on Build GRCh38
rs767132728 Chr.9: 32571056-32571056 on Build GRCh38
rs539103278 Chr.9: 32570799-32570799 on Build GRCh38
rs961509415 Chr.9: 32570780-32570781 on Build GRCh38
rs751371364 Chr.9: 32570926-32570926 on Build GRCh38
rs961162064 Chr.9: 32570813-32570813 on Build GRCh38
rs1037600945 Chr.9: 32571141-32571141 on Build GRCh38
rs531083959 Chr.9: 32571246-32571246 on Build GRCh38
rs746015407 Chr.9: 32570964-32570964 on Build GRCh38
rs1006947406 Chr.9: 32571228-32571228 on Build GRCh38
rs763810334 Chr.9: 32570923-32570923 on Build GRCh38
rs755753506 Chr.9: 32571061-32571061 on Build GRCh38
rs530383017 Chr.9: 32571139-32571139 on Build GRCh38
rs775438002 Chr.9: 32571039-32571039 on Build GRCh38
rs148891049 Chr.9: 32570991-32570991 on Build GRCh38
rs897224354 Chr.9: 32571134-32571134 on Build GRCh38
rs146518353 Chr.9: 32571041-32571041 on Build GRCh38
rs558935311 Chr.9: 32570800-32570800 on Build GRCh38
rs1019565860 Chr.9: 32571237-32571237 on Build GRCh38
rs748645977 Chr.9: 32570981-32570981 on Build GRCh38
rs779451513 Chr.9: 32570973-32570973 on Build GRCh38

View Full Product Details