Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 33922406-33922914 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBAP2
Entrez Gene ID
55833
Gene Name
ubiquitin associated protein 2
Gene Aliases
UBAP-2
UniGene
Hs.493739
Gene Chromosome Location
Chr.9: 33921693-34048949 on Build GRCh38
Transcripts
NM_001282529.1 NM_001282530.1 NM_018449.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5746975 c.3169C>A p.P1057T Chr.9: 33922782-33922782 on Build GRCh38
1292845 c.3140C>T p.S1047F Chr.9: 33922811-33922811 on Build GRCh38
6039205 c.3347A>G p.Y1116C Chr.9: 33922517-33922517 on Build GRCh38
264512 c.3280C>T p.R1094C Chr.9: 33922584-33922584 on Build GRCh38
1108386 c.3102G>A p.G1034G Chr.9: 33922849-33922849 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770457869 Chr.9: 33922447-33922447 on Build GRCh38
rs778396687 Chr.9: 33922626-33922626 on Build GRCh38
rs745449642 Chr.9: 33922607-33922607 on Build GRCh38
rs10971793 Chr.9: 33922628-33922628 on Build GRCh38
rs754551233 Chr.9: 33922789-33922789 on Build GRCh38
rs766563675 Chr.9: 33922514-33922514 on Build GRCh38
rs746282565 Chr.9: 33922808-33922808 on Build GRCh38
rs762246038 Chr.9: 33922531-33922531 on Build GRCh38
rs145638776 Chr.9: 33922701-33922701 on Build GRCh38
rs772271164 Chr.9: 33922592-33922592 on Build GRCh38
rs958975802 Chr.9: 33922510-33922510 on Build GRCh38
rs750238859 Chr.9: 33922774-33922774 on Build GRCh38
rs755650960 Chr.9: 33922611-33922611 on Build GRCh38
rs751481975 Chr.9: 33922664-33922664 on Build GRCh38
rs767712732 Chr.9: 33922479-33922479 on Build GRCh38
rs767370789 Chr.9: 33922773-33922773 on Build GRCh38
rs749444181 Chr.9: 33922823-33922823 on Build GRCh38
rs773576049 Chr.9: 33922480-33922480 on Build GRCh38
rs149408613 Chr.9: 33922875-33922875 on Build GRCh38
rs767228813 Chr.9: 33922673-33922673 on Build GRCh38
rs376765185 Chr.9: 33922461-33922461 on Build GRCh38
rs779395548 Chr.9: 33922615-33922615 on Build GRCh38
rs1015319543 Chr.9: 33922484-33922484 on Build GRCh38
rs771427881 Chr.9: 33922707-33922707 on Build GRCh38
rs758227401 Chr.9: 33922498-33922498 on Build GRCh38
rs201372104 Chr.9: 33922617-33922617 on Build GRCh38
rs35781362 Chr.9: 33922807-33922807 on Build GRCh38
rs748209060 Chr.9: 33922839-33922839 on Build GRCh38
rs753331597 Chr.9: 33922695-33922695 on Build GRCh38
rs201968722 Chr.9: 33922500-33922500 on Build GRCh38
rs919197593 Chr.9: 33922837-33922837 on Build GRCh38
rs774271971 Chr.9: 33922763-33922763 on Build GRCh38
rs773916321 Chr.9: 33922789-33922818 on Build GRCh38
rs534549028 Chr.9: 33922450-33922450 on Build GRCh38
rs747553647 Chr.9: 33922817-33922818 on Build GRCh38
rs375245273 Chr.9: 33922810-33922810 on Build GRCh38
rs760943172 Chr.9: 33922488-33922488 on Build GRCh38
rs754983146 Chr.9: 33922492-33922492 on Build GRCh38
rs769937768 Chr.9: 33922649-33922649 on Build GRCh38
rs779739415 Chr.9: 33922691-33922691 on Build GRCh38
rs375276983 Chr.9: 33922769-33922769 on Build GRCh38
rs776989578 Chr.9: 33922574-33922574 on Build GRCh38
rs752710693 Chr.9: 33922588-33922588 on Build GRCh38
rs753516283 Chr.9: 33922786-33922786 on Build GRCh38
rs571014698 Chr.9: 33922551-33922551 on Build GRCh38
rs192873385 Chr.9: 33922467-33922467 on Build GRCh38
rs140479290 Chr.9: 33922591-33922591 on Build GRCh38
rs756513939 Chr.9: 33922600-33922601 on Build GRCh38
rs989223860 Chr.9: 33922541-33922541 on Build GRCh38
rs966210844 Chr.9: 33922682-33922682 on Build GRCh38
rs556932450 Chr.9: 33922599-33922599 on Build GRCh38
rs775003963 Chr.9: 33922503-33922503 on Build GRCh38
rs138911165 Chr.9: 33922793-33922793 on Build GRCh38
rs745347097 Chr.9: 33922456-33922456 on Build GRCh38
rs377358049 Chr.9: 33922699-33922699 on Build GRCh38
rs775774831 Chr.9: 33922730-33922730 on Build GRCh38
rs766016402 Chr.9: 33922783-33922783 on Build GRCh38
rs565347052 Chr.9: 33922675-33922675 on Build GRCh38
rs759721895 Chr.9: 33922583-33922583 on Build GRCh38
rs745447785 Chr.9: 33922527-33922527 on Build GRCh38
rs369248975 Chr.9: 33922532-33922532 on Build GRCh38
rs759610789 Chr.9: 33922644-33922644 on Build GRCh38
rs376968021 Chr.9: 33922656-33922656 on Build GRCh38
rs561316385 Chr.9: 33922825-33922825 on Build GRCh38
rs771760254 Chr.9: 33922440-33922440 on Build GRCh38
rs966323533 Chr.9: 33922466-33922466 on Build GRCh38
rs763075750 Chr.9: 33922744-33922744 on Build GRCh38
rs899725416 Chr.9: 33922438-33922438 on Build GRCh38
rs769244530 Chr.9: 33922642-33922643 on Build GRCh38
rs756950516 Chr.9: 33922517-33922517 on Build GRCh38
rs754577964 Chr.9: 33922608-33922610 on Build GRCh38
rs752294529 Chr.9: 33922886-33922886 on Build GRCh38
rs536616785 Chr.9: 33922555-33922555 on Build GRCh38
rs368110650 Chr.9: 33922846-33922846 on Build GRCh38
rs752143649 Chr.9: 33922792-33922792 on Build GRCh38
rs141917867 Chr.9: 33922765-33922765 on Build GRCh38
rs760448754 Chr.9: 33922862-33922862 on Build GRCh38
rs1033333799 Chr.9: 33922698-33922698 on Build GRCh38
rs753968920 Chr.9: 33922704-33922704 on Build GRCh38
rs761608111 Chr.9: 33922669-33922669 on Build GRCh38
rs138274627 Chr.9: 33922713-33922713 on Build GRCh38
rs199859255 Chr.9: 33922700-33922700 on Build GRCh38
rs41313969 Chr.9: 33922659-33922659 on Build GRCh38
rs757038458 Chr.9: 33922598-33922598 on Build GRCh38
rs781253292 Chr.9: 33922440-33922440 on Build GRCh38
rs1036700769 Chr.9: 33922620-33922620 on Build GRCh38
rs901991064 Chr.9: 33922870-33922870 on Build GRCh38
rs778499089 Chr.9: 33922790-33922790 on Build GRCh38
rs761100974 Chr.9: 33922564-33922564 on Build GRCh38
rs370230611 Chr.9: 33922847-33922847 on Build GRCh38
rs768662520 Chr.9: 33922762-33922762 on Build GRCh38
rs547788118 Chr.9: 33922491-33922492 on Build GRCh38
rs771411622 Chr.9: 33922636-33922636 on Build GRCh38
rs746596776 Chr.9: 33922441-33922441 on Build GRCh38
rs780751571 Chr.9: 33922605-33922606 on Build GRCh38
rs185150870 Chr.9: 33922625-33922625 on Build GRCh38
rs752564823 Chr.9: 33922496-33922496 on Build GRCh38
rs754023879 Chr.9: 33922491-33922491 on Build GRCh38
rs781701472 Chr.9: 33922802-33922802 on Build GRCh38
rs780789852 Chr.9: 33922519-33922519 on Build GRCh38
rs150662040 Chr.9: 33922775-33922775 on Build GRCh38
rs989582656 Chr.9: 33922737-33922737 on Build GRCh38
rs762292774 Chr.9: 33922469-33922469 on Build GRCh38
rs776429442 Chr.9: 33922874-33922874 on Build GRCh38
rs371674945 Chr.9: 33922543-33922543 on Build GRCh38
rs977533999 Chr.9: 33922748-33922748 on Build GRCh38
rs774361919 Chr.9: 33922831-33922831 on Build GRCh38
rs755677865 Chr.9: 33922683-33922683 on Build GRCh38
rs756486152 Chr.9: 33922817-33922817 on Build GRCh38
rs968858433 Chr.9: 33922746-33922746 on Build GRCh38
rs138140692 Chr.9: 33922584-33922584 on Build GRCh38
rs200919379 Chr.9: 33922643-33922643 on Build GRCh38
rs1007616859 Chr.9: 33922867-33922867 on Build GRCh38
rs143780579 Chr.9: 33922868-33922868 on Build GRCh38
rs957894806 Chr.9: 33922883-33922883 on Build GRCh38
rs765287942 Chr.9: 33922495-33922495 on Build GRCh38
rs746221826 Chr.9: 33922715-33922715 on Build GRCh38
rs188768044 Chr.9: 33922822-33922822 on Build GRCh38
rs748018487 Chr.9: 33922434-33922434 on Build GRCh38
rs766590716 Chr.9: 33922489-33922489 on Build GRCh38
rs775480917 Chr.9: 33922652-33922652 on Build GRCh38

View Full Product Details