Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 34254994-34255503 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF24
Entrez Gene ID
347240
Gene Name
kinesin family member 24
Gene Aliases
C9orf48, bA571F15.4
UniGene
Hs.710246
Gene Chromosome Location
Chr.9: 34252380-34332972 on Build GRCh38
Transcripts
NM_194313.2 XM_005251451.2 XM_006716764.2 XM_011517861.1 XM_011517863.2 XM_017014697.1 XM_017014698.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1108419 c.3924C>T p.L1308L Chr.9: 34255114-34255114 on Build GRCh38
1472150 c.3941C>T p.T1314M Chr.9: 34255097-34255097 on Build GRCh38
1472151 c.2387C>T p.T796M Chr.9: 34255097-34255097 on Build GRCh38
1108420 c.2370C>T p.L790L Chr.9: 34255114-34255114 on Build GRCh38
5411015 c.3882C>T p.V1294V Chr.9: 34255156-34255156 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770333393 Chr.9: 34255063-34255063 on Build GRCh38
rs528254075 Chr.9: 34255404-34255404 on Build GRCh38
rs949813995 Chr.9: 34255385-34255385 on Build GRCh38
rs772462087 Chr.9: 34255319-34255319 on Build GRCh38
rs553561001 Chr.9: 34255079-34255079 on Build GRCh38
rs375506200 Chr.9: 34255124-34255124 on Build GRCh38
rs1038688240 Chr.9: 34255444-34255444 on Build GRCh38
rs1054478211 Chr.9: 34255279-34255279 on Build GRCh38
rs1015825556 Chr.9: 34255369-34255369 on Build GRCh38
rs774808437 Chr.9: 34255072-34255072 on Build GRCh38
rs143611896 Chr.9: 34255039-34255040 on Build GRCh38
rs77384420 Chr.9: 34255228-34255228 on Build GRCh38
rs746568082 Chr.9: 34255057-34255057 on Build GRCh38
rs778707752 Chr.9: 34255138-34255138 on Build GRCh38
rs915656263 Chr.9: 34255419-34255419 on Build GRCh38
rs371161721 Chr.9: 34255097-34255097 on Build GRCh38
rs750652909 Chr.9: 34255089-34255089 on Build GRCh38
rs913918803 Chr.9: 34255037-34255037 on Build GRCh38
rs747818188 Chr.9: 34255022-34255022 on Build GRCh38
rs138166811 Chr.9: 34255362-34255362 on Build GRCh38
rs767894069 Chr.9: 34255083-34255083 on Build GRCh38
rs867763583 Chr.9: 34255476-34255476 on Build GRCh38
rs775243991 Chr.9: 34255275-34255275 on Build GRCh38
rs79120725 Chr.9: 34255359-34255359 on Build GRCh38
rs924416429 Chr.9: 34255437-34255437 on Build GRCh38
rs987460844 Chr.9: 34255035-34255035 on Build GRCh38
rs10972024 Chr.9: 34255317-34255317 on Build GRCh38
rs770417380 Chr.9: 34255091-34255091 on Build GRCh38
rs777457806 Chr.9: 34255176-34255176 on Build GRCh38
rs997346971 Chr.9: 34255342-34255342 on Build GRCh38
rs749823916 Chr.9: 34255203-34255203 on Build GRCh38
rs770477281 Chr.9: 34255180-34255180 on Build GRCh38
rs763302512 Chr.9: 34255067-34255067 on Build GRCh38
rs915733552 Chr.9: 34255226-34255226 on Build GRCh38
rs996720186 Chr.9: 34255268-34255268 on Build GRCh38
rs913854530 Chr.9: 34255337-34255337 on Build GRCh38
rs542510188 Chr.9: 34255363-34255363 on Build GRCh38
rs963862106 Chr.9: 34255249-34255249 on Build GRCh38
rs747285908 Chr.9: 34255046-34255060 on Build GRCh38
rs866562264 Chr.9: 34255143-34255143 on Build GRCh38
rs754818374 Chr.9: 34255134-34255134 on Build GRCh38
rs753823453 Chr.9: 34255123-34255123 on Build GRCh38
rs140475225 Chr.9: 34255205-34255205 on Build GRCh38
rs566131291 Chr.9: 34255378-34255378 on Build GRCh38
rs980085561 Chr.9: 34255353-34255353 on Build GRCh38
rs773657991 Chr.9: 34255367-34255367 on Build GRCh38
rs946936265 Chr.9: 34255041-34255041 on Build GRCh38
rs559162712 Chr.9: 34255377-34255377 on Build GRCh38
rs373713874 Chr.9: 34255052-34255052 on Build GRCh38
rs955415597 Chr.9: 34255308-34255308 on Build GRCh38
rs367826400 Chr.9: 34255065-34255065 on Build GRCh38
rs80112237 Chr.9: 34255475-34255475 on Build GRCh38
rs764532598 Chr.9: 34255068-34255068 on Build GRCh38
rs746659730 Chr.9: 34255179-34255179 on Build GRCh38
rs918098308 Chr.9: 34255384-34255384 on Build GRCh38
rs114372480 Chr.9: 34255427-34255427 on Build GRCh38
rs920240220 Chr.9: 34255113-34255113 on Build GRCh38
rs771515517 Chr.9: 34255188-34255189 on Build GRCh38
rs769004927 Chr.9: 34255204-34255204 on Build GRCh38
rs934577933 Chr.9: 34255454-34255454 on Build GRCh38
rs908788841 Chr.9: 34255045-34255046 on Build GRCh38
rs762319144 Chr.9: 34255209-34255209 on Build GRCh38
rs762366536 Chr.9: 34255379-34255379 on Build GRCh38
rs752525182 Chr.9: 34255151-34255151 on Build GRCh38
rs372218054 Chr.9: 34255096-34255096 on Build GRCh38
rs575615691 Chr.9: 34255291-34255291 on Build GRCh38
rs968085692 Chr.9: 34255340-34255340 on Build GRCh38
rs949952596 Chr.9: 34255425-34255425 on Build GRCh38
rs1055467582 Chr.9: 34255086-34255086 on Build GRCh38
rs771418666 Chr.9: 34255384-34255385 on Build GRCh38
rs35693912 Chr.9: 34255269-34255269 on Build GRCh38
rs1025702993 Chr.9: 34255135-34255135 on Build GRCh38
rs1046096329 Chr.9: 34255235-34255235 on Build GRCh38
rs55684896 Chr.9: 34255140-34255140 on Build GRCh38
rs10972025 Chr.9: 34255415-34255415 on Build GRCh38
rs780510545 Chr.9: 34255194-34255194 on Build GRCh38
rs758164703 Chr.9: 34255162-34255162 on Build GRCh38
rs181302696 Chr.9: 34255039-34255039 on Build GRCh38

View Full Product Details