Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 34398477-34398966 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf24
Entrez Gene ID
84688
Gene Name
chromosome 9 open reading frame 24
Gene Aliases
CBE1, NYD-SP22, SMRP1, bA573M23.4
UniGene
Hs.50334
Gene Chromosome Location
Chr.9: 34379019-34400833 on Build GRCh38
Transcripts
XM_005251616.4 NM_001184940.1 NM_001184941.1 NM_001184942.1 NM_001184943.1 NM_001184945.1 NM_147202.1 XM_005251402.4 XM_011517797.2 XM_011517798.2 XM_011517799.2

Gene Symbol
FAM219A
Entrez Gene ID
203259
Gene Name
family with sequence similarity 219 member A
Gene Aliases
C9orf25
UniGene
Hs.493771
Gene Chromosome Location
Chr.9: 34398184-34458610 on Build GRCh38
Transcripts
XM_005251616.4 NM_001184940.1 NM_001184941.1 NM_001184942.1 NM_001184943.1 NM_001184945.1 NM_147202.1 XM_005251402.4 XM_011517797.2 XM_011517798.2 XM_011517799.2

SNP Detail

SNP ID Chromosome Location
rs181763314 Chr.9: 34398590-34398590 on Build GRCh38
rs988533602 Chr.9: 34398856-34398856 on Build GRCh38
rs964077834 Chr.9: 34398561-34398561 on Build GRCh38
rs899247158 Chr.9: 34398770-34398770 on Build GRCh38
rs562480149 Chr.9: 34398540-34398540 on Build GRCh38
rs1034523718 Chr.9: 34398588-34398588 on Build GRCh38
rs975433995 Chr.9: 34398803-34398803 on Build GRCh38
rs979660736 Chr.9: 34398941-34398941 on Build GRCh38
rs564362768 Chr.9: 34398639-34398639 on Build GRCh38
rs895446284 Chr.9: 34398689-34398689 on Build GRCh38
rs887009521 Chr.9: 34398736-34398736 on Build GRCh38
rs763771022 Chr.9: 34398767-34398767 on Build GRCh38
rs977841168 Chr.9: 34398658-34398658 on Build GRCh38
rs937068841 Chr.9: 34398676-34398676 on Build GRCh38
rs946958827 Chr.9: 34398921-34398921 on Build GRCh38
rs1036424660 Chr.9: 34398747-34398748 on Build GRCh38
rs930626142 Chr.9: 34398838-34398839 on Build GRCh38
rs922597126 Chr.9: 34398579-34398579 on Build GRCh38
rs201705766 Chr.9: 34398511-34398511 on Build GRCh38
rs923618331 Chr.9: 34398581-34398581 on Build GRCh38
rs931886098 Chr.9: 34398710-34398710 on Build GRCh38
rs527988099 Chr.9: 34398587-34398587 on Build GRCh38
rs917631775 Chr.9: 34398533-34398533 on Build GRCh38
rs748952136 Chr.9: 34398771-34398771 on Build GRCh38
rs1055714144 Chr.9: 34398876-34398876 on Build GRCh38
rs113390585 Chr.9: 34398635-34398635 on Build GRCh38
rs1017045793 Chr.9: 34398543-34398543 on Build GRCh38
rs1055477814 Chr.9: 34398682-34398682 on Build GRCh38
rs1014603222 Chr.9: 34398706-34398706 on Build GRCh38
rs41314238 Chr.9: 34398573-34398573 on Build GRCh38
rs921851143 Chr.9: 34398585-34398585 on Build GRCh38
rs548047972 Chr.9: 34398797-34398797 on Build GRCh38
rs964129618 Chr.9: 34398787-34398787 on Build GRCh38
rs934251091 Chr.9: 34398584-34398584 on Build GRCh38
rs1029637855 Chr.9: 34398817-34398817 on Build GRCh38
rs1005406548 Chr.9: 34398760-34398760 on Build GRCh38
rs911713834 Chr.9: 34398724-34398724 on Build GRCh38
rs957689695 Chr.9: 34398684-34398684 on Build GRCh38
rs550333203 Chr.9: 34398785-34398785 on Build GRCh38
rs914182648 Chr.9: 34398868-34398868 on Build GRCh38
rs940530826 Chr.9: 34398731-34398731 on Build GRCh38
rs561299446 Chr.9: 34398586-34398586 on Build GRCh38
rs967979856 Chr.9: 34398619-34398619 on Build GRCh38
rs955347836 Chr.9: 34398819-34398819 on Build GRCh38
rs533388324 Chr.9: 34398717-34398717 on Build GRCh38
rs541509873 Chr.9: 34398585-34398585 on Build GRCh38
rs770663884 Chr.9: 34398584-34398585 on Build GRCh38
rs904314569 Chr.9: 34398592-34398592 on Build GRCh38

View Full Product Details